Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 133199501-133200008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSTL4
Entrez Gene ID
23105
Gene Name
follistatin like 4
Gene Aliases
-
UniGene
Hs.483390
Gene Chromosome Location
Chr.5: 133194827-133842066 on Build GRCh38
Transcripts
NM_015082.1 XM_011543283.1 XM_011543284.1 XM_011543286.2 XM_011543287.2 XM_017009251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5555935 c.2092G>A p.G698R Chr.5: 133199532-133199532 on Build GRCh38
4186757 c.1982G>T p.R661L Chr.5: 133199642-133199642 on Build GRCh38
6297497 c.2095C>A p.R699S Chr.5: 133199529-133199529 on Build GRCh38
6322730 c.2049C>A p.G683G Chr.5: 133199575-133199575 on Build GRCh38
1433115 c.1858G>A p.A620T Chr.5: 133199766-133199766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761592405 Chr.5: 133199804-133199804 on Build GRCh38
rs996470244 Chr.5: 133199880-133199880 on Build GRCh38
rs775440704 Chr.5: 133199641-133199641 on Build GRCh38
rs983204748 Chr.5: 133199945-133199945 on Build GRCh38
rs773844080 Chr.5: 133199791-133199791 on Build GRCh38
rs776682386 Chr.5: 133199628-133199628 on Build GRCh38
rs375158054 Chr.5: 133199974-133199974 on Build GRCh38
rs3749816 Chr.5: 133199967-133199967 on Build GRCh38
rs142569430 Chr.5: 133199870-133199870 on Build GRCh38
rs1053636392 Chr.5: 133199802-133199802 on Build GRCh38
rs748386748 Chr.5: 133199576-133199576 on Build GRCh38
rs149501035 Chr.5: 133199528-133199528 on Build GRCh38
rs775387357 Chr.5: 133199758-133199758 on Build GRCh38
rs574540197 Chr.5: 133199632-133199632 on Build GRCh38
rs61740558 Chr.5: 133199533-133199533 on Build GRCh38
rs376343094 Chr.5: 133199568-133199568 on Build GRCh38
rs367547751 Chr.5: 133199635-133199635 on Build GRCh38
rs867775539 Chr.5: 133199643-133199643 on Build GRCh38
rs748463387 Chr.5: 133199688-133199688 on Build GRCh38
rs140560463 Chr.5: 133199642-133199642 on Build GRCh38
rs749842205 Chr.5: 133199562-133199562 on Build GRCh38
rs112340619 Chr.5: 133199684-133199684 on Build GRCh38
rs376592323 Chr.5: 133199535-133199535 on Build GRCh38
rs185850111 Chr.5: 133199664-133199664 on Build GRCh38
rs745876363 Chr.5: 133199570-133199570 on Build GRCh38
rs989077270 Chr.5: 133199840-133199840 on Build GRCh38
rs1018963439 Chr.5: 133199835-133199835 on Build GRCh38
rs947974542 Chr.5: 133199713-133199713 on Build GRCh38
rs1009130013 Chr.5: 133199551-133199551 on Build GRCh38
rs896145658 Chr.5: 133199728-133199728 on Build GRCh38
rs916470632 Chr.5: 133199698-133199698 on Build GRCh38
rs745435685 Chr.5: 133199787-133199787 on Build GRCh38
rs139046172 Chr.5: 133199731-133199731 on Build GRCh38
rs771134047 Chr.5: 133199718-133199718 on Build GRCh38
rs147873655 Chr.5: 133199741-133199741 on Build GRCh38
rs770848990 Chr.5: 133199616-133199616 on Build GRCh38
rs763998335 Chr.5: 133199784-133199784 on Build GRCh38
rs763687780 Chr.5: 133199649-133199649 on Build GRCh38
rs777877397 Chr.5: 133199697-133199697 on Build GRCh38
rs747301547 Chr.5: 133199702-133199702 on Build GRCh38
rs375012898 Chr.5: 133199610-133199610 on Build GRCh38
rs190274064 Chr.5: 133199670-133199670 on Build GRCh38
rs1019589650 Chr.5: 133199567-133199567 on Build GRCh38
rs550808563 Chr.5: 133199823-133199823 on Build GRCh38
rs560153644 Chr.5: 133199665-133199665 on Build GRCh38
rs544121824 Chr.5: 133199536-133199536 on Build GRCh38
rs370607272 Chr.5: 133199575-133199575 on Build GRCh38
rs754339021 Chr.5: 133199668-133199668 on Build GRCh38
rs561147619 Chr.5: 133199838-133199838 on Build GRCh38
rs748954143 Chr.5: 133199604-133199604 on Build GRCh38
rs1015067123 Chr.5: 133199958-133199958 on Build GRCh38
rs144664634 Chr.5: 133199690-133199690 on Build GRCh38
rs944828623 Chr.5: 133199596-133199596 on Build GRCh38
rs967745554 Chr.5: 133199752-133199752 on Build GRCh38
rs974702592 Chr.5: 133199933-133199933 on Build GRCh38
rs745746121 Chr.5: 133199740-133199740 on Build GRCh38
rs199510505 Chr.5: 133199615-133199615 on Build GRCh38
rs374862530 Chr.5: 133199529-133199529 on Build GRCh38
rs148542400 Chr.5: 133199780-133199780 on Build GRCh38
rs762679724 Chr.5: 133199772-133199772 on Build GRCh38
rs779188824 Chr.5: 133199674-133199674 on Build GRCh38
rs866493334 Chr.5: 133199879-133199879 on Build GRCh38
rs952342063 Chr.5: 133199904-133199904 on Build GRCh38
rs749123109 Chr.5: 133199896-133199896 on Build GRCh38

View Full Product Details