Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141526107-141526508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIAPH1
Entrez Gene ID
1729
Gene Name
diaphanous related formin 1
Gene Aliases
DFNA1, DIA1, DRF1, LFHL1, SCBMS, hDIA1
UniGene
Hs.529451
Gene Chromosome Location
Chr.5: 141515021-141619055 on Build GRCh38
Transcripts
NM_001079812.2 NM_001314007.1 NM_005219.4 XM_011537572.2 XM_011537573.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7158606 c.3293A>C p.Q1098P Chr.5: 141526442-141526442 on Build GRCh38
5649913 c.3318G>A p.M1106I Chr.5: 141526417-141526417 on Build GRCh38
1063474 c.3352C>T p.L1118L Chr.5: 141526383-141526383 on Build GRCh38
6447145 c.3274-1C>T p.? Chr.5: 141526462-141526462 on Build GRCh38
7221758 c.3430A>T p.M1144L Chr.5: 141526305-141526305 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764475981 Chr.5: 141526473-141526473 on Build GRCh38
rs758163133 Chr.5: 141526320-141526320 on Build GRCh38
rs762290499 Chr.5: 141526479-141526479 on Build GRCh38
rs376863281 Chr.5: 141526283-141526283 on Build GRCh38
rs761026699 Chr.5: 141526264-141526264 on Build GRCh38
rs369559394 Chr.5: 141526459-141526459 on Build GRCh38
rs754521222 Chr.5: 141526339-141526339 on Build GRCh38
rs756003432 Chr.5: 141526377-141526377 on Build GRCh38
rs115666914 Chr.5: 141526200-141526200 on Build GRCh38
rs183167083 Chr.5: 141526350-141526350 on Build GRCh38
rs727504670 Chr.5: 141526375-141526375 on Build GRCh38
rs762394819 Chr.5: 141526347-141526349 on Build GRCh38
rs766106223 Chr.5: 141526321-141526321 on Build GRCh38
rs774467412 Chr.5: 141526212-141526212 on Build GRCh38
rs756529055 Chr.5: 141526142-141526142 on Build GRCh38
rs949439587 Chr.5: 141526205-141526205 on Build GRCh38
rs376435335 Chr.5: 141526183-141526183 on Build GRCh38
rs200419320 Chr.5: 141526310-141526310 on Build GRCh38
rs371161306 Chr.5: 141526404-141526404 on Build GRCh38
rs997606547 Chr.5: 141526442-141526442 on Build GRCh38
rs548105319 Chr.5: 141526425-141526425 on Build GRCh38
rs977061013 Chr.5: 141526357-141526357 on Build GRCh38
rs751207442 Chr.5: 141526327-141526327 on Build GRCh38
rs999653581 Chr.5: 141526260-141526260 on Build GRCh38
rs748520819 Chr.5: 141526466-141526466 on Build GRCh38
rs199609749 Chr.5: 141526215-141526215 on Build GRCh38
rs369467830 Chr.5: 141526149-141526149 on Build GRCh38
rs764296073 Chr.5: 141526145-141526145 on Build GRCh38
rs771113954 Chr.5: 141526210-141526210 on Build GRCh38
rs576123582 Chr.5: 141526397-141526397 on Build GRCh38
rs779775889 Chr.5: 141526148-141526148 on Build GRCh38
rs768948411 Chr.5: 141526178-141526178 on Build GRCh38
rs761310118 Chr.5: 141526462-141526462 on Build GRCh38
rs772204178 Chr.5: 141526433-141526433 on Build GRCh38
rs750111294 Chr.5: 141526146-141526146 on Build GRCh38
rs749063652 Chr.5: 141526413-141526413 on Build GRCh38
rs928380301 Chr.5: 141526360-141526360 on Build GRCh38
rs776065119 Chr.5: 141526263-141526263 on Build GRCh38
rs747970514 Chr.5: 141526173-141526173 on Build GRCh38
rs373930400 Chr.5: 141526162-141526162 on Build GRCh38
rs373713667 Chr.5: 141526209-141526209 on Build GRCh38
rs767451157 Chr.5: 141526224-141526224 on Build GRCh38
rs768077893 Chr.5: 141526477-141526477 on Build GRCh38
rs779733072 Chr.5: 141526474-141526474 on Build GRCh38
rs902773161 Chr.5: 141526221-141526221 on Build GRCh38
rs964330253 Chr.5: 141526188-141526188 on Build GRCh38
rs764575477 Chr.5: 141526274-141526274 on Build GRCh38
rs377665243 Chr.5: 141526147-141526147 on Build GRCh38
rs777183651 Chr.5: 141526475-141526475 on Build GRCh38
rs893998939 Chr.5: 141526403-141526403 on Build GRCh38
rs781577050 Chr.5: 141526422-141526422 on Build GRCh38
rs750971019 Chr.5: 141526478-141526479 on Build GRCh38
rs770668053 Chr.5: 141526421-141526421 on Build GRCh38
rs754920334 Chr.5: 141526150-141526150 on Build GRCh38
rs916076581 Chr.5: 141526476-141526476 on Build GRCh38
rs777586861 Chr.5: 141526383-141526383 on Build GRCh38
rs761110996 Chr.5: 141526480-141526481 on Build GRCh38

View Full Product Details