Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151049636-151050146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNIP1
Entrez Gene ID
10318
Gene Name
TNFAIP3 interacting protein 1
Gene Aliases
ABIN-1, NAF1, VAN, nip40-1
UniGene
Hs.355141
Gene Chromosome Location
Chr.5: 151029943-151087660 on Build GRCh38
Transcripts
NM_001252385.1 NM_001252386.1 NM_001252390.1 NM_001252391.1 NM_001252392.1 NM_001252393.1 NM_001258454.1 NM_001258455.1 NM_001258456.1 NM_006058.4 XM_005268355.1 XM_006714751.1 XM_006714752.2 XM_017008945.1 XM_017008946.1 XM_017008947.1 XM_017008948.1 XM_017008949.1 XM_017008950.1 XM_017008951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5026442 c.801G>A p.P267P Chr.5: 151049869-151049869 on Build GRCh38
6562975 c.804G>T p.K268N Chr.5: 151049866-151049866 on Build GRCh38
4988937 c.800C>T p.P267L Chr.5: 151049870-151049870 on Build GRCh38
1723318 c.771_772insA p.E258fs*27 Chr.5: 151049898-151049899 on Build GRCh38
6562976 c.804G>T p.K268N Chr.5: 151049866-151049866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569039631 Chr.5: 151049707-151049707 on Build GRCh38
rs747647386 Chr.5: 151049966-151049966 on Build GRCh38
rs780838663 Chr.5: 151049951-151049951 on Build GRCh38
rs528664852 Chr.5: 151050119-151050119 on Build GRCh38
rs747665714 Chr.5: 151049843-151049843 on Build GRCh38
rs2233297 Chr.5: 151049679-151049679 on Build GRCh38
rs1037526665 Chr.5: 151050084-151050084 on Build GRCh38
rs770230799 Chr.5: 151049869-151049869 on Build GRCh38
rs1044185216 Chr.5: 151049950-151049950 on Build GRCh38
rs375892201 Chr.5: 151049937-151049937 on Build GRCh38
rs778913499 Chr.5: 151049790-151049790 on Build GRCh38
rs867930084 Chr.5: 151049693-151049693 on Build GRCh38
rs767169472 Chr.5: 151050052-151050052 on Build GRCh38
rs978280847 Chr.5: 151049774-151049774 on Build GRCh38
rs2233294 Chr.5: 151050002-151050002 on Build GRCh38
rs139634771 Chr.5: 151049882-151049882 on Build GRCh38
rs749846383 Chr.5: 151049983-151049983 on Build GRCh38
rs567295943 Chr.5: 151049742-151049742 on Build GRCh38
rs200832817 Chr.5: 151049896-151049896 on Build GRCh38
rs1016550028 Chr.5: 151049663-151049663 on Build GRCh38
rs964816382 Chr.5: 151049668-151049668 on Build GRCh38
rs945789901 Chr.5: 151049900-151049900 on Build GRCh38
rs765071000 Chr.5: 151049894-151049894 on Build GRCh38
rs746713152 Chr.5: 151049819-151049819 on Build GRCh38
rs745590011 Chr.5: 151049797-151049797 on Build GRCh38
rs1047639929 Chr.5: 151050030-151050030 on Build GRCh38
rs778107449 Chr.5: 151049982-151049982 on Build GRCh38
rs759534636 Chr.5: 151050036-151050036 on Build GRCh38
rs779851891 Chr.5: 151049909-151049909 on Build GRCh38
rs529909021 Chr.5: 151049708-151049708 on Build GRCh38
rs767881838 Chr.5: 151049890-151049890 on Build GRCh38
rs367620766 Chr.5: 151049789-151049789 on Build GRCh38
rs938333181 Chr.5: 151050020-151050020 on Build GRCh38
rs371782529 Chr.5: 151049873-151049873 on Build GRCh38
rs780227485 Chr.5: 151049690-151049690 on Build GRCh38
rs117663772 Chr.5: 151049883-151049883 on Build GRCh38
rs189304727 Chr.5: 151050121-151050121 on Build GRCh38
rs768320624 Chr.5: 151049832-151049832 on Build GRCh38
rs751387931 Chr.5: 151049914-151049914 on Build GRCh38
rs766565443 Chr.5: 151049965-151049966 on Build GRCh38
rs758270360 Chr.5: 151049800-151049800 on Build GRCh38
rs779543371 Chr.5: 151049801-151049801 on Build GRCh38
rs774812630 Chr.5: 151049995-151049995 on Build GRCh38
rs769415553 Chr.5: 151049980-151049980 on Build GRCh38
rs776082808 Chr.5: 151049837-151049837 on Build GRCh38
rs758106396 Chr.5: 151049905-151049905 on Build GRCh38
rs563424120 Chr.5: 151050102-151050102 on Build GRCh38
rs1030266967 Chr.5: 151049689-151049689 on Build GRCh38
rs2233295 Chr.5: 151049891-151049891 on Build GRCh38
rs2233296 Chr.5: 151049835-151049835 on Build GRCh38
rs575804388 Chr.5: 151050005-151050005 on Build GRCh38
rs768848133 Chr.5: 151049794-151049794 on Build GRCh38
rs754562369 Chr.5: 151049918-151049918 on Build GRCh38
rs994918181 Chr.5: 151050081-151050081 on Build GRCh38
rs550490018 Chr.5: 151049694-151049694 on Build GRCh38
rs889103642 Chr.5: 151050101-151050101 on Build GRCh38
rs766804511 Chr.5: 151049880-151049880 on Build GRCh38
rs548481454 Chr.5: 151049741-151049741 on Build GRCh38
rs542916738 Chr.5: 151050046-151050046 on Build GRCh38
rs377727559 Chr.5: 151049870-151049870 on Build GRCh38
rs771361669 Chr.5: 151049990-151049990 on Build GRCh38
rs754033722 Chr.5: 151049992-151050000 on Build GRCh38
rs925607693 Chr.5: 151049964-151049964 on Build GRCh38
rs533355788 Chr.5: 151049734-151049734 on Build GRCh38
rs57701664 Chr.5: 151049684-151049684 on Build GRCh38
rs200474545 Chr.5: 151049836-151049836 on Build GRCh38

View Full Product Details