Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180590672-180591168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCGB3A1
Entrez Gene ID
92304
Gene Name
secretoglobin family 3A member 1
Gene Aliases
HIN-1, HIN1, LU105, PnSP-2, UGRP2
UniGene
Hs.62492
Gene Chromosome Location
Chr.5: 180590105-180591487 on Build GRCh38
Transcripts
NM_052863.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4595507 c.151C>T p.L51F Chr.5: 180590740-180590740 on Build GRCh38
4595893 c.128G>A p.G43E Chr.5: 180590763-180590763 on Build GRCh38
1724271 c.63C>T p.F21F Chr.5: 180590828-180590828 on Build GRCh38
3012341 c.167C>T p.P56L Chr.5: 180590724-180590724 on Build GRCh38
737474 c.85G>T p.V29L Chr.5: 180590806-180590806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771508324 Chr.5: 180590935-180590935 on Build GRCh38
rs748746447 Chr.5: 180590909-180590909 on Build GRCh38
rs753351103 Chr.5: 180590815-180590815 on Build GRCh38
rs936573372 Chr.5: 180590912-180590912 on Build GRCh38
rs996419318 Chr.5: 180590998-180590998 on Build GRCh38
rs764951005 Chr.5: 180590817-180590817 on Build GRCh38
rs1020379034 Chr.5: 180590993-180590993 on Build GRCh38
rs990694150 Chr.5: 180591128-180591128 on Build GRCh38
rs886153015 Chr.5: 180590949-180590949 on Build GRCh38
rs763602544 Chr.5: 180590729-180590729 on Build GRCh38
rs549066053 Chr.5: 180591027-180591027 on Build GRCh38
rs749540547 Chr.5: 180590774-180590774 on Build GRCh38
rs1046573388 Chr.5: 180590945-180590945 on Build GRCh38
rs117170752 Chr.5: 180590927-180590927 on Build GRCh38
rs553072572 Chr.5: 180591073-180591073 on Build GRCh38
rs936402929 Chr.5: 180591020-180591020 on Build GRCh38
rs756600640 Chr.5: 180590869-180590869 on Build GRCh38
rs371005942 Chr.5: 180590932-180590932 on Build GRCh38
rs753145638 Chr.5: 180590864-180590864 on Build GRCh38
rs759325239 Chr.5: 180590786-180590786 on Build GRCh38
rs925039151 Chr.5: 180591013-180591013 on Build GRCh38
rs759521302 Chr.5: 180590714-180590714 on Build GRCh38
rs771768539 Chr.5: 180590879-180590879 on Build GRCh38
rs576021464 Chr.5: 180590740-180590740 on Build GRCh38
rs1029201434 Chr.5: 180591017-180591017 on Build GRCh38
rs563272924 Chr.5: 180590794-180590794 on Build GRCh38
rs781545845 Chr.5: 180590751-180590751 on Build GRCh38
rs746036190 Chr.5: 180590877-180590877 on Build GRCh38
rs954856870 Chr.5: 180591021-180591021 on Build GRCh38
rs760594889 Chr.5: 180590803-180590803 on Build GRCh38
rs774175548 Chr.5: 180590777-180590777 on Build GRCh38
rs925220733 Chr.5: 180590906-180590906 on Build GRCh38
rs985524114 Chr.5: 180590787-180590787 on Build GRCh38
rs767328587 Chr.5: 180590715-180590715 on Build GRCh38
rs967273494 Chr.5: 180590765-180590765 on Build GRCh38
rs755570373 Chr.5: 180590721-180590721 on Build GRCh38
rs373108626 Chr.5: 180590799-180590799 on Build GRCh38
rs536283333 Chr.5: 180591029-180591029 on Build GRCh38
rs967785683 Chr.5: 180590858-180590858 on Build GRCh38
rs867364290 Chr.5: 180590731-180590731 on Build GRCh38
rs773272977 Chr.5: 180590772-180590772 on Build GRCh38
rs530717590 Chr.5: 180590994-180590994 on Build GRCh38
rs892853209 Chr.5: 180591125-180591125 on Build GRCh38
rs777850373 Chr.5: 180590874-180590874 on Build GRCh38
rs909862808 Chr.5: 180590797-180590797 on Build GRCh38
rs151275301 Chr.5: 180590716-180590716 on Build GRCh38
rs998413178 Chr.5: 180590999-180590999 on Build GRCh38
rs954806841 Chr.5: 180590782-180590782 on Build GRCh38
rs550508085 Chr.5: 180591024-180591024 on Build GRCh38
rs757017012 Chr.5: 180590816-180590816 on Build GRCh38
rs1001302085 Chr.5: 180590733-180590733 on Build GRCh38
rs957980491 Chr.5: 180591114-180591114 on Build GRCh38
rs747353871 Chr.5: 180590744-180590744 on Build GRCh38
rs972718118 Chr.5: 180591068-180591068 on Build GRCh38
rs927093393 Chr.5: 180591086-180591086 on Build GRCh38
rs748494183 Chr.5: 180590753-180590755 on Build GRCh38
rs377764708 Chr.5: 180590809-180590809 on Build GRCh38
rs1033061269 Chr.5: 180590737-180590737 on Build GRCh38
rs949918401 Chr.5: 180591078-180591078 on Build GRCh38
rs369844606 Chr.5: 180590841-180590841 on Build GRCh38
rs755581339 Chr.5: 180590748-180590748 on Build GRCh38
rs753547037 Chr.5: 180590738-180590738 on Build GRCh38
rs748317251 Chr.5: 180590759-180590759 on Build GRCh38
rs1009939563 Chr.5: 180590957-180590957 on Build GRCh38
rs963124607 Chr.5: 180591051-180591051 on Build GRCh38
rs981384271 Chr.5: 180591080-180591080 on Build GRCh38
rs949175768 Chr.5: 180590944-180590944 on Build GRCh38
rs566975609 Chr.5: 180591144-180591144 on Build GRCh38
rs369637956 Chr.5: 180590790-180590790 on Build GRCh38
rs571314529 Chr.5: 180590941-180590941 on Build GRCh38
rs528151303 Chr.5: 180590924-180590924 on Build GRCh38
rs749338464 Chr.5: 180590875-180590875 on Build GRCh38
rs528941918 Chr.5: 180590997-180590997 on Build GRCh38
rs1021246451 Chr.5: 180590757-180590757 on Build GRCh38
rs1045565317 Chr.5: 180590899-180590899 on Build GRCh38
rs1004966004 Chr.5: 180590967-180590967 on Build GRCh38
rs781557334 Chr.5: 180590857-180590857 on Build GRCh38
rs978752842 Chr.5: 180590871-180590871 on Build GRCh38
rs905707510 Chr.5: 180590911-180590911 on Build GRCh38
rs374562892 Chr.5: 180590741-180590741 on Build GRCh38
rs925943189 Chr.5: 180591103-180591103 on Build GRCh38
rs780589927 Chr.5: 180590743-180590743 on Build GRCh38
rs559432495 Chr.5: 180590876-180590876 on Build GRCh38
rs769772742 Chr.5: 180590762-180590762 on Build GRCh38
rs755283773 Chr.5: 180590852-180590852 on Build GRCh38
rs937196694 Chr.5: 180591102-180591102 on Build GRCh38

View Full Product Details