Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 3153577-3154072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBB2A
Entrez Gene ID
7280
Gene Name
tubulin beta 2A class IIa
Gene Aliases
CDCBM5, TUBB, TUBB2
UniGene
Hs.654543
Gene Chromosome Location
Chr.6: 3153666-3157549 on Build GRCh38
Transcripts
NM_001069.2 NM_001310315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3624577 c.1300G>A p.G434R Chr.6: 3153901-3153901 on Build GRCh38
1077590 c.1186C>T p.H396Y Chr.6: 3154015-3154015 on Build GRCh38
4153244 c.1287G>A p.T429T Chr.6: 3153914-3153914 on Build GRCh38
741379 c.1320G>C p.E440D Chr.6: 3153881-3153881 on Build GRCh38
1077588 c.1260C>A p.S420S Chr.6: 3153941-3153941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540952596 Chr.6: 3153736-3153737 on Build GRCh38
rs987749526 Chr.6: 3153632-3153632 on Build GRCh38
rs62637713 Chr.6: 3153878-3153878 on Build GRCh38
rs956741889 Chr.6: 3153843-3153843 on Build GRCh38
rs1003032383 Chr.6: 3153807-3153807 on Build GRCh38
rs760987108 Chr.6: 3153845-3153848 on Build GRCh38
rs1038351981 Chr.6: 3153757-3153757 on Build GRCh38
rs759149588 Chr.6: 3153857-3153857 on Build GRCh38
rs1054419 Chr.6: 3153904-3153904 on Build GRCh38
rs766895377 Chr.6: 3153855-3153857 on Build GRCh38
rs749283603 Chr.6: 3153740-3153740 on Build GRCh38
rs1049199649 Chr.6: 3153611-3153611 on Build GRCh38
rs3205007 Chr.6: 3153756-3153756 on Build GRCh38
rs762463562 Chr.6: 3153845-3153845 on Build GRCh38
rs369148878 Chr.6: 3153892-3153892 on Build GRCh38
rs772818777 Chr.6: 3153701-3153701 on Build GRCh38
rs1028233753 Chr.6: 3153974-3153974 on Build GRCh38
rs774107403 Chr.6: 3153941-3153941 on Build GRCh38
rs745518154 Chr.6: 3153989-3153989 on Build GRCh38
rs898507927 Chr.6: 3153762-3153762 on Build GRCh38
rs765549114 Chr.6: 3153870-3153870 on Build GRCh38
rs770480973 Chr.6: 3153913-3153913 on Build GRCh38
rs533112511 Chr.6: 3153666-3153666 on Build GRCh38
rs548563768 Chr.6: 3153651-3153651 on Build GRCh38
rs768447032 Chr.6: 3153823-3153823 on Build GRCh38
rs1005075801 Chr.6: 3153629-3153629 on Build GRCh38
rs777633833 Chr.6: 3153718-3153718 on Build GRCh38
rs773170649 Chr.6: 3153842-3153842 on Build GRCh38
rs781346126 Chr.6: 3153824-3153824 on Build GRCh38
rs755145441 Chr.6: 3153890-3153890 on Build GRCh38
rs771502017 Chr.6: 3154004-3154004 on Build GRCh38
rs747853799 Chr.6: 3153665-3153665 on Build GRCh38
rs914926045 Chr.6: 3153639-3153639 on Build GRCh38
rs72843326 Chr.6: 3153825-3153825 on Build GRCh38
rs200677577 Chr.6: 3154043-3154043 on Build GRCh38
rs756141916 Chr.6: 3153908-3153908 on Build GRCh38
rs762262687 Chr.6: 3154040-3154040 on Build GRCh38
rs562932953 Chr.6: 3153808-3153808 on Build GRCh38
rs866944719 Chr.6: 3153709-3153709 on Build GRCh38
rs1057518044 Chr.6: 3153961-3153961 on Build GRCh38
rs145215507 Chr.6: 3153773-3153773 on Build GRCh38
rs1044651891 Chr.6: 3153798-3153798 on Build GRCh38
rs267600945 Chr.6: 3153911-3153911 on Build GRCh38
rs531681539 Chr.6: 3153642-3153642 on Build GRCh38
rs754289959 Chr.6: 3153880-3153882 on Build GRCh38
rs202207883 Chr.6: 3153871-3153871 on Build GRCh38
rs887928613 Chr.6: 3153613-3153613 on Build GRCh38
rs933837370 Chr.6: 3153712-3153713 on Build GRCh38
rs773699055 Chr.6: 3153851-3153851 on Build GRCh38
rs572259304 Chr.6: 3153604-3153609 on Build GRCh38
rs375083268 Chr.6: 3153814-3153814 on Build GRCh38
rs975362341 Chr.6: 3153689-3153689 on Build GRCh38
rs770665341 Chr.6: 3153847-3153847 on Build GRCh38
rs3205008 Chr.6: 3153754-3153754 on Build GRCh38
rs777062028 Chr.6: 3153861-3153861 on Build GRCh38
rs1036570957 Chr.6: 3153634-3153634 on Build GRCh38
rs921055081 Chr.6: 3153708-3153708 on Build GRCh38
rs758531793 Chr.6: 3153872-3153872 on Build GRCh38
rs969477381 Chr.6: 3153802-3153802 on Build GRCh38
rs764492313 Chr.6: 3153858-3153858 on Build GRCh38
rs762314156 Chr.6: 3153869-3153869 on Build GRCh38
rs769656196 Chr.6: 3153828-3153828 on Build GRCh38
rs956486463 Chr.6: 3153609-3153609 on Build GRCh38
rs574707391 Chr.6: 3153884-3153884 on Build GRCh38

View Full Product Details