Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 11713869-11714380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADTRP
Entrez Gene ID
84830
Gene Name
androgen dependent TFPI regulating protein
Gene Aliases
AIG1L, C6orf105, dJ413H6.1
UniGene
Hs.126409
Gene Chromosome Location
Chr.6: 11713523-11779628 on Build GRCh38
Transcripts
NM_001143948.1 NM_032744.3 XM_005249454.3 XM_011514956.1 XM_017011377.1

SNP Detail

SNP ID Chromosome Location
rs869170692 Chr.6: 11713968-11713969 on Build GRCh38
rs530013105 Chr.6: 11714241-11714244 on Build GRCh38
rs3069335 Chr.6: 11713981-11713981 on Build GRCh38
rs952800803 Chr.6: 11714147-11714147 on Build GRCh38
rs904297330 Chr.6: 11713948-11713948 on Build GRCh38
rs169790 Chr.6: 11714058-11714058 on Build GRCh38
rs139878588 Chr.6: 11713896-11713896 on Build GRCh38
rs397703615 Chr.6: 11713967-11713968 on Build GRCh38
rs1009271640 Chr.6: 11714000-11714000 on Build GRCh38
rs1042757121 Chr.6: 11713923-11713923 on Build GRCh38
rs1009910738 Chr.6: 11713921-11713921 on Build GRCh38
rs868282309 Chr.6: 11714225-11714225 on Build GRCh38
rs757072071 Chr.6: 11713908-11713908 on Build GRCh38
rs750246592 Chr.6: 11714079-11714079 on Build GRCh38
rs914876341 Chr.6: 11714109-11714109 on Build GRCh38
rs56750142 Chr.6: 11713992-11713993 on Build GRCh38
rs1026586252 Chr.6: 11713981-11713981 on Build GRCh38
rs995185303 Chr.6: 11714309-11714309 on Build GRCh38
rs758283326 Chr.6: 11714139-11714139 on Build GRCh38
rs1018643988 Chr.6: 11714054-11714054 on Build GRCh38
rs34508693 Chr.6: 11713904-11713904 on Build GRCh38
rs1057495686 Chr.6: 11714197-11714197 on Build GRCh38
rs112324696 Chr.6: 11714140-11714140 on Build GRCh38
rs1018696860 Chr.6: 11714356-11714357 on Build GRCh38
rs913495103 Chr.6: 11714228-11714228 on Build GRCh38
rs945690358 Chr.6: 11714271-11714271 on Build GRCh38
rs972742013 Chr.6: 11714083-11714083 on Build GRCh38
rs948798934 Chr.6: 11714235-11714235 on Build GRCh38
rs758473602 Chr.6: 11713982-11713983 on Build GRCh38
rs780883932 Chr.6: 11714337-11714337 on Build GRCh38
rs886306230 Chr.6: 11714331-11714331 on Build GRCh38
rs950537259 Chr.6: 11713983-11713983 on Build GRCh38
rs934801926 Chr.6: 11714148-11714148 on Build GRCh38
rs869222090 Chr.6: 11713991-11713992 on Build GRCh38
rs369956854 Chr.6: 11714283-11714286 on Build GRCh38
rs201873314 Chr.6: 11714280-11714283 on Build GRCh38
rs112834653 Chr.6: 11713950-11713950 on Build GRCh38
rs1026174728 Chr.6: 11714324-11714324 on Build GRCh38
rs895901425 Chr.6: 11714185-11714185 on Build GRCh38
rs746494271 Chr.6: 11714188-11714188 on Build GRCh38
rs980257914 Chr.6: 11714161-11714161 on Build GRCh38
rs534527152 Chr.6: 11714276-11714276 on Build GRCh38
rs1042725940 Chr.6: 11714275-11714275 on Build GRCh38
rs200555116 Chr.6: 11714281-11714284 on Build GRCh38
rs869235863 Chr.6: 11713993-11713993 on Build GRCh38
rs536908895 Chr.6: 11713982-11713982 on Build GRCh38
rs904269669 Chr.6: 11714329-11714329 on Build GRCh38
rs1028814192 Chr.6: 11713980-11713980 on Build GRCh38
rs1057271110 Chr.6: 11714174-11714174 on Build GRCh38
rs752516523 Chr.6: 11713981-11713983 on Build GRCh38
rs954643305 Chr.6: 11713989-11713989 on Build GRCh38
rs979011635 Chr.6: 11714146-11714147 on Build GRCh38
rs869067941 Chr.6: 11713991-11713993 on Build GRCh38
rs780002086 Chr.6: 11714175-11714175 on Build GRCh38
rs924795791 Chr.6: 11714147-11714148 on Build GRCh38
rs532543278 Chr.6: 11713981-11713982 on Build GRCh38
rs5874334 Chr.6: 11713964-11713965 on Build GRCh38

View Full Product Details