Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26598921-26599430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABT1
Entrez Gene ID
29777
Gene Name
activator of basal transcription 1
Gene Aliases
hABT1
UniGene
Hs.254406
Gene Chromosome Location
Chr.6: 26596943-26600050 on Build GRCh38
Transcripts
NM_013375.3

SNP Detail

SNP ID Chromosome Location
rs527681003 Chr.6: 26598991-26598991 on Build GRCh38
rs895914694 Chr.6: 26599097-26599097 on Build GRCh38
rs929271395 Chr.6: 26599060-26599060 on Build GRCh38
rs546449352 Chr.6: 26598999-26598999 on Build GRCh38
rs954465850 Chr.6: 26599338-26599338 on Build GRCh38
rs553844288 Chr.6: 26599348-26599348 on Build GRCh38
rs551893680 Chr.6: 26599321-26599321 on Build GRCh38
rs533782293 Chr.6: 26599332-26599332 on Build GRCh38
rs537913519 Chr.6: 26599215-26599215 on Build GRCh38
rs750673356 Chr.6: 26598997-26598997 on Build GRCh38
rs956599204 Chr.6: 26599264-26599264 on Build GRCh38
rs552324058 Chr.6: 26599008-26599008 on Build GRCh38
rs1045302585 Chr.6: 26599129-26599129 on Build GRCh38
rs45527431 Chr.6: 26599281-26599281 on Build GRCh38
rs180692287 Chr.6: 26599378-26599378 on Build GRCh38
rs919277896 Chr.6: 26599013-26599013 on Build GRCh38
rs781842144 Chr.6: 26599159-26599159 on Build GRCh38
rs986272729 Chr.6: 26598947-26598947 on Build GRCh38
rs369983822 Chr.6: 26599105-26599105 on Build GRCh38
rs1032243080 Chr.6: 26599223-26599223 on Build GRCh38
rs529249489 Chr.6: 26599000-26599000 on Build GRCh38
rs1001223960 Chr.6: 26599185-26599185 on Build GRCh38
rs910747101 Chr.6: 26599350-26599350 on Build GRCh38
rs905022628 Chr.6: 26599143-26599143 on Build GRCh38
rs782659382 Chr.6: 26599294-26599294 on Build GRCh38
rs73739030 Chr.6: 26599004-26599004 on Build GRCh38
rs1009202227 Chr.6: 26599279-26599279 on Build GRCh38
rs1057130031 Chr.6: 26599088-26599088 on Build GRCh38
rs554729175 Chr.6: 26599249-26599249 on Build GRCh38
rs368622746 Chr.6: 26599045-26599045 on Build GRCh38
rs1012971126 Chr.6: 26599107-26599107 on Build GRCh38
rs189939133 Chr.6: 26599002-26599002 on Build GRCh38
rs568989334 Chr.6: 26599076-26599076 on Build GRCh38

View Full Product Details