Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153930865-153931357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND4B
Entrez Gene ID
9909
Gene Name
DENN domain containing 4B
Gene Aliases
KIAA0476
UniGene
Hs.744878
Gene Chromosome Location
Chr.1: 153929501-153946681 on Build GRCh38
Transcripts
NM_014856.2 XM_005245678.4 XM_005245679.2 XM_011510219.1 XM_011510220.2 XM_011510221.2 XM_017002993.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1203364 c.4043G>A p.R1348Q Chr.1: 153931018-153931018 on Build GRCh38
4677018 c.3998C>A p.A1333D Chr.1: 153931063-153931063 on Build GRCh38
1319937 c.3661-3_3661-2insC p.? Chr.1: 153931066-153931067 on Build GRCh38
1203365 c.3707G>A p.R1236Q Chr.1: 153931018-153931018 on Build GRCh38
4677019 c.3662C>A p.A1221D Chr.1: 153931063-153931063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996433439 Chr.1: 153931128-153931128 on Build GRCh38
rs781753616 Chr.1: 153931004-153931004 on Build GRCh38
rs757602960 Chr.1: 153931072-153931072 on Build GRCh38
rs762613294 Chr.1: 153930954-153930954 on Build GRCh38
rs764657194 Chr.1: 153931087-153931087 on Build GRCh38
rs1041232689 Chr.1: 153931106-153931106 on Build GRCh38
rs750034641 Chr.1: 153930944-153930944 on Build GRCh38
rs953605710 Chr.1: 153931184-153931184 on Build GRCh38
rs749527610 Chr.1: 153930902-153930902 on Build GRCh38
rs989378695 Chr.1: 153931160-153931160 on Build GRCh38
rs1027854064 Chr.1: 153931304-153931304 on Build GRCh38
rs372457135 Chr.1: 153930940-153930940 on Build GRCh38
rs76216422 Chr.1: 153931006-153931006 on Build GRCh38
rs1021780241 Chr.1: 153931252-153931252 on Build GRCh38
rs150831120 Chr.1: 153931080-153931080 on Build GRCh38
rs900934103 Chr.1: 153931116-153931116 on Build GRCh38
rs571802931 Chr.1: 153931240-153931240 on Build GRCh38
rs994824639 Chr.1: 153931273-153931273 on Build GRCh38
rs767725042 Chr.1: 153931311-153931311 on Build GRCh38
rs748064755 Chr.1: 153931098-153931098 on Build GRCh38
rs34834138 Chr.1: 153931247-153931247 on Build GRCh38
rs1018437069 Chr.1: 153930918-153930918 on Build GRCh38
rs977528767 Chr.1: 153931272-153931272 on Build GRCh38
rs370247245 Chr.1: 153930922-153930922 on Build GRCh38
rs935003443 Chr.1: 153931000-153931000 on Build GRCh38
rs183373310 Chr.1: 153931162-153931162 on Build GRCh38
rs200600366 Chr.1: 153930964-153930964 on Build GRCh38
rs748401005 Chr.1: 153931127-153931127 on Build GRCh38
rs774595759 Chr.1: 153930913-153930913 on Build GRCh38
rs771095154 Chr.1: 153930903-153930903 on Build GRCh38
rs923572672 Chr.1: 153930996-153930996 on Build GRCh38
rs778314875 Chr.1: 153931053-153931053 on Build GRCh38
rs915804176 Chr.1: 153930915-153930915 on Build GRCh38
rs939419554 Chr.1: 153931224-153931225 on Build GRCh38
rs755439162 Chr.1: 153930984-153930984 on Build GRCh38
rs756436696 Chr.1: 153931037-153931037 on Build GRCh38
rs769527145 Chr.1: 153931102-153931102 on Build GRCh38
rs778085882 Chr.1: 153930892-153930892 on Build GRCh38
rs375631387 Chr.1: 153930985-153930985 on Build GRCh38
rs753159925 Chr.1: 153931019-153931019 on Build GRCh38
rs746017232 Chr.1: 153931082-153931082 on Build GRCh38
rs947873522 Chr.1: 153931164-153931164 on Build GRCh38
rs751124817 Chr.1: 153930965-153930965 on Build GRCh38
rs761608952 Chr.1: 153930937-153930937 on Build GRCh38
rs1044946430 Chr.1: 153931200-153931200 on Build GRCh38
rs749474369 Chr.1: 153931061-153931061 on Build GRCh38
rs760498000 Chr.1: 153930921-153930921 on Build GRCh38
rs757653243 Chr.1: 153931257-153931257 on Build GRCh38
rs776563575 Chr.1: 153931096-153931096 on Build GRCh38
rs372670116 Chr.1: 153930952-153930952 on Build GRCh38
rs551511498 Chr.1: 153931213-153931213 on Build GRCh38
rs569739218 Chr.1: 153931310-153931310 on Build GRCh38
rs1055813617 Chr.1: 153930911-153930911 on Build GRCh38
rs776276303 Chr.1: 153930935-153930935 on Build GRCh38
rs965116249 Chr.1: 153930966-153930966 on Build GRCh38

View Full Product Details