Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41934872-41935367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCND3
Entrez Gene ID
896
Gene Name
cyclin D3
Gene Aliases
-
UniGene
Hs.534307
Gene Chromosome Location
Chr.6: 41934933-42048894 on Build GRCh38
Transcripts
NM_001136017.3 NM_001136125.2 NM_001136126.2 NM_001287427.1 NM_001287434.1 NM_001760.4 XM_011514971.1 XM_011514972.1

SNP Detail

SNP ID Chromosome Location
rs879739915 Chr.6: 41935067-41935068 on Build GRCh38
rs903789585 Chr.6: 41935036-41935036 on Build GRCh38
rs527697377 Chr.6: 41935147-41935147 on Build GRCh38
rs780467730 Chr.6: 41934919-41934920 on Build GRCh38
rs1050933003 Chr.6: 41934958-41934958 on Build GRCh38
rs866375719 Chr.6: 41934932-41934932 on Build GRCh38
rs534111328 Chr.6: 41935018-41935018 on Build GRCh38
rs527958754 Chr.6: 41935291-41935291 on Build GRCh38
rs9529 Chr.6: 41935269-41935269 on Build GRCh38
rs747739901 Chr.6: 41935325-41935325 on Build GRCh38
rs1006803059 Chr.6: 41934964-41934964 on Build GRCh38
rs762409757 Chr.6: 41935232-41935232 on Build GRCh38
rs1024458526 Chr.6: 41935156-41935156 on Build GRCh38
rs999109988 Chr.6: 41935040-41935040 on Build GRCh38
rs547720067 Chr.6: 41935304-41935304 on Build GRCh38
rs776899963 Chr.6: 41935294-41935295 on Build GRCh38
rs1032762048 Chr.6: 41935248-41935248 on Build GRCh38
rs3218106 Chr.6: 41935200-41935200 on Build GRCh38
rs749130363 Chr.6: 41935043-41935043 on Build GRCh38
rs889060379 Chr.6: 41935076-41935076 on Build GRCh38
rs889617163 Chr.6: 41935286-41935286 on Build GRCh38
rs1051171 Chr.6: 41935108-41935108 on Build GRCh38
rs970670964 Chr.6: 41935169-41935169 on Build GRCh38
rs934075149 Chr.6: 41934929-41934929 on Build GRCh38
rs897218621 Chr.6: 41934927-41934927 on Build GRCh38
rs941610682 Chr.6: 41935237-41935237 on Build GRCh38
rs3218107 Chr.6: 41934962-41934962 on Build GRCh38
rs892865649 Chr.6: 41935249-41935249 on Build GRCh38
rs1023122279 Chr.6: 41935311-41935311 on Build GRCh38
rs982095390 Chr.6: 41935336-41935336 on Build GRCh38
rs1030644717 Chr.6: 41935106-41935106 on Build GRCh38
rs563241000 Chr.6: 41934998-41934998 on Build GRCh38
rs1051186 Chr.6: 41935068-41935068 on Build GRCh38
rs1043545586 Chr.6: 41935330-41935330 on Build GRCh38
rs11552785 Chr.6: 41935210-41935210 on Build GRCh38
rs1022669539 Chr.6: 41935010-41935010 on Build GRCh38
rs902837375 Chr.6: 41935161-41935161 on Build GRCh38
rs1051654595 Chr.6: 41935273-41935273 on Build GRCh38
rs960416831 Chr.6: 41935138-41935138 on Build GRCh38
rs755870776 Chr.6: 41934900-41934900 on Build GRCh38
rs941308124 Chr.6: 41934913-41934913 on Build GRCh38
rs768339846 Chr.6: 41935115-41935115 on Build GRCh38
rs972699302 Chr.6: 41935247-41935247 on Build GRCh38
rs910095407 Chr.6: 41935205-41935205 on Build GRCh38
rs1040078560 Chr.6: 41935153-41935153 on Build GRCh38
rs572165991 Chr.6: 41934921-41934921 on Build GRCh38
rs944602982 Chr.6: 41935083-41935083 on Build GRCh38
rs545214043 Chr.6: 41935255-41935255 on Build GRCh38
rs1050187102 Chr.6: 41935065-41935066 on Build GRCh38
rs3218108 Chr.6: 41934917-41934917 on Build GRCh38

View Full Product Details