Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44172223-44172721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN11
Entrez Gene ID
11131
Gene Name
calpain 11
Gene Aliases
calpain11
UniGene
Hs.225953
Gene Chromosome Location
Chr.6: 44158811-44184402 on Build GRCh38
Transcripts
NM_007058.3 XM_006714984.1 XM_006714985.1 XM_006714987.1 XM_011514274.1 XM_011514275.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5961248 c.494T>C p.F165S Chr.6: 44172386-44172386 on Build GRCh38
3352282 c.435C>G p.I145M Chr.6: 44172327-44172327 on Build GRCh38
4603135 c.425_426insGGCTGCC p.I145fs*48 Chr.6: 44172317-44172318 on Build GRCh38
6381382 c.416G>A p.C139Y Chr.6: 44172308-44172308 on Build GRCh38
5961249 c.494T>C p.F165S Chr.6: 44172386-44172386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997911041 Chr.6: 44172264-44172264 on Build GRCh38
rs762610901 Chr.6: 44172363-44172363 on Build GRCh38
rs183410962 Chr.6: 44172675-44172675 on Build GRCh38
rs773278222 Chr.6: 44172390-44172390 on Build GRCh38
rs114969347 Chr.6: 44172621-44172621 on Build GRCh38
rs905463740 Chr.6: 44172260-44172260 on Build GRCh38
rs201360460 Chr.6: 44172278-44172278 on Build GRCh38
rs1010580120 Chr.6: 44172544-44172544 on Build GRCh38
rs1418487 Chr.6: 44172538-44172538 on Build GRCh38
rs767643842 Chr.6: 44172389-44172389 on Build GRCh38
rs767432787 Chr.6: 44172254-44172254 on Build GRCh38
rs754636663 Chr.6: 44172328-44172328 on Build GRCh38
rs376991093 Chr.6: 44172339-44172339 on Build GRCh38
rs1049491864 Chr.6: 44172398-44172398 on Build GRCh38
rs147029811 Chr.6: 44172515-44172515 on Build GRCh38
rs867849105 Chr.6: 44172347-44172347 on Build GRCh38
rs930686152 Chr.6: 44172672-44172672 on Build GRCh38
rs187597064 Chr.6: 44172345-44172345 on Build GRCh38
rs536441657 Chr.6: 44172367-44172367 on Build GRCh38
rs978297388 Chr.6: 44172618-44172618 on Build GRCh38
rs111320370 Chr.6: 44172317-44172317 on Build GRCh38
rs534051096 Chr.6: 44172501-44172501 on Build GRCh38
rs1418488 Chr.6: 44172583-44172583 on Build GRCh38
rs978245200 Chr.6: 44172336-44172336 on Build GRCh38
rs866138097 Chr.6: 44172324-44172326 on Build GRCh38
rs960855078 Chr.6: 44172625-44172625 on Build GRCh38
rs905430971 Chr.6: 44172566-44172566 on Build GRCh38
rs576347559 Chr.6: 44172369-44172369 on Build GRCh38
rs370784911 Chr.6: 44172461-44172461 on Build GRCh38
rs61403960 Chr.6: 44172659-44172659 on Build GRCh38
rs74993668 Chr.6: 44172437-44172437 on Build GRCh38
rs761942642 Chr.6: 44172669-44172669 on Build GRCh38
rs546211585 Chr.6: 44172251-44172251 on Build GRCh38
rs777715152 Chr.6: 44172361-44172361 on Build GRCh38
rs1028430359 Chr.6: 44172268-44172268 on Build GRCh38
rs527919779 Chr.6: 44172541-44172541 on Build GRCh38
rs766281978 Chr.6: 44172424-44172424 on Build GRCh38
rs751249452 Chr.6: 44172305-44172306 on Build GRCh38
rs397947482 Chr.6: 44172324-44172325 on Build GRCh38
rs1022548316 Chr.6: 44172325-44172325 on Build GRCh38
rs57288791 Chr.6: 44172317-44172318 on Build GRCh38
rs900988056 Chr.6: 44172505-44172505 on Build GRCh38
rs747688113 Chr.6: 44172340-44172340 on Build GRCh38
rs939889861 Chr.6: 44172477-44172477 on Build GRCh38
rs768866187 Chr.6: 44172364-44172364 on Build GRCh38
rs767096762 Chr.6: 44172389-44172390 on Build GRCh38
rs746442126 Chr.6: 44172358-44172358 on Build GRCh38
rs966953930 Chr.6: 44172611-44172611 on Build GRCh38
rs566775227 Chr.6: 44172457-44172457 on Build GRCh38
rs959797247 Chr.6: 44172307-44172307 on Build GRCh38
rs143129879 Chr.6: 44172247-44172247 on Build GRCh38
rs754807237 Chr.6: 44172436-44172436 on Build GRCh38
rs202080178 Chr.6: 44172304-44172304 on Build GRCh38
rs374346436 Chr.6: 44172362-44172362 on Build GRCh38
rs953724059 Chr.6: 44172290-44172290 on Build GRCh38
rs774641753 Chr.6: 44172366-44172366 on Build GRCh38
rs760647465 Chr.6: 44172395-44172395 on Build GRCh38
rs753670184 Chr.6: 44172434-44172434 on Build GRCh38
rs991349897 Chr.6: 44172656-44172656 on Build GRCh38
rs766230788 Chr.6: 44172308-44172308 on Build GRCh38
rs757937446 Chr.6: 44172344-44172344 on Build GRCh38
rs190904283 Chr.6: 44172574-44172574 on Build GRCh38
rs753601236 Chr.6: 44172327-44172327 on Build GRCh38

View Full Product Details