Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49866188-49866699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISP1
Entrez Gene ID
167
Gene Name
cysteine rich secretory protein 1
Gene Aliases
AEGL1, ARP, CRISP-1, HEL-S-57, HSCRISP1D, HSCRISP1G, HUMARP
UniGene
Hs.109620
Gene Chromosome Location
Chr.6: 49834257-49877096 on Build GRCh38
Transcripts
NM_001131.2 NM_001205220.1 NM_170609.1

SNP Detail

SNP ID Chromosome Location
rs528186609 Chr.6: 49866560-49866560 on Build GRCh38
rs561616251 Chr.6: 49866389-49866389 on Build GRCh38
rs888003245 Chr.6: 49866592-49866592 on Build GRCh38
rs964810821 Chr.6: 49866549-49866549 on Build GRCh38
rs926266527 Chr.6: 49866641-49866641 on Build GRCh38
rs573029543 Chr.6: 49866219-49866219 on Build GRCh38
rs17663968 Chr.6: 49866575-49866575 on Build GRCh38
rs956842376 Chr.6: 49866519-49866519 on Build GRCh38
rs912401656 Chr.6: 49866538-49866538 on Build GRCh38
rs943770048 Chr.6: 49866540-49866540 on Build GRCh38
rs1013696330 Chr.6: 49866407-49866407 on Build GRCh38
rs933822871 Chr.6: 49866282-49866282 on Build GRCh38
rs1056575371 Chr.6: 49866420-49866420 on Build GRCh38
rs553724325 Chr.6: 49866579-49866579 on Build GRCh38
rs896671283 Chr.6: 49866643-49866643 on Build GRCh38
rs896669561 Chr.6: 49866463-49866463 on Build GRCh38
rs150761997 Chr.6: 49866601-49866601 on Build GRCh38
rs960795271 Chr.6: 49866357-49866358 on Build GRCh38
rs769333116 Chr.6: 49866514-49866514 on Build GRCh38
rs141771621 Chr.6: 49866584-49866584 on Build GRCh38
rs35069279 Chr.6: 49866656-49866657 on Build GRCh38
rs559472730 Chr.6: 49866499-49866499 on Build GRCh38
rs1023759849 Chr.6: 49866657-49866657 on Build GRCh38
rs1017589798 Chr.6: 49866516-49866516 on Build GRCh38
rs887996726 Chr.6: 49866504-49866504 on Build GRCh38
rs571354285 Chr.6: 49866583-49866583 on Build GRCh38
rs912278761 Chr.6: 49866220-49866220 on Build GRCh38
rs1048484856 Chr.6: 49866484-49866484 on Build GRCh38
rs189679309 Chr.6: 49866308-49866308 on Build GRCh38
rs552971807 Chr.6: 49866668-49866668 on Build GRCh38
rs975257774 Chr.6: 49866276-49866276 on Build GRCh38
rs542681922 Chr.6: 49866478-49866478 on Build GRCh38
rs922406256 Chr.6: 49866281-49866281 on Build GRCh38
rs770501119 Chr.6: 49866589-49866589 on Build GRCh38
rs904957526 Chr.6: 49866408-49866408 on Build GRCh38
rs937659199 Chr.6: 49866651-49866651 on Build GRCh38
rs550590192 Chr.6: 49866587-49866587 on Build GRCh38
rs759464801 Chr.6: 49866629-49866629 on Build GRCh38
rs967902707 Chr.6: 49866617-49866617 on Build GRCh38
rs536223090 Chr.6: 49866623-49866623 on Build GRCh38
rs144686568 Chr.6: 49866268-49866268 on Build GRCh38
rs1015517194 Chr.6: 49866333-49866333 on Build GRCh38
rs913814012 Chr.6: 49866310-49866310 on Build GRCh38
rs747634896 Chr.6: 49866315-49866315 on Build GRCh38
rs541987500 Chr.6: 49866612-49866612 on Build GRCh38
rs987963099 Chr.6: 49866532-49866532 on Build GRCh38
rs1030405536 Chr.6: 49866581-49866581 on Build GRCh38
rs937732646 Chr.6: 49866414-49866414 on Build GRCh38
rs545554922 Chr.6: 49866266-49866266 on Build GRCh38
rs780593668 Chr.6: 49866244-49866244 on Build GRCh38
rs528889423 Chr.6: 49866430-49866430 on Build GRCh38
rs138559076 Chr.6: 49866332-49866332 on Build GRCh38
rs993667595 Chr.6: 49866483-49866483 on Build GRCh38
rs964757090 Chr.6: 49866274-49866274 on Build GRCh38

View Full Product Details