Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 121449253-121449756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJA1
Entrez Gene ID
2697
Gene Name
gap junction protein alpha 1
Gene Aliases
AVSD3, CMDR, CX43, EKVP, GJAL, HLHS1, HSS, ODDD, PPKCA
UniGene
Hs.700699
Gene Chromosome Location
Chr.6: 121435577-121449744 on Build GRCh38
Transcripts
NM_000165.4

SNP Detail

SNP ID Chromosome Location
rs907137387 Chr.6: 121449488-121449488 on Build GRCh38
rs1045586301 Chr.6: 121449483-121449492 on Build GRCh38
rs540478741 Chr.6: 121449514-121449514 on Build GRCh38
rs1030552210 Chr.6: 121449659-121449659 on Build GRCh38
rs749953074 Chr.6: 121449330-121449330 on Build GRCh38
rs750807625 Chr.6: 121449703-121449704 on Build GRCh38
rs12212865 Chr.6: 121449318-121449318 on Build GRCh38
rs1017806248 Chr.6: 121449641-121449641 on Build GRCh38
rs945317035 Chr.6: 121449295-121449295 on Build GRCh38
rs111485072 Chr.6: 121449682-121449682 on Build GRCh38
rs1005956301 Chr.6: 121449623-121449623 on Build GRCh38
rs1025955323 Chr.6: 121449519-121449519 on Build GRCh38
rs978365477 Chr.6: 121449701-121449701 on Build GRCh38
rs374104162 Chr.6: 121449703-121449703 on Build GRCh38
rs34127088 Chr.6: 121449495-121449495 on Build GRCh38
rs556587636 Chr.6: 121449643-121449643 on Build GRCh38
rs567727782 Chr.6: 121449503-121449503 on Build GRCh38
rs536283987 Chr.6: 121449568-121449568 on Build GRCh38
rs936703761 Chr.6: 121449718-121449718 on Build GRCh38
rs547546766 Chr.6: 121449413-121449413 on Build GRCh38
rs545138467 Chr.6: 121449495-121449495 on Build GRCh38
rs565222467 Chr.6: 121449580-121449580 on Build GRCh38
rs78394273 Chr.6: 121449729-121449729 on Build GRCh38
rs976656840 Chr.6: 121449647-121449647 on Build GRCh38
rs758792254 Chr.6: 121449556-121449556 on Build GRCh38
rs531165846 Chr.6: 121449581-121449581 on Build GRCh38
rs534316459 Chr.6: 121449402-121449402 on Build GRCh38
rs1049349 Chr.6: 121449496-121449496 on Build GRCh38
rs948601789 Chr.6: 121449417-121449417 on Build GRCh38
rs1055649347 Chr.6: 121449365-121449365 on Build GRCh38
rs11541536 Chr.6: 121449600-121449600 on Build GRCh38
rs755560671 Chr.6: 121449371-121449371 on Build GRCh38
rs573053530 Chr.6: 121449328-121449328 on Build GRCh38

View Full Product Details