Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 139178869-139179373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECA
Entrez Gene ID
51696
Gene Name
hdc homolog, cell cycle regulator
Gene Aliases
HDC, HDCL, HHDC, dJ225E12.1
UniGene
Hs.197644
Gene Chromosome Location
Chr.6: 139135112-139180809 on Build GRCh38
Transcripts
NM_016217.2

SNP Detail

SNP ID Chromosome Location
rs1018055348 Chr.6: 139179169-139179169 on Build GRCh38
rs181815548 Chr.6: 139179173-139179173 on Build GRCh38
rs992055200 Chr.6: 139179282-139179282 on Build GRCh38
rs995187924 Chr.6: 139179184-139179184 on Build GRCh38
rs774742287 Chr.6: 139178937-139178937 on Build GRCh38
rs917822369 Chr.6: 139179331-139179331 on Build GRCh38
rs981322518 Chr.6: 139179022-139179022 on Build GRCh38
rs566511091 Chr.6: 139179007-139179007 on Build GRCh38
rs1010256796 Chr.6: 139179309-139179309 on Build GRCh38
rs927440984 Chr.6: 139179347-139179347 on Build GRCh38
rs775615248 Chr.6: 139178999-139178999 on Build GRCh38
rs1040575020 Chr.6: 139179202-139179202 on Build GRCh38
rs542560874 Chr.6: 139179326-139179326 on Build GRCh38
rs900670814 Chr.6: 139179228-139179228 on Build GRCh38
rs958647153 Chr.6: 139179057-139179057 on Build GRCh38
rs1051223165 Chr.6: 139179070-139179070 on Build GRCh38
rs73556663 Chr.6: 139179229-139179229 on Build GRCh38
rs760941996 Chr.6: 139179144-139179144 on Build GRCh38
rs563935233 Chr.6: 139179295-139179295 on Build GRCh38
rs918553390 Chr.6: 139178943-139178943 on Build GRCh38
rs7745259 Chr.6: 139178998-139178998 on Build GRCh38
rs936051162 Chr.6: 139179020-139179020 on Build GRCh38
rs1020686718 Chr.6: 139179310-139179310 on Build GRCh38
rs554909668 Chr.6: 139179349-139179349 on Build GRCh38
rs148047269 Chr.6: 139179053-139179053 on Build GRCh38
rs973985421 Chr.6: 139179060-139179060 on Build GRCh38
rs527284456 Chr.6: 139178924-139178924 on Build GRCh38
rs1054643282 Chr.6: 139179230-139179230 on Build GRCh38
rs760045028 Chr.6: 139178979-139178979 on Build GRCh38
rs537903292 Chr.6: 139179016-139179016 on Build GRCh38
rs754004115 Chr.6: 139179252-139179252 on Build GRCh38
rs1047139008 Chr.6: 139179100-139179101 on Build GRCh38
rs141738650 Chr.6: 139179334-139179334 on Build GRCh38
rs919800416 Chr.6: 139179066-139179066 on Build GRCh38
rs538863685 Chr.6: 139179152-139179152 on Build GRCh38
rs797002889 Chr.6: 139179139-139179150 on Build GRCh38
rs571437191 Chr.6: 139179067-139179067 on Build GRCh38
rs981006382 Chr.6: 139179344-139179344 on Build GRCh38
rs917729171 Chr.6: 139178991-139178991 on Build GRCh38
rs951240735 Chr.6: 139178952-139178952 on Build GRCh38
rs1003363703 Chr.6: 139179341-139179341 on Build GRCh38
rs974136297 Chr.6: 139178930-139178930 on Build GRCh38
rs1054823876 Chr.6: 139179041-139179041 on Build GRCh38

View Full Product Details