Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 160722316-160722826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLG
Entrez Gene ID
5340
Gene Name
plasminogen
Gene Aliases
-
UniGene
Hs.143436
Gene Chromosome Location
Chr.6: 160702193-160754054 on Build GRCh38
Transcripts
NM_000301.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6750092 c.1224G>T p.R408R Chr.6: 160722535-160722535 on Build GRCh38
4586442 c.1111A>G p.T371A Chr.6: 160722422-160722422 on Build GRCh38
6750093 c.1112C>T p.T371I Chr.6: 160722423-160722423 on Build GRCh38
6750094 c.1256+1G>A p.? Chr.6: 160722568-160722568 on Build GRCh38
6448019 c.1175C>T p.T392I Chr.6: 160722486-160722486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768154023 Chr.6: 160722366-160722366 on Build GRCh38
rs767145040 Chr.6: 160722385-160722385 on Build GRCh38
rs4252121 Chr.6: 160722602-160722602 on Build GRCh38
rs762821111 Chr.6: 160722365-160722365 on Build GRCh38
rs754725376 Chr.6: 160722708-160722709 on Build GRCh38
rs569230380 Chr.6: 160722352-160722352 on Build GRCh38
rs758597842 Chr.6: 160722425-160722425 on Build GRCh38
rs370893923 Chr.6: 160722588-160722588 on Build GRCh38
rs769370242 Chr.6: 160722457-160722457 on Build GRCh38
rs577403521 Chr.6: 160722733-160722733 on Build GRCh38
rs4252117 Chr.6: 160722344-160722344 on Build GRCh38
rs765725347 Chr.6: 160722593-160722593 on Build GRCh38
rs771610738 Chr.6: 160722467-160722467 on Build GRCh38
rs199914831 Chr.6: 160722607-160722607 on Build GRCh38
rs900579426 Chr.6: 160722355-160722355 on Build GRCh38
rs763563451 Chr.6: 160722595-160722595 on Build GRCh38
rs771522609 Chr.6: 160722565-160722565 on Build GRCh38
rs773855126 Chr.6: 160722369-160722369 on Build GRCh38
rs776405398 Chr.6: 160722580-160722580 on Build GRCh38
rs969828765 Chr.6: 160722499-160722499 on Build GRCh38
rs121918028 Chr.6: 160722431-160722431 on Build GRCh38
rs780940385 Chr.6: 160722423-160722423 on Build GRCh38
rs778793475 Chr.6: 160722559-160722559 on Build GRCh38
rs1015329067 Chr.6: 160722430-160722430 on Build GRCh38
rs747572282 Chr.6: 160722439-160722439 on Build GRCh38
rs754253835 Chr.6: 160722360-160722361 on Build GRCh38
rs760014589 Chr.6: 160722487-160722487 on Build GRCh38
rs770488932 Chr.6: 160722579-160722579 on Build GRCh38
rs142086839 Chr.6: 160722435-160722435 on Build GRCh38
rs760271152 Chr.6: 160722475-160722475 on Build GRCh38
rs996386174 Chr.6: 160722731-160722731 on Build GRCh38
rs1014945935 Chr.6: 160722764-160722764 on Build GRCh38
rs1027776438 Chr.6: 160722374-160722374 on Build GRCh38
rs552318802 Chr.6: 160722348-160722348 on Build GRCh38
rs773226077 Chr.6: 160722509-160722509 on Build GRCh38
rs1005137939 Chr.6: 160722757-160722757 on Build GRCh38
rs758508931 Chr.6: 160722523-160722523 on Build GRCh38
rs73017951 Chr.6: 160722754-160722754 on Build GRCh38
rs1039524398 Chr.6: 160722618-160722618 on Build GRCh38
rs769578807 Chr.6: 160722449-160722449 on Build GRCh38
rs4252118 Chr.6: 160722360-160722360 on Build GRCh38
rs370326625 Chr.6: 160722459-160722459 on Build GRCh38
rs757413544 Chr.6: 160722604-160722604 on Build GRCh38
rs397843659 Chr.6: 160722367-160722367 on Build GRCh38
rs900610546 Chr.6: 160722634-160722634 on Build GRCh38
rs772811538 Chr.6: 160722468-160722468 on Build GRCh38
rs754160565 Chr.6: 160722482-160722482 on Build GRCh38
rs751773919 Chr.6: 160722534-160722534 on Build GRCh38
rs1016967321 Chr.6: 160722760-160722760 on Build GRCh38
rs768546781 Chr.6: 160722547-160722547 on Build GRCh38
rs749021358 Chr.6: 160722545-160722545 on Build GRCh38
rs4252120 Chr.6: 160722576-160722576 on Build GRCh38
rs1049677544 Chr.6: 160722732-160722732 on Build GRCh38
rs748115521 Chr.6: 160722564-160722564 on Build GRCh38
rs776088327 Chr.6: 160722594-160722594 on Build GRCh38
rs772727378 Chr.6: 160722568-160722568 on Build GRCh38
rs376572197 Chr.6: 160722452-160722452 on Build GRCh38
rs778073217 Chr.6: 160722461-160722461 on Build GRCh38
rs960953621 Chr.6: 160722772-160722772 on Build GRCh38
rs749182016 Chr.6: 160722454-160722454 on Build GRCh38
rs751585449 Chr.6: 160722603-160722603 on Build GRCh38
rs766585160 Chr.6: 160722359-160722360 on Build GRCh38
rs765815120 Chr.6: 160722515-160722515 on Build GRCh38
rs372689013 Chr.6: 160722478-160722478 on Build GRCh38
rs1018142719 Chr.6: 160722353-160722353 on Build GRCh38
rs1016440595 Chr.6: 160722409-160722409 on Build GRCh38
rs753294113 Chr.6: 160722521-160722521 on Build GRCh38
rs754325596 Chr.6: 160722418-160722418 on Build GRCh38
rs761377369 Chr.6: 160722380-160722380 on Build GRCh38
rs781542891 Chr.6: 160722543-160722543 on Build GRCh38
rs963656260 Chr.6: 160722763-160722763 on Build GRCh38
rs374260556 Chr.6: 160722614-160722614 on Build GRCh38
rs754567261 Chr.6: 160722357-160722357 on Build GRCh38
rs754728286 Chr.6: 160722612-160722612 on Build GRCh38
rs757425055 Chr.6: 160722540-160722540 on Build GRCh38
rs766685527 Chr.6: 160722408-160722408 on Build GRCh38
rs750126916 Chr.6: 160722390-160722390 on Build GRCh38
rs4252119 Chr.6: 160722533-160722533 on Build GRCh38
rs191668994 Chr.6: 160722407-160722407 on Build GRCh38
rs1005349860 Chr.6: 160722755-160722755 on Build GRCh38

View Full Product Details