Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 166307346-166307840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR18
Entrez Gene ID
285800
Gene Name
proline rich 18
Gene Aliases
-
UniGene
Hs.731862
Gene Chromosome Location
Chr.6: 166305680-166308383 on Build GRCh38
Transcripts
NM_175922.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4638622 c.376G>T p.G126W Chr.6: 166307767-166307767 on Build GRCh38
6756856 c.650G>A p.G217D Chr.6: 166307493-166307493 on Build GRCh38
4718090 c.383G>C p.C128S Chr.6: 166307760-166307760 on Build GRCh38
4770535 c.495delC p.S166fs*127 Chr.6: 166307648-166307648 on Build GRCh38
1744191 c.679C>T p.R227W Chr.6: 166307464-166307464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749809611 Chr.6: 166307775-166307777 on Build GRCh38
rs745408167 Chr.6: 166307680-166307680 on Build GRCh38
rs151246255 Chr.6: 166307495-166307495 on Build GRCh38
rs566981011 Chr.6: 166307413-166307413 on Build GRCh38
rs555771783 Chr.6: 166307463-166307463 on Build GRCh38
rs1033711023 Chr.6: 166307524-166307524 on Build GRCh38
rs758218727 Chr.6: 166307482-166307482 on Build GRCh38
rs776333433 Chr.6: 166307417-166307417 on Build GRCh38
rs777105560 Chr.6: 166307406-166307406 on Build GRCh38
rs972929057 Chr.6: 166307550-166307550 on Build GRCh38
rs1049423984 Chr.6: 166307423-166307423 on Build GRCh38
rs761967501 Chr.6: 166307579-166307579 on Build GRCh38
rs959424787 Chr.6: 166307530-166307530 on Build GRCh38
rs765951027 Chr.6: 166307819-166307819 on Build GRCh38
rs910941291 Chr.6: 166307446-166307446 on Build GRCh38
rs771092605 Chr.6: 166307788-166307788 on Build GRCh38
rs933612454 Chr.6: 166307420-166307420 on Build GRCh38
rs774077818 Chr.6: 166307801-166307801 on Build GRCh38
rs746549198 Chr.6: 166307408-166307408 on Build GRCh38
rs746498931 Chr.6: 166307493-166307493 on Build GRCh38
rs775671112 Chr.6: 166307512-166307512 on Build GRCh38
rs751565954 Chr.6: 166307659-166307659 on Build GRCh38
rs1055125661 Chr.6: 166307654-166307654 on Build GRCh38
rs760078791 Chr.6: 166307811-166307811 on Build GRCh38
rs577902206 Chr.6: 166307560-166307560 on Build GRCh38
rs749679057 Chr.6: 166307523-166307523 on Build GRCh38
rs765163754 Chr.6: 166307477-166307477 on Build GRCh38
rs769936339 Chr.6: 166307800-166307800 on Build GRCh38
rs200995730 Chr.6: 166307620-166307620 on Build GRCh38
rs534030399 Chr.6: 166307445-166307445 on Build GRCh38
rs1017104205 Chr.6: 166307545-166307546 on Build GRCh38
rs868812742 Chr.6: 166307715-166307715 on Build GRCh38
rs768565484 Chr.6: 166307795-166307796 on Build GRCh38
rs766729445 Chr.6: 166307618-166307618 on Build GRCh38
rs556456266 Chr.6: 166307540-166307540 on Build GRCh38
rs755709585 Chr.6: 166307681-166307681 on Build GRCh38
rs752567613 Chr.6: 166307479-166307479 on Build GRCh38
rs773375703 Chr.6: 166307722-166307722 on Build GRCh38
rs773537010 Chr.6: 166307612-166307612 on Build GRCh38
rs1046023754 Chr.6: 166307513-166307513 on Build GRCh38
rs774695390 Chr.6: 166307704-166307704 on Build GRCh38
rs769119126 Chr.6: 166307690-166307690 on Build GRCh38
rs753572265 Chr.6: 166307753-166307753 on Build GRCh38
rs945848657 Chr.6: 166307604-166307604 on Build GRCh38
rs751387681 Chr.6: 166307739-166307739 on Build GRCh38
rs763925761 Chr.6: 166307643-166307643 on Build GRCh38
rs771578999 Chr.6: 166307387-166307387 on Build GRCh38
rs973427312 Chr.6: 166307381-166307381 on Build GRCh38
rs772648679 Chr.6: 166307775-166307775 on Build GRCh38
rs922886372 Chr.6: 166307404-166307404 on Build GRCh38
rs747074038 Chr.6: 166307785-166307785 on Build GRCh38
rs763333136 Chr.6: 166307447-166307447 on Build GRCh38
rs754794387 Chr.6: 166307468-166307468 on Build GRCh38
rs369396640 Chr.6: 166307764-166307764 on Build GRCh38
rs755263070 Chr.6: 166307756-166307756 on Build GRCh38
rs537800335 Chr.6: 166307509-166307509 on Build GRCh38
rs13204594 Chr.6: 166307723-166307723 on Build GRCh38
rs1054099396 Chr.6: 166307578-166307578 on Build GRCh38
rs756729523 Chr.6: 166307662-166307662 on Build GRCh38
rs772736959 Chr.6: 166307716-166307716 on Build GRCh38
rs770193945 Chr.6: 166307442-166307442 on Build GRCh38
rs746211088 Chr.6: 166307421-166307435 on Build GRCh38
rs935745794 Chr.6: 166307572-166307572 on Build GRCh38
rs774240520 Chr.6: 166307458-166307458 on Build GRCh38
rs201295448 Chr.6: 166307732-166307732 on Build GRCh38
rs779555801 Chr.6: 166307749-166307749 on Build GRCh38
rs758282551 Chr.6: 166307627-166307627 on Build GRCh38
rs745560683 Chr.6: 166307502-166307502 on Build GRCh38
rs200470806 Chr.6: 166307760-166307760 on Build GRCh38
rs768974534 Chr.6: 166307538-166307538 on Build GRCh38
rs754884357 Chr.6: 166307365-166307365 on Build GRCh38
rs533053957 Chr.6: 166307795-166307795 on Build GRCh38
rs767459528 Chr.6: 166307744-166307744 on Build GRCh38
rs961807306 Chr.6: 166307546-166307546 on Build GRCh38
rs767844908 Chr.6: 166307464-166307464 on Build GRCh38
rs200347308 Chr.6: 166307372-166307372 on Build GRCh38
rs375458610 Chr.6: 166307615-166307615 on Build GRCh38
rs777777819 Chr.6: 166307483-166307483 on Build GRCh38
rs1019071631 Chr.6: 166307803-166307803 on Build GRCh38
rs776038357 Chr.6: 166307444-166307444 on Build GRCh38
rs779764072 Chr.6: 166307682-166307682 on Build GRCh38
rs750415482 Chr.6: 166307808-166307808 on Build GRCh38
rs762634933 Chr.6: 166307733-166307733 on Build GRCh38
rs767441150 Chr.6: 166307806-166307806 on Build GRCh38
rs748701015 Chr.6: 166307706-166307706 on Build GRCh38
rs907138317 Chr.6: 166307517-166307517 on Build GRCh38
rs770989824 Chr.6: 166307728-166307728 on Build GRCh38
rs761694090 Chr.6: 166307743-166307743 on Build GRCh38
rs756819681 Chr.6: 166307494-166307494 on Build GRCh38
rs749871133 Chr.6: 166307747-166307747 on Build GRCh38
rs774441320 Chr.6: 166307806-166307808 on Build GRCh38
rs7757150 Chr.6: 166307736-166307736 on Build GRCh38
rs141594777 Chr.6: 166307374-166307374 on Build GRCh38
rs866289085 Chr.6: 166307661-166307661 on Build GRCh38
rs769471812 Chr.6: 166307505-166307505 on Build GRCh38
rs750728495 Chr.6: 166307466-166307466 on Build GRCh38
rs756557275 Chr.6: 166307451-166307452 on Build GRCh38
rs776687690 Chr.6: 166307792-166307792 on Build GRCh38
rs373872044 Chr.6: 166307478-166307478 on Build GRCh38
rs759881293 Chr.6: 166307425-166307425 on Build GRCh38
rs749609110 Chr.6: 166307683-166307683 on Build GRCh38
rs1045584102 Chr.6: 166307613-166307613 on Build GRCh38
rs562802984 Chr.6: 166307777-166307777 on Build GRCh38
rs780424571 Chr.6: 166307484-166307495 on Build GRCh38
rs372264427 Chr.6: 166307585-166307585 on Build GRCh38
rs780644610 Chr.6: 166307672-166307672 on Build GRCh38
rs544192282 Chr.6: 166307774-166307774 on Build GRCh38
rs1005561700 Chr.6: 166307679-166307679 on Build GRCh38
rs764687059 Chr.6: 166307454-166307454 on Build GRCh38
rs755685248 Chr.6: 166307748-166307748 on Build GRCh38
rs922820323 Chr.6: 166307553-166307553 on Build GRCh38

View Full Product Details