Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 28299234-28299743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB5
Entrez Gene ID
9586
Gene Name
cAMP responsive element binding protein 5
Gene Aliases
CRE-BPA, CREB-5
UniGene
Hs.437075
Gene Chromosome Location
Chr.7: 28299321-28825894 on Build GRCh38
Transcripts
NM_182899.4

SNP Detail

SNP ID Chromosome Location
rs754154429 Chr.7: 28299279-28299279 on Build GRCh38
rs185023547 Chr.7: 28299379-28299379 on Build GRCh38
rs543581414 Chr.7: 28299601-28299601 on Build GRCh38
rs980169058 Chr.7: 28299690-28299690 on Build GRCh38
rs914763370 Chr.7: 28299316-28299316 on Build GRCh38
rs987473046 Chr.7: 28299619-28299619 on Build GRCh38
rs531768405 Chr.7: 28299573-28299573 on Build GRCh38
rs757595773 Chr.7: 28299336-28299336 on Build GRCh38
rs56273634 Chr.7: 28299324-28299324 on Build GRCh38
rs899028466 Chr.7: 28299446-28299446 on Build GRCh38
rs541810785 Chr.7: 28299616-28299616 on Build GRCh38
rs1007913971 Chr.7: 28299397-28299397 on Build GRCh38
rs1033785738 Chr.7: 28299509-28299509 on Build GRCh38
rs978837314 Chr.7: 28299489-28299489 on Build GRCh38
rs1017637094 Chr.7: 28299459-28299459 on Build GRCh38
rs1009320914 Chr.7: 28299456-28299456 on Build GRCh38
rs551291884 Chr.7: 28299396-28299396 on Build GRCh38
rs1032793193 Chr.7: 28299510-28299510 on Build GRCh38
rs947465700 Chr.7: 28299321-28299321 on Build GRCh38
rs926987469 Chr.7: 28299692-28299692 on Build GRCh38
rs890421844 Chr.7: 28299415-28299415 on Build GRCh38
rs955883375 Chr.7: 28299584-28299584 on Build GRCh38
rs541969360 Chr.7: 28299345-28299345 on Build GRCh38
rs193259289 Chr.7: 28299367-28299367 on Build GRCh38
rs1037989667 Chr.7: 28299683-28299683 on Build GRCh38
rs1044115198 Chr.7: 28299307-28299307 on Build GRCh38
rs867518238 Chr.7: 28299558-28299558 on Build GRCh38
rs1037872217 Chr.7: 28299342-28299342 on Build GRCh38
rs1047623843 Chr.7: 28299712-28299712 on Build GRCh38
rs964761265 Chr.7: 28299463-28299463 on Build GRCh38
rs764952523 Chr.7: 28299325-28299325 on Build GRCh38
rs1398725 Chr.7: 28299637-28299637 on Build GRCh38
rs1052215362 Chr.7: 28299370-28299370 on Build GRCh38
rs187344022 Chr.7: 28299651-28299651 on Build GRCh38
rs138836200 Chr.7: 28299366-28299366 on Build GRCh38
rs745962980 Chr.7: 28299572-28299572 on Build GRCh38
rs991653223 Chr.7: 28299290-28299290 on Build GRCh38
rs977575084 Chr.7: 28299577-28299577 on Build GRCh38
rs576214461 Chr.7: 28299337-28299337 on Build GRCh38
rs992183067 Chr.7: 28299544-28299544 on Build GRCh38
rs929101174 Chr.7: 28299706-28299706 on Build GRCh38
rs1021884746 Chr.7: 28299563-28299563 on Build GRCh38
rs547982077 Chr.7: 28299638-28299638 on Build GRCh38
rs564661686 Chr.7: 28299391-28299391 on Build GRCh38

View Full Product Details