Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 45104460-45104961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNORA5A
Entrez Gene ID
654319
Gene Name
small nucleolar RNA, H/ACA box 5A
Gene Aliases
ACA5, ACA5A/B/C
UniGene
Hs.689711
Gene Chromosome Location
Chr.7: 45104349-45104482 on Build GRCh38
Transcripts
NR_002919.1 NR_002991.1 NM_001261834.1 NM_004749.3 NM_030900.3 NM_199122.2

Gene Symbol
SNORA5C
Entrez Gene ID
677796
Gene Name
small nucleolar RNA, H/ACA box 5C
Gene Aliases
ACA5c
UniGene
Hs.692718
Gene Chromosome Location
Chr.7: 45104906-45105042 on Build GRCh38
Transcripts
NR_002919.1 NR_002991.1 NM_001261834.1 NM_004749.3 NM_030900.3 NM_199122.2

Gene Symbol
TBRG4
Entrez Gene ID
9238
Gene Name
transforming growth factor beta regulator 4
Gene Aliases
CPR2, FASTKD4
UniGene
Hs.231411
Gene Chromosome Location
Chr.7: 45100100-45111747 on Build GRCh38
Transcripts
NR_002919.1 NR_002991.1 NM_001261834.1 NM_004749.3 NM_030900.3 NM_199122.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3929060 c.800C>T p.A267V Chr.7: 45104645-45104645 on Build GRCh38
7258261 c.829C>T p.R277W Chr.7: 45104616-45104616 on Build GRCh38
6797843 c.890A>G p.D297G Chr.7: 45104555-45104555 on Build GRCh38
5872770 c.821C>T p.P274L Chr.7: 45104624-45104624 on Build GRCh38
4162245 c.871A>G p.T291A Chr.7: 45104574-45104574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961645544 Chr.7: 45104938-45104938 on Build GRCh38
rs772958693 Chr.7: 45104637-45104637 on Build GRCh38
rs776985504 Chr.7: 45104628-45104628 on Build GRCh38
rs1046974270 Chr.7: 45104507-45104508 on Build GRCh38
rs747719901 Chr.7: 45104507-45104507 on Build GRCh38
rs374933985 Chr.7: 45104500-45104500 on Build GRCh38
rs370066970 Chr.7: 45104749-45104749 on Build GRCh38
rs373330133 Chr.7: 45104725-45104725 on Build GRCh38
rs766775394 Chr.7: 45104636-45104636 on Build GRCh38
rs777393084 Chr.7: 45104583-45104583 on Build GRCh38
rs532744186 Chr.7: 45104743-45104743 on Build GRCh38
rs201556853 Chr.7: 45104678-45104678 on Build GRCh38
rs377359964 Chr.7: 45104786-45104786 on Build GRCh38
rs779538171 Chr.7: 45104495-45104495 on Build GRCh38
rs1056228606 Chr.7: 45104814-45104814 on Build GRCh38
rs1034717390 Chr.7: 45104596-45104596 on Build GRCh38
rs372855977 Chr.7: 45104517-45104517 on Build GRCh38
rs775382726 Chr.7: 45104540-45104540 on Build GRCh38
rs753304099 Chr.7: 45104859-45104859 on Build GRCh38
rs775711965 Chr.7: 45104928-45104936 on Build GRCh38
rs749008576 Chr.7: 45104591-45104591 on Build GRCh38
rs780645949 Chr.7: 45104571-45104571 on Build GRCh38
rs768415098 Chr.7: 45104909-45104909 on Build GRCh38
rs770056741 Chr.7: 45104899-45104899 on Build GRCh38
rs759924107 Chr.7: 45104919-45104919 on Build GRCh38
rs774073359 Chr.7: 45104605-45104605 on Build GRCh38
rs757680481 Chr.7: 45104554-45104554 on Build GRCh38
rs762071924 Chr.7: 45104629-45104629 on Build GRCh38
rs765581334 Chr.7: 45104630-45104630 on Build GRCh38
rs750832942 Chr.7: 45104568-45104568 on Build GRCh38
rs777196714 Chr.7: 45104649-45104649 on Build GRCh38
rs1018296994 Chr.7: 45104560-45104560 on Build GRCh38
rs41280672 Chr.7: 45104514-45104514 on Build GRCh38
rs930501683 Chr.7: 45104526-45104526 on Build GRCh38
rs750144457 Chr.7: 45104869-45104869 on Build GRCh38
rs190179117 Chr.7: 45104806-45104806 on Build GRCh38
rs538060989 Chr.7: 45104811-45104811 on Build GRCh38
rs267601522 Chr.7: 45104611-45104611 on Build GRCh38
rs755699853 Chr.7: 45104573-45104573 on Build GRCh38
rs760836953 Chr.7: 45104546-45104546 on Build GRCh38
rs146485929 Chr.7: 45104645-45104645 on Build GRCh38
rs778566814 Chr.7: 45104687-45104687 on Build GRCh38
rs764289642 Chr.7: 45104626-45104626 on Build GRCh38
rs955485480 Chr.7: 45104724-45104724 on Build GRCh38
rs760645047 Chr.7: 45104617-45104617 on Build GRCh38
rs201938531 Chr.7: 45104750-45104750 on Build GRCh38
rs2289375 Chr.7: 45104804-45104804 on Build GRCh38
rs566422848 Chr.7: 45104760-45104760 on Build GRCh38
rs776297882 Chr.7: 45104595-45104595 on Build GRCh38
rs376659192 Chr.7: 45104572-45104572 on Build GRCh38
rs770232127 Chr.7: 45104881-45104881 on Build GRCh38
rs775401803 Chr.7: 45104865-45104865 on Build GRCh38
rs1044343280 Chr.7: 45104886-45104886 on Build GRCh38
rs752884619 Chr.7: 45104727-45104727 on Build GRCh38
rs1008229558 Chr.7: 45104903-45104903 on Build GRCh38
rs2289376 Chr.7: 45104895-45104895 on Build GRCh38
rs542979670 Chr.7: 45104905-45104905 on Build GRCh38
rs569339246 Chr.7: 45104857-45104857 on Build GRCh38
rs763214544 Chr.7: 45104740-45104740 on Build GRCh38
rs539447895 Chr.7: 45104873-45104873 on Build GRCh38
rs775533686 Chr.7: 45104616-45104616 on Build GRCh38
rs775274259 Chr.7: 45104719-45104719 on Build GRCh38
rs758786928 Chr.7: 45104570-45104570 on Build GRCh38
rs778511406 Chr.7: 45104866-45104866 on Build GRCh38
rs897300775 Chr.7: 45104509-45104509 on Build GRCh38
rs1048632042 Chr.7: 45104516-45104516 on Build GRCh38
rs747032040 Chr.7: 45104720-45104720 on Build GRCh38
rs562296179 Chr.7: 45104530-45104530 on Build GRCh38
rs201310402 Chr.7: 45104898-45104898 on Build GRCh38
rs182364100 Chr.7: 45104931-45104931 on Build GRCh38
rs143689271 Chr.7: 45104634-45104634 on Build GRCh38
rs768678211 Chr.7: 45104891-45104891 on Build GRCh38
rs753248606 Chr.7: 45104940-45104940 on Build GRCh38
rs767981285 Chr.7: 45104924-45104924 on Build GRCh38
rs764187647 Chr.7: 45104550-45104550 on Build GRCh38
rs1006856058 Chr.7: 45104542-45104542 on Build GRCh38
rs920536922 Chr.7: 45104744-45104744 on Build GRCh38
rs557135166 Chr.7: 45104856-45104856 on Build GRCh38
rs35312515 Chr.7: 45104942-45104943 on Build GRCh38
rs892090081 Chr.7: 45104820-45104820 on Build GRCh38
rs200776996 Chr.7: 45104504-45104504 on Build GRCh38
rs11541070 Chr.7: 45104606-45104606 on Build GRCh38
rs201920015 Chr.7: 45104501-45104501 on Build GRCh38
rs767503586 Chr.7: 45104531-45104531 on Build GRCh38
rs746805401 Chr.7: 45104881-45104881 on Build GRCh38
rs918366035 Chr.7: 45104765-45104765 on Build GRCh38
rs200081299 Chr.7: 45104717-45104717 on Build GRCh38
rs779829415 Chr.7: 45104877-45104877 on Build GRCh38
rs1048595627 Chr.7: 45104805-45104806 on Build GRCh38
rs754159048 Chr.7: 45104553-45104553 on Build GRCh38
rs527616648 Chr.7: 45104794-45104794 on Build GRCh38
rs776806871 Chr.7: 45104726-45104726 on Build GRCh38
rs761295620 Chr.7: 45104942-45104942 on Build GRCh38
rs770574123 Chr.7: 45104593-45104593 on Build GRCh38
rs374234109 Chr.7: 45104900-45104900 on Build GRCh38
rs997041113 Chr.7: 45104648-45104648 on Build GRCh38
rs745698557 Chr.7: 45104613-45104613 on Build GRCh38
rs529533304 Chr.7: 45104633-45104633 on Build GRCh38
rs140097576 Chr.7: 45104565-45104565 on Build GRCh38
rs371266326 Chr.7: 45104737-45104737 on Build GRCh38
rs771864304 Chr.7: 45104615-45104615 on Build GRCh38
rs748808163 Chr.7: 45104668-45104668 on Build GRCh38
rs759088932 Chr.7: 45104865-45104865 on Build GRCh38
rs770772425 Chr.7: 45104518-45104518 on Build GRCh38
rs745614597 Chr.7: 45104697-45104697 on Build GRCh38
rs774265854 Chr.7: 45104529-45104529 on Build GRCh38

View Full Product Details