Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100574479-100574988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRCH4
Entrez Gene ID
4034
Gene Name
leucine rich repeats and calponin homology domain containing 4
Gene Aliases
LRN, LRRN1, LRRN4, PP14183
UniGene
Hs.125742
Gene Chromosome Location
Chr.7: 100574011-100586188 on Build GRCh38
Transcripts
NM_001289934.1 NM_002319.4 NM_001289933.1

Gene Symbol
ZASP
Entrez Gene ID
101927655
Gene Name
ZO-2 associated speckle protein
Gene Aliases
-
UniGene
Hs.125742
Gene Chromosome Location
Chr.7: 100574035-100574733 on Build GRCh38
Transcripts
NM_001289934.1 NM_002319.4 NM_001289933.1

SNP Detail

SNP ID Chromosome Location
rs527998424 Chr.7: 100574586-100574586 on Build GRCh38
rs921818466 Chr.7: 100574834-100574834 on Build GRCh38
rs995923130 Chr.7: 100574614-100574614 on Build GRCh38
rs770710630 Chr.7: 100574555-100574555 on Build GRCh38
rs551008802 Chr.7: 100574568-100574568 on Build GRCh38
rs144874653 Chr.7: 100574618-100574619 on Build GRCh38
rs944631567 Chr.7: 100574797-100574797 on Build GRCh38
rs996358686 Chr.7: 100574651-100574651 on Build GRCh38
rs1051312132 Chr.7: 100574619-100574622 on Build GRCh38
rs561768975 Chr.7: 100574592-100574592 on Build GRCh38
rs184063729 Chr.7: 100574600-100574600 on Build GRCh38
rs74987581 Chr.7: 100574936-100574936 on Build GRCh38
rs577358634 Chr.7: 100574938-100574938 on Build GRCh38
rs778437467 Chr.7: 100574775-100574775 on Build GRCh38
rs376207067 Chr.7: 100574742-100574743 on Build GRCh38
rs962864593 Chr.7: 100574705-100574705 on Build GRCh38
rs987065894 Chr.7: 100574742-100574742 on Build GRCh38
rs555239576 Chr.7: 100574776-100574776 on Build GRCh38
rs1040272294 Chr.7: 100574647-100574647 on Build GRCh38
rs117649730 Chr.7: 100574632-100574632 on Build GRCh38
rs200891670 Chr.7: 100574632-100574633 on Build GRCh38
rs947146419 Chr.7: 100574761-100574761 on Build GRCh38
rs201832534 Chr.7: 100574741-100574742 on Build GRCh38
rs540617737 Chr.7: 100574774-100574775 on Build GRCh38
rs887569180 Chr.7: 100574905-100574905 on Build GRCh38
rs750668674 Chr.7: 100574835-100574835 on Build GRCh38
rs573766222 Chr.7: 100574818-100574818 on Build GRCh38
rs571816568 Chr.7: 100574627-100574627 on Build GRCh38
rs751716869 Chr.7: 100574682-100574682 on Build GRCh38
rs112916968 Chr.7: 100574634-100574634 on Build GRCh38
rs1035546842 Chr.7: 100574671-100574675 on Build GRCh38
rs537780084 Chr.7: 100574630-100574631 on Build GRCh38
rs1008736998 Chr.7: 100574599-100574599 on Build GRCh38
rs748909919 Chr.7: 100574542-100574543 on Build GRCh38
rs1053188033 Chr.7: 100574628-100574628 on Build GRCh38
rs925395741 Chr.7: 100574757-100574757 on Build GRCh38
rs900285291 Chr.7: 100574610-100574610 on Build GRCh38
rs960910769 Chr.7: 100574624-100574624 on Build GRCh38
rs117136190 Chr.7: 100574636-100574636 on Build GRCh38
rs903836480 Chr.7: 100574552-100574552 on Build GRCh38
rs960982901 Chr.7: 100574688-100574688 on Build GRCh38
rs371729570 Chr.7: 100574527-100574527 on Build GRCh38
rs887499950 Chr.7: 100574664-100574664 on Build GRCh38
rs954099036 Chr.7: 100574881-100574881 on Build GRCh38
rs961002967 Chr.7: 100574686-100574686 on Build GRCh38
rs1044157059 Chr.7: 100574540-100574540 on Build GRCh38
rs979526315 Chr.7: 100574629-100574629 on Build GRCh38
rs112238718 Chr.7: 100574902-100574902 on Build GRCh38
rs552159529 Chr.7: 100574699-100574699 on Build GRCh38
rs73711137 Chr.7: 100574872-100574872 on Build GRCh38
rs983493101 Chr.7: 100574621-100574621 on Build GRCh38
rs956974788 Chr.7: 100574759-100574759 on Build GRCh38
rs1034993262 Chr.7: 100574577-100574577 on Build GRCh38
rs988195208 Chr.7: 100574762-100574762 on Build GRCh38
rs547421596 Chr.7: 100574626-100574626 on Build GRCh38
rs866545385 Chr.7: 100574630-100574630 on Build GRCh38
rs983288577 Chr.7: 100574583-100574583 on Build GRCh38
rs934892230 Chr.7: 100574749-100574749 on Build GRCh38
rs1028385012 Chr.7: 100574879-100574879 on Build GRCh38
rs529166555 Chr.7: 100574618-100574618 on Build GRCh38
rs763072491 Chr.7: 100574959-100574959 on Build GRCh38
rs560613700 Chr.7: 100574576-100574576 on Build GRCh38
rs775494586 Chr.7: 100574649-100574649 on Build GRCh38
rs981317657 Chr.7: 100574552-100574552 on Build GRCh38
rs935361888 Chr.7: 100574631-100574631 on Build GRCh38
rs1040303254 Chr.7: 100574799-100574799 on Build GRCh38
rs896141549 Chr.7: 100574924-100574924 on Build GRCh38
rs982831970 Chr.7: 100574691-100574691 on Build GRCh38
rs979983945 Chr.7: 100574748-100574748 on Build GRCh38
rs969539769 Chr.7: 100574711-100574711 on Build GRCh38
rs188852580 Chr.7: 100574617-100574617 on Build GRCh38
rs921438526 Chr.7: 100574713-100574713 on Build GRCh38
rs1048817417 Chr.7: 100574663-100574663 on Build GRCh38
rs1004560364 Chr.7: 100574669-100574669 on Build GRCh38
rs1052828981 Chr.7: 100574755-100574756 on Build GRCh38
rs931853580 Chr.7: 100574861-100574861 on Build GRCh38
rs912566471 Chr.7: 100574777-100574777 on Build GRCh38
rs767612040 Chr.7: 100574802-100574802 on Build GRCh38
rs1005035934 Chr.7: 100574907-100574907 on Build GRCh38
rs770117081 Chr.7: 100574582-100574582 on Build GRCh38
rs562279883 Chr.7: 100574504-100574504 on Build GRCh38
rs375091814 Chr.7: 100574536-100574536 on Build GRCh38
rs550275620 Chr.7: 100574625-100574625 on Build GRCh38
rs1011739585 Chr.7: 100574633-100574636 on Build GRCh38
rs534114518 Chr.7: 100574687-100574687 on Build GRCh38
rs886381900 Chr.7: 100574780-100574784 on Build GRCh38
rs1023786229 Chr.7: 100574709-100574709 on Build GRCh38
rs1044086726 Chr.7: 100574769-100574769 on Build GRCh38
rs941429545 Chr.7: 100574590-100574590 on Build GRCh38
rs1015179663 Chr.7: 100574623-100574623 on Build GRCh38
rs557738767 Chr.7: 100574515-100574515 on Build GRCh38
rs1036009044 Chr.7: 100574913-100574913 on Build GRCh38
rs986993032 Chr.7: 100574899-100574899 on Build GRCh38
rs80089855 Chr.7: 100574775-100574776 on Build GRCh38

View Full Product Details