Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158849108-158849609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MNDA
Entrez Gene ID
4332
Gene Name
myeloid cell nuclear differentiation antigen
Gene Aliases
PYHIN3
UniGene
Hs.153837
Gene Chromosome Location
Chr.1: 158831378-158849480 on Build GRCh38
Transcripts
NM_002432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1639568 c.1216G>A p.V406I Chr.1: 158849229-158849229 on Build GRCh38
6507977 c.1207C>A p.P403T Chr.1: 158849220-158849220 on Build GRCh38
5895504 c.1205G>A p.G402E Chr.1: 158849218-158849218 on Build GRCh38
424268 c.1185delG p.A396fs*9 Chr.1: 158849198-158849198 on Build GRCh38
4446523 c.1177-3A>G p.? Chr.1: 158849187-158849187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567160173 Chr.1: 158849488-158849488 on Build GRCh38
rs998542192 Chr.1: 158849165-158849165 on Build GRCh38
rs532989730 Chr.1: 158849260-158849260 on Build GRCh38
rs892274077 Chr.1: 158849449-158849449 on Build GRCh38
rs182868751 Chr.1: 158849169-158849169 on Build GRCh38
rs984934828 Chr.1: 158849543-158849543 on Build GRCh38
rs116085800 Chr.1: 158849392-158849392 on Build GRCh38
rs943333823 Chr.1: 158849344-158849344 on Build GRCh38
rs373639519 Chr.1: 158849173-158849173 on Build GRCh38
rs929491319 Chr.1: 158849284-158849284 on Build GRCh38
rs753166099 Chr.1: 158849249-158849249 on Build GRCh38
rs1009421015 Chr.1: 158849460-158849460 on Build GRCh38
rs193037875 Chr.1: 158849226-158849226 on Build GRCh38
rs532631846 Chr.1: 158849556-158849556 on Build GRCh38
rs190290242 Chr.1: 158849140-158849140 on Build GRCh38
rs367659098 Chr.1: 158849213-158849213 on Build GRCh38
rs772016102 Chr.1: 158849179-158849179 on Build GRCh38
rs746483414 Chr.1: 158849202-158849202 on Build GRCh38
rs755945134 Chr.1: 158849280-158849283 on Build GRCh38
rs749868974 Chr.1: 158849503-158849503 on Build GRCh38
rs978386483 Chr.1: 158849270-158849270 on Build GRCh38
rs1039989449 Chr.1: 158849132-158849132 on Build GRCh38
rs751427712 Chr.1: 158849285-158849285 on Build GRCh38
rs199984814 Chr.1: 158849223-158849223 on Build GRCh38
rs74122496 Chr.1: 158849512-158849512 on Build GRCh38
rs374008772 Chr.1: 158849271-158849271 on Build GRCh38
rs80254262 Chr.1: 158849483-158849483 on Build GRCh38
rs41264047 Chr.1: 158849489-158849489 on Build GRCh38
rs992844361 Chr.1: 158849245-158849245 on Build GRCh38
rs147701010 Chr.1: 158849221-158849221 on Build GRCh38
rs745700528 Chr.1: 158849174-158849174 on Build GRCh38
rs959094380 Chr.1: 158849515-158849515 on Build GRCh38
rs544888072 Chr.1: 158849291-158849291 on Build GRCh38
rs980464829 Chr.1: 158849222-158849222 on Build GRCh38
rs530152871 Chr.1: 158849431-158849431 on Build GRCh38
rs1026544145 Chr.1: 158849529-158849529 on Build GRCh38
rs760777259 Chr.1: 158849254-158849254 on Build GRCh38
rs185813417 Chr.1: 158849350-158849350 on Build GRCh38
rs781200649 Chr.1: 158849305-158849305 on Build GRCh38
rs899136007 Chr.1: 158849452-158849452 on Build GRCh38
rs909305327 Chr.1: 158849311-158849311 on Build GRCh38
rs200798015 Chr.1: 158849199-158849199 on Build GRCh38
rs950830301 Chr.1: 158849162-158849162 on Build GRCh38
rs761448098 Chr.1: 158849217-158849217 on Build GRCh38
rs933787457 Chr.1: 158849294-158849294 on Build GRCh38
rs768171362 Chr.1: 158849203-158849203 on Build GRCh38
rs933903342 Chr.1: 158849396-158849396 on Build GRCh38
rs960682140 Chr.1: 158849505-158849505 on Build GRCh38
rs1009305078 Chr.1: 158849172-158849172 on Build GRCh38
rs1043455666 Chr.1: 158849465-158849465 on Build GRCh38
rs943401747 Chr.1: 158849401-158849401 on Build GRCh38
rs540239500 Chr.1: 158849494-158849494 on Build GRCh38
rs768027494 Chr.1: 158849229-158849229 on Build GRCh38
rs754124199 Chr.1: 158849157-158849157 on Build GRCh38
rs2261161 Chr.1: 158849534-158849534 on Build GRCh38
rs911969359 Chr.1: 158849399-158849399 on Build GRCh38
rs1033333535 Chr.1: 158849496-158849496 on Build GRCh38
rs757045346 Chr.1: 158849166-158849166 on Build GRCh38
rs566254150 Chr.1: 158849564-158849564 on Build GRCh38
rs140803418 Chr.1: 158849228-158849228 on Build GRCh38
rs964819267 Chr.1: 158849558-158849558 on Build GRCh38

View Full Product Details