Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 106112173-106112661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYPL1
Entrez Gene ID
6856
Gene Name
synaptophysin like 1
Gene Aliases
H-SP1, SYPL
UniGene
Hs.80919
Gene Chromosome Location
Chr.7: 106090368-106112647 on Build GRCh38
Transcripts
NM_006754.3 NM_182715.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5997248 c.33G>T p.R11R Chr.7: 106112497-106112497 on Build GRCh38
3778001 c.72C>T p.I24I Chr.7: 106112191-106112191 on Build GRCh38
1312562 c.87C>G p.L29L Chr.7: 106112176-106112176 on Build GRCh38
7392773 c.69G>C p.Q23H Chr.7: 106112194-106112194 on Build GRCh38
4412976 c.46G>A p.G16S Chr.7: 106112484-106112484 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs542494618 Chr.7: 106112417-106112417 on Build GRCh38
rs572311580 Chr.7: 106112618-106112618 on Build GRCh38
rs1052093431 Chr.7: 106112266-106112266 on Build GRCh38
rs893106179 Chr.7: 106112264-106112264 on Build GRCh38
rs376130471 Chr.7: 106112442-106112442 on Build GRCh38
rs748668915 Chr.7: 106112209-106112209 on Build GRCh38
rs751167848 Chr.7: 106112553-106112553 on Build GRCh38
rs772308930 Chr.7: 106112465-106112465 on Build GRCh38
rs376663730 Chr.7: 106112258-106112258 on Build GRCh38
rs531418959 Chr.7: 106112349-106112349 on Build GRCh38
rs770080004 Chr.7: 106112210-106112210 on Build GRCh38
rs779378012 Chr.7: 106112521-106112521 on Build GRCh38
rs763800615 Chr.7: 106112250-106112250 on Build GRCh38
rs766915615 Chr.7: 106112556-106112556 on Build GRCh38
rs758682523 Chr.7: 106112573-106112573 on Build GRCh38
rs762611278 Chr.7: 106112549-106112549 on Build GRCh38
rs764146438 Chr.7: 106112480-106112480 on Build GRCh38
rs765816089 Chr.7: 106112494-106112494 on Build GRCh38
rs780360003 Chr.7: 106112576-106112576 on Build GRCh38
rs766117364 Chr.7: 106112550-106112550 on Build GRCh38
rs1005857704 Chr.7: 106112295-106112295 on Build GRCh38
rs762280301 Chr.7: 106112226-106112227 on Build GRCh38
rs553750684 Chr.7: 106112472-106112472 on Build GRCh38
rs771418030 Chr.7: 106112453-106112453 on Build GRCh38
rs746067265 Chr.7: 106112460-106112460 on Build GRCh38
rs372856747 Chr.7: 106112542-106112542 on Build GRCh38
rs1041768237 Chr.7: 106112464-106112466 on Build GRCh38
rs750875492 Chr.7: 106112571-106112571 on Build GRCh38
rs776676628 Chr.7: 106112535-106112535 on Build GRCh38
rs754420564 Chr.7: 106112558-106112558 on Build GRCh38
rs750686658 Chr.7: 106112574-106112574 on Build GRCh38
rs927623119 Chr.7: 106112197-106112197 on Build GRCh38
rs373449152 Chr.7: 106112265-106112265 on Build GRCh38
rs887070318 Chr.7: 106112445-106112445 on Build GRCh38
rs927636085 Chr.7: 106112517-106112517 on Build GRCh38
rs758009323 Chr.7: 106112493-106112493 on Build GRCh38
rs760619388 Chr.7: 106112249-106112249 on Build GRCh38
rs1014585700 Chr.7: 106112585-106112585 on Build GRCh38
rs1050969724 Chr.7: 106112552-106112552 on Build GRCh38
rs13246433 Chr.7: 106112358-106112358 on Build GRCh38
rs1039522454 Chr.7: 106112290-106112290 on Build GRCh38
rs751027121 Chr.7: 106112499-106112499 on Build GRCh38
rs928827719 Chr.7: 106112348-106112352 on Build GRCh38
rs377219931 Chr.7: 106112554-106112554 on Build GRCh38
rs981633438 Chr.7: 106112347-106112348 on Build GRCh38
rs541724729 Chr.7: 106112633-106112633 on Build GRCh38
rs530280636 Chr.7: 106112276-106112276 on Build GRCh38
rs946512407 Chr.7: 106112285-106112285 on Build GRCh38
rs971550730 Chr.7: 106112625-106112625 on Build GRCh38
rs567147151 Chr.7: 106112483-106112483 on Build GRCh38
rs369661398 Chr.7: 106112540-106112540 on Build GRCh38
rs944154163 Chr.7: 106112509-106112509 on Build GRCh38
rs998105984 Chr.7: 106112299-106112299 on Build GRCh38
rs776424542 Chr.7: 106112481-106112481 on Build GRCh38
rs575045341 Chr.7: 106112291-106112291 on Build GRCh38
rs866204607 Chr.7: 106112504-106112504 on Build GRCh38
rs749729141 Chr.7: 106112446-106112446 on Build GRCh38
rs200891171 Chr.7: 106112534-106112534 on Build GRCh38
rs746091652 Chr.7: 106112523-106112523 on Build GRCh38
rs561539383 Chr.7: 106112247-106112247 on Build GRCh38
rs17426359 Chr.7: 106112636-106112636 on Build GRCh38
rs759345923 Chr.7: 106112282-106112282 on Build GRCh38
rs576172548 Chr.7: 106112544-106112544 on Build GRCh38
rs780787022 Chr.7: 106112567-106112567 on Build GRCh38
rs1012992893 Chr.7: 106112598-106112598 on Build GRCh38
rs753610989 Chr.7: 106112255-106112255 on Build GRCh38
rs200172873 Chr.7: 106112565-106112565 on Build GRCh38
rs774757124 Chr.7: 106112455-106112455 on Build GRCh38
rs772278094 Chr.7: 106112527-106112527 on Build GRCh38
rs764992362 Chr.7: 106112485-106112485 on Build GRCh38
rs11545140 Chr.7: 106112246-106112246 on Build GRCh38
rs951692360 Chr.7: 106112294-106112294 on Build GRCh38
rs903281403 Chr.7: 106112268-106112268 on Build GRCh38
rs999352299 Chr.7: 106112273-106112273 on Build GRCh38
rs961759484 Chr.7: 106112587-106112587 on Build GRCh38
rs757029973 Chr.7: 106112257-106112257 on Build GRCh38
rs176501 Chr.7: 106112245-106112245 on Build GRCh38
rs763092781 Chr.7: 106112234-106112234 on Build GRCh38
rs757747921 Chr.7: 106112514-106112514 on Build GRCh38
rs139360207 Chr.7: 106112578-106112578 on Build GRCh38
rs947340655 Chr.7: 106112424-106112424 on Build GRCh38
rs370234036 Chr.7: 106112235-106112235 on Build GRCh38
rs756581995 Chr.7: 106112440-106112440 on Build GRCh38
rs987862761 Chr.7: 106112531-106112531 on Build GRCh38
rs775572127 Chr.7: 106112467-106112467 on Build GRCh38
rs992109832 Chr.7: 106112388-106112388 on Build GRCh38
rs893647185 Chr.7: 106112283-106112283 on Build GRCh38
rs1004309538 Chr.7: 106112451-106112451 on Build GRCh38
rs767795567 Chr.7: 106112484-106112485 on Build GRCh38
rs748636940 Chr.7: 106112262-106112262 on Build GRCh38
rs773589373 Chr.7: 106112223-106112223 on Build GRCh38
rs568998091 Chr.7: 106112320-106112320 on Build GRCh38
rs6962415 Chr.7: 106112259-106112259 on Build GRCh38
rs775560792 Chr.7: 106112529-106112529 on Build GRCh38
rs536071079 Chr.7: 106112513-106112513 on Build GRCh38
rs176502 Chr.7: 106112374-106112374 on Build GRCh38
rs373409803 Chr.7: 106112237-106112237 on Build GRCh38
rs778412371 Chr.7: 106112501-106112501 on Build GRCh38
rs1005826411 Chr.7: 106112584-106112584 on Build GRCh38
rs530769292 Chr.7: 106112545-106112545 on Build GRCh38
rs1029707199 Chr.7: 106112302-106112302 on Build GRCh38
rs996463254 Chr.7: 106112284-106112284 on Build GRCh38
rs972088072 Chr.7: 106112511-106112511 on Build GRCh38
rs981712183 Chr.7: 106112512-106112512 on Build GRCh38
rs978375088 Chr.7: 106112422-106112422 on Build GRCh38

View Full Product Details