Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117381783-117382288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASZ1
Entrez Gene ID
136991
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Gene Aliases
ALP1, ANKL1, C7orf7, CT1.19, GASZ, Orf3
UniGene
Hs.352412
Gene Chromosome Location
Chr.7: 117363222-117427523 on Build GRCh38
Transcripts
NM_001301821.1 NM_001301822.1 NM_130768.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3662874 c.813-6A>G p.? Chr.7: 117382150-117382150 on Build GRCh38
3257188 c.832G>A p.D278N Chr.7: 117382125-117382125 on Build GRCh38
1488155 c.821C>G p.T274R Chr.7: 117382136-117382136 on Build GRCh38
1084459 c.878A>G p.D293G Chr.7: 117382079-117382079 on Build GRCh38
1755041 c.857T>A p.L286H Chr.7: 117382100-117382100 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751842190 Chr.7: 117382078-117382078 on Build GRCh38
rs554383492 Chr.7: 117382106-117382106 on Build GRCh38
rs1042405158 Chr.7: 117382030-117382031 on Build GRCh38
rs993964061 Chr.7: 117381917-117381917 on Build GRCh38
rs777563848 Chr.7: 117382184-117382184 on Build GRCh38
rs932940928 Chr.7: 117382219-117382219 on Build GRCh38
rs899227043 Chr.7: 117381898-117381898 on Build GRCh38
rs922831568 Chr.7: 117382179-117382179 on Build GRCh38
rs915447655 Chr.7: 117382227-117382227 on Build GRCh38
rs1012481753 Chr.7: 117382097-117382097 on Build GRCh38
rs199772988 Chr.7: 117382099-117382099 on Build GRCh38
rs946749756 Chr.7: 117382231-117382233 on Build GRCh38
rs927784116 Chr.7: 117382241-117382241 on Build GRCh38
rs760776291 Chr.7: 117382225-117382225 on Build GRCh38
rs547163722 Chr.7: 117381944-117381944 on Build GRCh38
rs558683755 Chr.7: 117382236-117382236 on Build GRCh38
rs781482198 Chr.7: 117382092-117382092 on Build GRCh38
rs529028004 Chr.7: 117381863-117381863 on Build GRCh38
rs757696915 Chr.7: 117382137-117382138 on Build GRCh38
rs1003968845 Chr.7: 117382243-117382243 on Build GRCh38
rs964766582 Chr.7: 117382088-117382088 on Build GRCh38
rs951789472 Chr.7: 117382120-117382120 on Build GRCh38
rs765117647 Chr.7: 117382052-117382052 on Build GRCh38
rs773050044 Chr.7: 117382032-117382032 on Build GRCh38
rs1047900467 Chr.7: 117381934-117381934 on Build GRCh38
rs769084575 Chr.7: 117382195-117382195 on Build GRCh38
rs965699375 Chr.7: 117382161-117382161 on Build GRCh38
rs907507291 Chr.7: 117382160-117382160 on Build GRCh38
rs890591861 Chr.7: 117381998-117381998 on Build GRCh38
rs766209385 Chr.7: 117382037-117382037 on Build GRCh38
rs570809693 Chr.7: 117382245-117382245 on Build GRCh38
rs781558494 Chr.7: 117382181-117382183 on Build GRCh38
rs764284179 Chr.7: 117382072-117382072 on Build GRCh38
rs771767183 Chr.7: 117382163-117382163 on Build GRCh38
rs960525473 Chr.7: 117382256-117382256 on Build GRCh38
rs762989561 Chr.7: 117382063-117382063 on Build GRCh38
rs771820565 Chr.7: 117382027-117382027 on Build GRCh38
rs977001389 Chr.7: 117382177-117382177 on Build GRCh38
rs768674677 Chr.7: 117382247-117382247 on Build GRCh38
rs372808264 Chr.7: 117382017-117382017 on Build GRCh38
rs879563904 Chr.7: 117382197-117382197 on Build GRCh38
rs192454506 Chr.7: 117381893-117381893 on Build GRCh38
rs146322411 Chr.7: 117382116-117382116 on Build GRCh38
rs376694351 Chr.7: 117382189-117382189 on Build GRCh38
rs747673498 Chr.7: 117382158-117382158 on Build GRCh38
rs757479192 Chr.7: 117382089-117382089 on Build GRCh38
rs377254051 Chr.7: 117382193-117382193 on Build GRCh38
rs1042498851 Chr.7: 117382240-117382240 on Build GRCh38
rs545662789 Chr.7: 117382048-117382048 on Build GRCh38
rs752788327 Chr.7: 117382058-117382058 on Build GRCh38
rs187935385 Chr.7: 117381881-117381881 on Build GRCh38
rs534147279 Chr.7: 117382223-117382223 on Build GRCh38
rs932826242 Chr.7: 117381933-117381933 on Build GRCh38
rs566754172 Chr.7: 117382119-117382119 on Build GRCh38
rs760534183 Chr.7: 117382035-117382035 on Build GRCh38
rs1003423334 Chr.7: 117382070-117382070 on Build GRCh38
rs983169901 Chr.7: 117382155-117382155 on Build GRCh38
rs776878098 Chr.7: 117382264-117382264 on Build GRCh38
rs111285711 Chr.7: 117382206-117382206 on Build GRCh38
rs901315609 Chr.7: 117381928-117381928 on Build GRCh38
rs773865053 Chr.7: 117382021-117382021 on Build GRCh38
rs747645815 Chr.7: 117382022-117382022 on Build GRCh38
rs1010558871 Chr.7: 117382185-117382185 on Build GRCh38
rs74720429 Chr.7: 117381841-117381841 on Build GRCh38

View Full Product Details