Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161122843-161123346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEDD
Entrez Gene ID
9191
Gene Name
death effector domain containing
Gene Aliases
CASP8IP1, DEDD1, DEFT, FLDED1, KE05
UniGene
Hs.744092
Gene Chromosome Location
Chr.1: 161120977-161132783 on Build GRCh38
Transcripts
NM_001039711.1 NM_001039712.1 NM_032998.2 XM_005245597.3 XM_005245598.4 XM_005245599.2 XM_005245600.2 XM_017002802.1 NM_001185092.1 XM_006711341.3 XM_017001371.1

Gene Symbol
NIT1
Entrez Gene ID
4817
Gene Name
nitrilase 1
Gene Aliases
-
UniGene
Hs.146406
Gene Chromosome Location
Chr.1: 161118072-161125446 on Build GRCh38
Transcripts
NM_001039711.1 NM_001039712.1 NM_032998.2 XM_005245597.3 XM_005245598.4 XM_005245599.2 XM_005245600.2 XM_017002802.1 NM_001185092.1 XM_006711341.3 XM_017001371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7135553 c.592C>A p.Q198K Chr.1: 161123063-161123063 on Build GRCh38
5430515 c.502C>T p.R168* Chr.1: 161123153-161123153 on Build GRCh38
2086736 c.530G>A p.R177Q Chr.1: 161123125-161123125 on Build GRCh38
4224229 c.530G>A p.R177Q Chr.1: 161123125-161123125 on Build GRCh38
4499239 c.537C>T p.R179R Chr.1: 161123118-161123118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921371919 Chr.1: 161123040-161123040 on Build GRCh38
rs766710268 Chr.1: 161123272-161123272 on Build GRCh38
rs558356123 Chr.1: 161123115-161123115 on Build GRCh38
rs772988092 Chr.1: 161123007-161123007 on Build GRCh38
rs920103902 Chr.1: 161122897-161122897 on Build GRCh38
rs780445260 Chr.1: 161123067-161123067 on Build GRCh38
rs749463178 Chr.1: 161123217-161123217 on Build GRCh38
rs1053881869 Chr.1: 161123252-161123254 on Build GRCh38
rs778004213 Chr.1: 161123207-161123207 on Build GRCh38
rs1056207579 Chr.1: 161123235-161123235 on Build GRCh38
rs534874719 Chr.1: 161123092-161123092 on Build GRCh38
rs766997282 Chr.1: 161123137-161123137 on Build GRCh38
rs769589484 Chr.1: 161123003-161123003 on Build GRCh38
rs762918432 Chr.1: 161123014-161123014 on Build GRCh38
rs760440014 Chr.1: 161123152-161123152 on Build GRCh38
rs368552890 Chr.1: 161123256-161123256 on Build GRCh38
rs763812597 Chr.1: 161123155-161123155 on Build GRCh38
rs759002967 Chr.1: 161123036-161123036 on Build GRCh38
rs748205290 Chr.1: 161123190-161123190 on Build GRCh38
rs373852129 Chr.1: 161123050-161123050 on Build GRCh38
rs370630271 Chr.1: 161123161-161123161 on Build GRCh38
rs201370857 Chr.1: 161123038-161123038 on Build GRCh38
rs374707294 Chr.1: 161123120-161123120 on Build GRCh38
rs926267043 Chr.1: 161123175-161123175 on Build GRCh38
rs775880098 Chr.1: 161122997-161122997 on Build GRCh38
rs766988478 Chr.1: 161123037-161123037 on Build GRCh38
rs759463050 Chr.1: 161123135-161123135 on Build GRCh38
rs937796246 Chr.1: 161123185-161123185 on Build GRCh38
rs751119955 Chr.1: 161123030-161123032 on Build GRCh38
rs781289830 Chr.1: 161123187-161123187 on Build GRCh38
rs746439985 Chr.1: 161122947-161122947 on Build GRCh38
rs763654942 Chr.1: 161123261-161123261 on Build GRCh38
rs776425371 Chr.1: 161123264-161123264 on Build GRCh38
rs768620883 Chr.1: 161123090-161123090 on Build GRCh38
rs760076059 Chr.1: 161123279-161123279 on Build GRCh38
rs766206679 Chr.1: 161123017-161123017 on Build GRCh38
rs775186397 Chr.1: 161123255-161123255 on Build GRCh38
rs191962431 Chr.1: 161122879-161122879 on Build GRCh38
rs768730469 Chr.1: 161122972-161122972 on Build GRCh38
rs752019537 Chr.1: 161123173-161123173 on Build GRCh38
rs923468936 Chr.1: 161123119-161123119 on Build GRCh38
rs765984803 Chr.1: 161122966-161122969 on Build GRCh38
rs555435758 Chr.1: 161122899-161122899 on Build GRCh38
rs775139574 Chr.1: 161123151-161123151 on Build GRCh38
rs909858744 Chr.1: 161123118-161123118 on Build GRCh38
rs780218843 Chr.1: 161122966-161122966 on Build GRCh38
rs769972871 Chr.1: 161123097-161123097 on Build GRCh38
rs750781889 Chr.1: 161123061-161123061 on Build GRCh38
rs755936414 Chr.1: 161122875-161122878 on Build GRCh38
rs375821148 Chr.1: 161123201-161123201 on Build GRCh38
rs887706602 Chr.1: 161123322-161123322 on Build GRCh38
rs755497175 Chr.1: 161123183-161123183 on Build GRCh38
rs1035266837 Chr.1: 161122913-161122913 on Build GRCh38
rs377566442 Chr.1: 161123125-161123125 on Build GRCh38
rs773041210 Chr.1: 161123108-161123108 on Build GRCh38
rs774186944 Chr.1: 161123236-161123236 on Build GRCh38
rs763229280 Chr.1: 161123162-161123162 on Build GRCh38
rs74124643 Chr.1: 161123063-161123063 on Build GRCh38
rs182968623 Chr.1: 161123058-161123058 on Build GRCh38
rs993406565 Chr.1: 161122993-161122993 on Build GRCh38
rs747507944 Chr.1: 161123068-161123068 on Build GRCh38
rs17389237 Chr.1: 161122971-161122971 on Build GRCh38
rs756139518 Chr.1: 161123196-161123196 on Build GRCh38
rs766698121 Chr.1: 161123168-161123168 on Build GRCh38
rs772698164 Chr.1: 161122959-161122959 on Build GRCh38
rs761469811 Chr.1: 161123267-161123267 on Build GRCh38
rs375587444 Chr.1: 161123252-161123252 on Build GRCh38
rs372379351 Chr.1: 161123226-161123226 on Build GRCh38
rs745863968 Chr.1: 161123251-161123251 on Build GRCh38
rs751466070 Chr.1: 161123020-161123020 on Build GRCh38

View Full Product Details