Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 165428627-165429131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RXRG
Entrez Gene ID
6258
Gene Name
retinoid X receptor gamma
Gene Aliases
NR2B3, RXRC
UniGene
Hs.26550
Gene Chromosome Location
Chr.1: 165400922-165445355 on Build GRCh38
Transcripts
NM_001256570.1 NM_006917.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4470579 c.166C>T p.L56L Chr.1: 165428850-165428850 on Build GRCh38
2088930 c.257C>T p.A86V Chr.1: 165428759-165428759 on Build GRCh38
424458 c.159A>C p.P53P Chr.1: 165428857-165428857 on Build GRCh38
6357162 c.199G>A p.G67S Chr.1: 165428817-165428817 on Build GRCh38
7326514 c.187_188insC p.L63fs*74 Chr.1: 165428828-165428829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770620106 Chr.1: 165428857-165428857 on Build GRCh38
rs369858210 Chr.1: 165428979-165428979 on Build GRCh38
rs776724134 Chr.1: 165428971-165428973 on Build GRCh38
rs149455886 Chr.1: 165428834-165428834 on Build GRCh38
rs781134922 Chr.1: 165428831-165428831 on Build GRCh38
rs145797395 Chr.1: 165428762-165428762 on Build GRCh38
rs975246590 Chr.1: 165429044-165429044 on Build GRCh38
rs370244132 Chr.1: 165428696-165428696 on Build GRCh38
rs142730991 Chr.1: 165428759-165428759 on Build GRCh38
rs750811270 Chr.1: 165428879-165428879 on Build GRCh38
rs574623447 Chr.1: 165428937-165428937 on Build GRCh38
rs995448667 Chr.1: 165428736-165428736 on Build GRCh38
rs117921572 Chr.1: 165428649-165428649 on Build GRCh38
rs753204832 Chr.1: 165428991-165428992 on Build GRCh38
rs780033756 Chr.1: 165428722-165428722 on Build GRCh38
rs749137726 Chr.1: 165428912-165428912 on Build GRCh38
rs968538320 Chr.1: 165429041-165429041 on Build GRCh38
rs773412624 Chr.1: 165428972-165428972 on Build GRCh38
rs779942426 Chr.1: 165428827-165428827 on Build GRCh38
rs768047738 Chr.1: 165428677-165428677 on Build GRCh38
rs1038114960 Chr.1: 165428943-165428943 on Build GRCh38
rs759609044 Chr.1: 165429000-165429000 on Build GRCh38
rs371375925 Chr.1: 165428957-165428957 on Build GRCh38
rs1036704593 Chr.1: 165428986-165428986 on Build GRCh38
rs758489485 Chr.1: 165428823-165428823 on Build GRCh38
rs560261474 Chr.1: 165429018-165429018 on Build GRCh38
rs750674966 Chr.1: 165428822-165428822 on Build GRCh38
rs986703903 Chr.1: 165429079-165429079 on Build GRCh38
rs774069121 Chr.1: 165429001-165429001 on Build GRCh38
rs766657681 Chr.1: 165428882-165428882 on Build GRCh38
rs778676053 Chr.1: 165428922-165428922 on Build GRCh38
rs886830623 Chr.1: 165428660-165428660 on Build GRCh38
rs764728266 Chr.1: 165428780-165428780 on Build GRCh38
rs764306208 Chr.1: 165428688-165428688 on Build GRCh38
rs371155454 Chr.1: 165428925-165428925 on Build GRCh38
rs143885966 Chr.1: 165428841-165428841 on Build GRCh38
rs375743449 Chr.1: 165428990-165428990 on Build GRCh38
rs368291917 Chr.1: 165428969-165428969 on Build GRCh38
rs773692997 Chr.1: 165428858-165428858 on Build GRCh38
rs367816854 Chr.1: 165428948-165428948 on Build GRCh38
rs1022375119 Chr.1: 165429036-165429036 on Build GRCh38
rs752699044 Chr.1: 165428898-165428898 on Build GRCh38
rs113471 Chr.1: 165428878-165428878 on Build GRCh38
rs764675336 Chr.1: 165428861-165428861 on Build GRCh38
rs546073425 Chr.1: 165429064-165429064 on Build GRCh38
rs757744281 Chr.1: 165428803-165428803 on Build GRCh38
rs151322161 Chr.1: 165428742-165428742 on Build GRCh38
rs754205838 Chr.1: 165428800-165428800 on Build GRCh38
rs781077674 Chr.1: 165428894-165428894 on Build GRCh38
rs905423656 Chr.1: 165428980-165428980 on Build GRCh38
rs746975087 Chr.1: 165428723-165428723 on Build GRCh38
rs539278154 Chr.1: 165428729-165428729 on Build GRCh38
rs370579158 Chr.1: 165428855-165428855 on Build GRCh38
rs751848503 Chr.1: 165428997-165428997 on Build GRCh38
rs376567163 Chr.1: 165428836-165428836 on Build GRCh38
rs150448748 Chr.1: 165428758-165428758 on Build GRCh38
rs908494026 Chr.1: 165429082-165429082 on Build GRCh38
rs1033295939 Chr.1: 165429106-165429106 on Build GRCh38
rs769392173 Chr.1: 165428755-165428755 on Build GRCh38
rs745916177 Chr.1: 165428697-165428697 on Build GRCh38
rs755892073 Chr.1: 165428904-165428904 on Build GRCh38
rs1028180522 Chr.1: 165429063-165429063 on Build GRCh38
rs397862617 Chr.1: 165428820-165428820 on Build GRCh38
rs777314160 Chr.1: 165428872-165428872 on Build GRCh38
rs199523000 Chr.1: 165428856-165428856 on Build GRCh38
rs375033777 Chr.1: 165428921-165428921 on Build GRCh38
rs749226423 Chr.1: 165428926-165428926 on Build GRCh38
rs1010013845 Chr.1: 165429026-165429026 on Build GRCh38
rs149766171 Chr.1: 165428962-165428962 on Build GRCh38
rs759107051 Chr.1: 165428860-165428860 on Build GRCh38
rs752789069 Chr.1: 165429002-165429002 on Build GRCh38
rs373070041 Chr.1: 165428825-165428825 on Build GRCh38
rs756002805 Chr.1: 165429017-165429017 on Build GRCh38
rs906054448 Chr.1: 165429032-165429032 on Build GRCh38
rs939696922 Chr.1: 165428942-165428942 on Build GRCh38
rs776346206 Chr.1: 165428730-165428730 on Build GRCh38
rs374496115 Chr.1: 165428935-165428935 on Build GRCh38
rs755103416 Chr.1: 165428885-165428885 on Build GRCh38
rs765520178 Chr.1: 165428812-165428812 on Build GRCh38
rs374305512 Chr.1: 165428905-165428905 on Build GRCh38
rs370210993 Chr.1: 165428994-165428994 on Build GRCh38
rs575591593 Chr.1: 165428829-165428829 on Build GRCh38
rs756498388 Chr.1: 165428684-165428684 on Build GRCh38
rs200482382 Chr.1: 165428683-165428683 on Build GRCh38
rs754258823 Chr.1: 165428691-165428691 on Build GRCh38
rs761832945 Chr.1: 165428993-165428994 on Build GRCh38
rs532208042 Chr.1: 165429061-165429061 on Build GRCh38
rs908192532 Chr.1: 165428938-165428938 on Build GRCh38
rs776054493 Chr.1: 165428766-165428766 on Build GRCh38

View Full Product Details