Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94882040-94882544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNE2
Entrez Gene ID
9134
Gene Name
cyclin E2
Gene Aliases
CYCE2
UniGene
Hs.521693
Gene Chromosome Location
Chr.8: 94880224-94896671 on Build GRCh38
Transcripts
NM_057749.2 XM_017013957.1 XM_017013958.1 XM_017013959.1 XM_017013616.1 XM_017013618.1 XM_017013619.1

Gene Symbol
INTS8
Entrez Gene ID
55656
Gene Name
integrator complex subunit 8
Gene Aliases
C8orf52, INT8
UniGene
Hs.727669
Gene Chromosome Location
Chr.8: 94823290-94894664 on Build GRCh38
Transcripts
NM_057749.2 XM_017013957.1 XM_017013958.1 XM_017013959.1 XM_017013616.1 XM_017013618.1 XM_017013619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1458740 c.1049C>A p.P350H Chr.8: 94882184-94882184 on Build GRCh38
1674016 c.979T>C p.W327R Chr.8: 94882254-94882254 on Build GRCh38
6348880 c.1082C>A p.T361K Chr.8: 94882151-94882151 on Build GRCh38
1102693 c.1090T>G p.L364V Chr.8: 94882143-94882143 on Build GRCh38
5779152 c.1051A>G p.M351V Chr.8: 94882182-94882182 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764698405 Chr.8: 94882090-94882090 on Build GRCh38
rs141602286 Chr.8: 94882266-94882266 on Build GRCh38
rs892012307 Chr.8: 94882423-94882423 on Build GRCh38
rs771557976 Chr.8: 94882307-94882310 on Build GRCh38
rs768535563 Chr.8: 94882330-94882330 on Build GRCh38
rs573167895 Chr.8: 94882436-94882436 on Build GRCh38
rs933783876 Chr.8: 94882235-94882235 on Build GRCh38
rs773066507 Chr.8: 94882317-94882317 on Build GRCh38
rs753273433 Chr.8: 94882255-94882255 on Build GRCh38
rs757453877 Chr.8: 94882299-94882299 on Build GRCh38
rs752072919 Chr.8: 94882095-94882095 on Build GRCh38
rs746224353 Chr.8: 94882127-94882127 on Build GRCh38
rs760523773 Chr.8: 94882326-94882329 on Build GRCh38
rs967814733 Chr.8: 94882167-94882167 on Build GRCh38
rs908420612 Chr.8: 94882272-94882272 on Build GRCh38
rs201837249 Chr.8: 94882170-94882170 on Build GRCh38
rs968225290 Chr.8: 94882446-94882446 on Build GRCh38
rs1002971577 Chr.8: 94882405-94882405 on Build GRCh38
rs779003245 Chr.8: 94882310-94882310 on Build GRCh38
rs771915643 Chr.8: 94882174-94882174 on Build GRCh38
rs754028664 Chr.8: 94882276-94882276 on Build GRCh38
rs988518334 Chr.8: 94882172-94882172 on Build GRCh38
rs751679116 Chr.8: 94882117-94882118 on Build GRCh38
rs921094517 Chr.8: 94882199-94882199 on Build GRCh38
rs907391214 Chr.8: 94882346-94882346 on Build GRCh38
rs376184791 Chr.8: 94882151-94882151 on Build GRCh38
rs756266274 Chr.8: 94882131-94882131 on Build GRCh38
rs956389384 Chr.8: 94882099-94882099 on Build GRCh38
rs562263218 Chr.8: 94882463-94882463 on Build GRCh38
rs914247083 Chr.8: 94882218-94882218 on Build GRCh38
rs868546251 Chr.8: 94882214-94882214 on Build GRCh38
rs761395311 Chr.8: 94882301-94882301 on Build GRCh38
rs746988191 Chr.8: 94882320-94882320 on Build GRCh38
rs138458120 Chr.8: 94882274-94882274 on Build GRCh38
rs773686788 Chr.8: 94882296-94882297 on Build GRCh38
rs776396741 Chr.8: 94882204-94882204 on Build GRCh38
rs529797759 Chr.8: 94882507-94882507 on Build GRCh38
rs756384170 Chr.8: 94882496-94882496 on Build GRCh38
rs373041033 Chr.8: 94882332-94882332 on Build GRCh38
rs781393912 Chr.8: 94882120-94882123 on Build GRCh38
rs987726627 Chr.8: 94882119-94882119 on Build GRCh38
rs146388666 Chr.8: 94882273-94882273 on Build GRCh38
rs576814851 Chr.8: 94882396-94882396 on Build GRCh38
rs1021360645 Chr.8: 94882482-94882482 on Build GRCh38
rs975690146 Chr.8: 94882169-94882169 on Build GRCh38
rs370082768 Chr.8: 94882284-94882284 on Build GRCh38
rs754637542 Chr.8: 94882065-94882065 on Build GRCh38
rs747709521 Chr.8: 94882306-94882307 on Build GRCh38
rs749837679 Chr.8: 94882290-94882291 on Build GRCh38
rs748528238 Chr.8: 94882106-94882106 on Build GRCh38
rs764039885 Chr.8: 94882086-94882087 on Build GRCh38
rs117719646 Chr.8: 94882418-94882418 on Build GRCh38
rs887805125 Chr.8: 94882356-94882356 on Build GRCh38
rs1006619067 Chr.8: 94882381-94882381 on Build GRCh38
rs60994907 Chr.8: 94882442-94882442 on Build GRCh38
rs929401382 Chr.8: 94882345-94882345 on Build GRCh38
rs554714016 Chr.8: 94882088-94882088 on Build GRCh38
rs775356601 Chr.8: 94882181-94882181 on Build GRCh38
rs753135943 Chr.8: 94882454-94882454 on Build GRCh38
rs865967928 Chr.8: 94882168-94882168 on Build GRCh38
rs768629344 Chr.8: 94882187-94882187 on Build GRCh38
rs781575344 Chr.8: 94882102-94882102 on Build GRCh38
rs948815499 Chr.8: 94882325-94882325 on Build GRCh38
rs760033034 Chr.8: 94882220-94882220 on Build GRCh38
rs770888764 Chr.8: 94882339-94882340 on Build GRCh38
rs769239668 Chr.8: 94882291-94882294 on Build GRCh38
rs745939237 Chr.8: 94882311-94882311 on Build GRCh38
rs757361691 Chr.8: 94882119-94882120 on Build GRCh38
rs373155995 Chr.8: 94882219-94882219 on Build GRCh38
rs764760850 Chr.8: 94882206-94882206 on Build GRCh38
rs374512238 Chr.8: 94882319-94882319 on Build GRCh38
rs755469367 Chr.8: 94882100-94882100 on Build GRCh38
rs756532128 Chr.8: 94882145-94882146 on Build GRCh38
rs371847039 Chr.8: 94882108-94882108 on Build GRCh38
rs574843451 Chr.8: 94882162-94882162 on Build GRCh38
rs767917366 Chr.8: 94882221-94882221 on Build GRCh38
rs780521636 Chr.8: 94882198-94882204 on Build GRCh38
rs749571065 Chr.8: 94882150-94882150 on Build GRCh38
rs534318388 Chr.8: 94882400-94882400 on Build GRCh38

View Full Product Details