Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 127736333-127736835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYC
Entrez Gene ID
4609
Gene Name
v-myc avian myelocytomatosis viral oncogene homolog
Gene Aliases
MRTL, MYCC, bHLHe39, c-Myc
UniGene
Hs.202453
Gene Chromosome Location
Chr.8: 127736069-127741434 on Build GRCh38
Transcripts
NM_002467.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6207185 c.27C>T p.N9N Chr.8: 127736620-127736620 on Build GRCh38
6955610 c.5_6insT p.R5fs*17 Chr.8: 127736598-127736599 on Build GRCh38
3316824 c.30G>C p.Q10H Chr.8: 127736623-127736623 on Build GRCh38
240777 c.30+1G>T p.? Chr.8: 127736624-127736624 on Build GRCh38
1161715 c.19G>A p.V7M Chr.8: 127736612-127736612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs935427530 Chr.8: 127736756-127736756 on Build GRCh38
rs557901268 Chr.8: 127736506-127736506 on Build GRCh38
rs774938117 Chr.8: 127736719-127736719 on Build GRCh38
rs747141352 Chr.8: 127736607-127736607 on Build GRCh38
rs766983499 Chr.8: 127736577-127736577 on Build GRCh38
rs1045323017 Chr.8: 127736788-127736793 on Build GRCh38
rs759986640 Chr.8: 127736616-127736616 on Build GRCh38
rs535709985 Chr.8: 127736481-127736481 on Build GRCh38
rs989745198 Chr.8: 127736385-127736385 on Build GRCh38
rs777604765 Chr.8: 127736656-127736657 on Build GRCh38
rs773453291 Chr.8: 127736547-127736547 on Build GRCh38
rs931755035 Chr.8: 127736398-127736398 on Build GRCh38
rs770464239 Chr.8: 127736648-127736652 on Build GRCh38
rs775761084 Chr.8: 127736631-127736631 on Build GRCh38
rs893949916 Chr.8: 127736530-127736530 on Build GRCh38
rs907691279 Chr.8: 127736405-127736405 on Build GRCh38
rs373577293 Chr.8: 127736571-127736571 on Build GRCh38
rs76318023 Chr.8: 127736536-127736536 on Build GRCh38
rs554049915 Chr.8: 127736482-127736482 on Build GRCh38
rs1031309075 Chr.8: 127736358-127736358 on Build GRCh38
rs928364521 Chr.8: 127736364-127736364 on Build GRCh38
rs772459508 Chr.8: 127736602-127736602 on Build GRCh38
rs753723304 Chr.8: 127736589-127736589 on Build GRCh38
rs878995495 Chr.8: 127736543-127736543 on Build GRCh38
rs985838399 Chr.8: 127736400-127736400 on Build GRCh38
rs765884995 Chr.8: 127736659-127736659 on Build GRCh38
rs979834632 Chr.8: 127736358-127736358 on Build GRCh38
rs973364711 Chr.8: 127736725-127736725 on Build GRCh38
rs529123559 Chr.8: 127736795-127736795 on Build GRCh38
rs1011185108 Chr.8: 127736537-127736537 on Build GRCh38
rs369952293 Chr.8: 127736550-127736550 on Build GRCh38
rs549914515 Chr.8: 127736782-127736782 on Build GRCh38
rs190092855 Chr.8: 127736504-127736504 on Build GRCh38
rs543513193 Chr.8: 127736520-127736520 on Build GRCh38
rs888609496 Chr.8: 127736507-127736507 on Build GRCh38
rs921807840 Chr.8: 127736733-127736733 on Build GRCh38
rs756035058 Chr.8: 127736583-127736583 on Build GRCh38
rs985890826 Chr.8: 127736692-127736692 on Build GRCh38
rs756894698 Chr.8: 127736574-127736574 on Build GRCh38
rs931948337 Chr.8: 127736469-127736469 on Build GRCh38
rs2071346 Chr.8: 127736777-127736777 on Build GRCh38
rs796121384 Chr.8: 127736388-127736388 on Build GRCh38
rs776052954 Chr.8: 127736448-127736448 on Build GRCh38
rs771277797 Chr.8: 127736644-127736644 on Build GRCh38
rs777141161 Chr.8: 127736609-127736609 on Build GRCh38
rs777974417 Chr.8: 127736605-127736605 on Build GRCh38
rs780034860 Chr.8: 127736585-127736585 on Build GRCh38
rs568267700 Chr.8: 127736462-127736462 on Build GRCh38
rs1051854440 Chr.8: 127736497-127736497 on Build GRCh38
rs533487784 Chr.8: 127736752-127736752 on Build GRCh38
rs767039286 Chr.8: 127736554-127736554 on Build GRCh38
rs78120979 Chr.8: 127736460-127736460 on Build GRCh38
rs915064846 Chr.8: 127736392-127736392 on Build GRCh38
rs767216614 Chr.8: 127736643-127736643 on Build GRCh38
rs759881591 Chr.8: 127736564-127736564 on Build GRCh38
rs748361858 Chr.8: 127736599-127736599 on Build GRCh38
rs747682614 Chr.8: 127736656-127736656 on Build GRCh38
rs749985481 Chr.8: 127736581-127736581 on Build GRCh38
rs140142417 Chr.8: 127736747-127736747 on Build GRCh38
rs956585583 Chr.8: 127736383-127736383 on Build GRCh38
rs1048576281 Chr.8: 127736505-127736505 on Build GRCh38
rs987504296 Chr.8: 127736749-127736749 on Build GRCh38
rs944342103 Chr.8: 127736416-127736416 on Build GRCh38
rs550000037 Chr.8: 127736449-127736449 on Build GRCh38
rs4645949 Chr.8: 127736498-127736498 on Build GRCh38
rs758079498 Chr.8: 127736657-127736657 on Build GRCh38
rs576124632 Chr.8: 127736514-127736514 on Build GRCh38
rs541331885 Chr.8: 127736746-127736746 on Build GRCh38
rs761433935 Chr.8: 127736633-127736633 on Build GRCh38
rs201238897 Chr.8: 127736626-127736626 on Build GRCh38
rs777120045 Chr.8: 127736562-127736562 on Build GRCh38
rs778595707 Chr.8: 127736597-127736597 on Build GRCh38
rs765535915 Chr.8: 127736569-127736569 on Build GRCh38
rs911568835 Chr.8: 127736414-127736414 on Build GRCh38
rs973717300 Chr.8: 127736447-127736447 on Build GRCh38
rs760109975 Chr.8: 127736655-127736655 on Build GRCh38
rs753759396 Chr.8: 127736675-127736675 on Build GRCh38
rs73707293 Chr.8: 127736772-127736772 on Build GRCh38
rs4645950 Chr.8: 127736794-127736794 on Build GRCh38
rs771557035 Chr.8: 127736608-127736608 on Build GRCh38
rs754742138 Chr.8: 127736592-127736592 on Build GRCh38

View Full Product Details