Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 142874878-142875384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP11B1
Entrez Gene ID
1584
Gene Name
cytochrome P450 family 11 subfamily B member 1
Gene Aliases
CPN1, CYP11B, FHI, P450C11
UniGene
Hs.184927
Gene Chromosome Location
Chr.8: 142872354-142879839 on Build GRCh38
Transcripts
NM_000497.3 NM_001026213.1 XM_011516874.2 XM_011516875.1 XM_017013146.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5808202 c.1334T>G p.F445C Chr.8: 142875021-142875021 on Build GRCh38
6180267 c.1298G>C p.G433A Chr.8: 142875057-142875057 on Build GRCh38
749822 c.1144C>T p.L382L Chr.8: 142875290-142875290 on Build GRCh38
5772409 c.1272C>T p.N424N Chr.8: 142875083-142875083 on Build GRCh38
2867749 c.1201-1G>T p.? Chr.8: 142875155-142875155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958715423 Chr.8: 142874903-142874903 on Build GRCh38
rs778663484 Chr.8: 142875149-142875149 on Build GRCh38
rs760339638 Chr.8: 142875116-142875116 on Build GRCh38
rs774352542 Chr.8: 142874939-142874939 on Build GRCh38
rs754240896 Chr.8: 142875121-142875121 on Build GRCh38
rs28934586 Chr.8: 142875012-142875012 on Build GRCh38
rs761488596 Chr.8: 142874962-142874964 on Build GRCh38
rs746697113 Chr.8: 142875159-142875159 on Build GRCh38
rs61752796 Chr.8: 142874949-142874949 on Build GRCh38
rs375224541 Chr.8: 142875052-142875052 on Build GRCh38
rs772169059 Chr.8: 142875033-142875033 on Build GRCh38
rs4998896 Chr.8: 142875144-142875144 on Build GRCh38
rs373191820 Chr.8: 142875228-142875228 on Build GRCh38
rs1031238917 Chr.8: 142875150-142875150 on Build GRCh38
rs779842583 Chr.8: 142875145-142875145 on Build GRCh38
rs749285481 Chr.8: 142875100-142875100 on Build GRCh38
rs564754333 Chr.8: 142875142-142875142 on Build GRCh38
rs781052848 Chr.8: 142875034-142875034 on Build GRCh38
rs764996968 Chr.8: 142875311-142875311 on Build GRCh38
rs775013399 Chr.8: 142874966-142874966 on Build GRCh38
rs748098106 Chr.8: 142875234-142875234 on Build GRCh38
rs5293 Chr.8: 142875290-142875290 on Build GRCh38
rs763699043 Chr.8: 142874932-142874932 on Build GRCh38
rs529700971 Chr.8: 142875264-142875264 on Build GRCh38
rs370772088 Chr.8: 142874924-142874924 on Build GRCh38
rs771775313 Chr.8: 142875115-142875115 on Build GRCh38
rs142887967 Chr.8: 142875276-142875276 on Build GRCh38
rs61752795 Chr.8: 142875325-142875325 on Build GRCh38
rs780845350 Chr.8: 142875331-142875331 on Build GRCh38
rs4998897 Chr.8: 142875143-142875143 on Build GRCh38
rs370757808 Chr.8: 142875304-142875304 on Build GRCh38
rs751424855 Chr.8: 142874944-142874945 on Build GRCh38
rs780198531 Chr.8: 142874942-142874942 on Build GRCh38
rs775901877 Chr.8: 142875172-142875172 on Build GRCh38
rs751256857 Chr.8: 142874988-142874988 on Build GRCh38
rs757479374 Chr.8: 142875212-142875212 on Build GRCh38
rs745610620 Chr.8: 142875035-142875035 on Build GRCh38
rs373649246 Chr.8: 142874991-142874991 on Build GRCh38
rs769070161 Chr.8: 142875351-142875351 on Build GRCh38
rs769028095 Chr.8: 142874945-142874945 on Build GRCh38
rs774884519 Chr.8: 142875352-142875352 on Build GRCh38
rs769517888 Chr.8: 142875091-142875091 on Build GRCh38
rs768525580 Chr.8: 142875106-142875106 on Build GRCh38
rs778735553 Chr.8: 142875233-142875233 on Build GRCh38
rs554178535 Chr.8: 142875021-142875021 on Build GRCh38
rs759939173 Chr.8: 142875163-142875163 on Build GRCh38
rs749627330 Chr.8: 142875343-142875343 on Build GRCh38
rs777844820 Chr.8: 142875281-142875284 on Build GRCh38
rs750344288 Chr.8: 142875060-142875061 on Build GRCh38
rs1024429962 Chr.8: 142874933-142874933 on Build GRCh38
rs775455382 Chr.8: 142875047-142875047 on Build GRCh38
rs749018677 Chr.8: 142875146-142875146 on Build GRCh38
rs780289850 Chr.8: 142875341-142875341 on Build GRCh38
rs767690646 Chr.8: 142875215-142875215 on Build GRCh38
rs777138805 Chr.8: 142875162-142875162 on Build GRCh38
rs760880418 Chr.8: 142875306-142875306 on Build GRCh38
rs61752794 Chr.8: 142875332-142875332 on Build GRCh38
rs763573023 Chr.8: 142874975-142874975 on Build GRCh38
rs565440875 Chr.8: 142875119-142875119 on Build GRCh38
rs765025654 Chr.8: 142874957-142874957 on Build GRCh38
rs752865698 Chr.8: 142875220-142875239 on Build GRCh38
rs5316 Chr.8: 142875002-142875002 on Build GRCh38
rs985837633 Chr.8: 142874958-142874958 on Build GRCh38
rs746452616 Chr.8: 142875246-142875246 on Build GRCh38
rs577022490 Chr.8: 142874955-142874955 on Build GRCh38
rs193922537 Chr.8: 142875324-142875324 on Build GRCh38
rs201300785 Chr.8: 142875154-142875154 on Build GRCh38
rs753651666 Chr.8: 142875223-142875223 on Build GRCh38
rs377100267 Chr.8: 142875064-142875064 on Build GRCh38
rs755536549 Chr.8: 142875076-142875076 on Build GRCh38
rs770647465 Chr.8: 142875161-142875161 on Build GRCh38
rs761835287 Chr.8: 142875302-142875302 on Build GRCh38
rs750679917 Chr.8: 142875305-142875305 on Build GRCh38
rs542739915 Chr.8: 142875057-142875057 on Build GRCh38
rs368155421 Chr.8: 142875058-142875058 on Build GRCh38
rs199688260 Chr.8: 142875067-142875067 on Build GRCh38
rs751135858 Chr.8: 142875329-142875329 on Build GRCh38
rs752395337 Chr.8: 142875241-142875241 on Build GRCh38
rs371855747 Chr.8: 142875060-142875060 on Build GRCh38
rs200342486 Chr.8: 142875056-142875056 on Build GRCh38
rs565297677 Chr.8: 142874898-142874898 on Build GRCh38
rs764138882 Chr.8: 142875197-142875197 on Build GRCh38
rs372693198 Chr.8: 142875210-142875210 on Build GRCh38
rs367634557 Chr.8: 142874994-142874994 on Build GRCh38
rs776284924 Chr.8: 142874951-142874951 on Build GRCh38
rs200711326 Chr.8: 142875269-142875269 on Build GRCh38
rs368954289 Chr.8: 142875158-142875158 on Build GRCh38
rs753984710 Chr.8: 142875020-142875020 on Build GRCh38
rs768226546 Chr.8: 142875053-142875053 on Build GRCh38
rs779077340 Chr.8: 142875126-142875127 on Build GRCh38
rs370528535 Chr.8: 142875059-142875059 on Build GRCh38
rs200770247 Chr.8: 142875127-142875127 on Build GRCh38
rs4541 Chr.8: 142875277-142875277 on Build GRCh38
rs73368712 Chr.8: 142874904-142874904 on Build GRCh38
rs779103938 Chr.8: 142875024-142875024 on Build GRCh38
rs369813641 Chr.8: 142875090-142875090 on Build GRCh38
rs368496071 Chr.8: 142874935-142874935 on Build GRCh38
rs769431602 Chr.8: 142875339-142875339 on Build GRCh38
rs756646001 Chr.8: 142875005-142875005 on Build GRCh38
rs765676068 Chr.8: 142875171-142875171 on Build GRCh38
rs767599353 Chr.8: 142874996-142874999 on Build GRCh38
rs374559854 Chr.8: 142875130-142875130 on Build GRCh38
rs532797257 Chr.8: 142875123-142875123 on Build GRCh38
rs5294 Chr.8: 142875040-142875040 on Build GRCh38
rs758714890 Chr.8: 142875253-142875254 on Build GRCh38
rs770737447 Chr.8: 142875251-142875251 on Build GRCh38
rs758052244 Chr.8: 142874931-142874931 on Build GRCh38
rs763645256 Chr.8: 142874913-142874923 on Build GRCh38
rs4998903 Chr.8: 142875083-142875083 on Build GRCh38
rs758341908 Chr.8: 142875238-142875238 on Build GRCh38
rs755128839 Chr.8: 142875063-142875063 on Build GRCh38
rs61752793 Chr.8: 142875353-142875353 on Build GRCh38
rs754941958 Chr.8: 142875148-142875148 on Build GRCh38
rs754432887 Chr.8: 142875075-142875075 on Build GRCh38
rs761186013 Chr.8: 142875108-142875108 on Build GRCh38
rs374421677 Chr.8: 142874934-142874934 on Build GRCh38
rs989804021 Chr.8: 142875230-142875231 on Build GRCh38
rs748447513 Chr.8: 142875025-142875025 on Build GRCh38
rs758458518 Chr.8: 142875140-142875140 on Build GRCh38
rs756568204 Chr.8: 142874941-142874941 on Build GRCh38
rs559331350 Chr.8: 142875173-142875173 on Build GRCh38
rs765770519 Chr.8: 142875128-142875128 on Build GRCh38
rs777160383 Chr.8: 142875242-142875242 on Build GRCh38
rs746458392 Chr.8: 142874940-142874940 on Build GRCh38
rs764598023 Chr.8: 142875283-142875283 on Build GRCh38
rs4998902 Chr.8: 142875089-142875089 on Build GRCh38
rs56715454 Chr.8: 142875360-142875360 on Build GRCh38
rs762402072 Chr.8: 142874967-142874967 on Build GRCh38
rs181764014 Chr.8: 142875094-142875094 on Build GRCh38
rs886062738 Chr.8: 142875150-142875150 on Build GRCh38
rs267606755 Chr.8: 142875086-142875086 on Build GRCh38
rs748092288 Chr.8: 142875151-142875151 on Build GRCh38
rs758271711 Chr.8: 142875312-142875312 on Build GRCh38
rs749310551 Chr.8: 142874944-142874944 on Build GRCh38

View Full Product Details