Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 142876074-142876562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP11B1
Entrez Gene ID
1584
Gene Name
cytochrome P450 family 11 subfamily B member 1
Gene Aliases
CPN1, CYP11B, FHI, P450C11
UniGene
Hs.184927
Gene Chromosome Location
Chr.8: 142872354-142879839 on Build GRCh38
Transcripts
NM_000497.3 NM_001026213.1 XM_011516874.2 XM_011516875.1 XM_017013146.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3951395 c.877C>T p.L293F Chr.8: 142876318-142876318 on Build GRCh38
749820 c.872C>T p.A291V Chr.8: 142876323-142876323 on Build GRCh38
6639010 c.842G>A p.S281N Chr.8: 142876353-142876353 on Build GRCh38
1489067 c.840C>T p.F280F Chr.8: 142876355-142876355 on Build GRCh38
6226726 c.920A>T p.N307I Chr.8: 142876275-142876275 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040219361 Chr.8: 142876233-142876233 on Build GRCh38
rs1007124772 Chr.8: 142876500-142876500 on Build GRCh38
rs763141139 Chr.8: 142876281-142876281 on Build GRCh38
rs779449583 Chr.8: 142876443-142876443 on Build GRCh38
rs781689489 Chr.8: 142876198-142876199 on Build GRCh38
rs1021808684 Chr.8: 142876497-142876497 on Build GRCh38
rs578061861 Chr.8: 142876398-142876398 on Build GRCh38
rs748180875 Chr.8: 142876393-142876393 on Build GRCh38
rs753598646 Chr.8: 142876291-142876291 on Build GRCh38
rs753601845 Chr.8: 142876348-142876348 on Build GRCh38
rs758902842 Chr.8: 142876093-142876093 on Build GRCh38
rs778295450 Chr.8: 142876247-142876247 on Build GRCh38
rs1035880476 Chr.8: 142876486-142876486 on Build GRCh38
rs553481635 Chr.8: 142876333-142876333 on Build GRCh38
rs779822554 Chr.8: 142876235-142876235 on Build GRCh38
rs387907572 Chr.8: 142876278-142876278 on Build GRCh38
rs372512957 Chr.8: 142876220-142876220 on Build GRCh38
rs557707841 Chr.8: 142876351-142876351 on Build GRCh38
rs62524999 Chr.8: 142876102-142876102 on Build GRCh38
rs115446538 Chr.8: 142876430-142876430 on Build GRCh38
rs887698889 Chr.8: 142876420-142876420 on Build GRCh38
rs530938850 Chr.8: 142876292-142876292 on Build GRCh38
rs751843934 Chr.8: 142876283-142876283 on Build GRCh38
rs985473407 Chr.8: 142876161-142876161 on Build GRCh38
rs104894061 Chr.8: 142876242-142876242 on Build GRCh38
rs370226163 Chr.8: 142876290-142876290 on Build GRCh38
rs372463434 Chr.8: 142876303-142876303 on Build GRCh38
rs387907573 Chr.8: 142876299-142876299 on Build GRCh38
rs770030737 Chr.8: 142876311-142876311 on Build GRCh38
rs749806219 Chr.8: 142876388-142876388 on Build GRCh38
rs777620711 Chr.8: 142876264-142876264 on Build GRCh38
rs528133024 Chr.8: 142876164-142876164 on Build GRCh38
rs981611979 Chr.8: 142876111-142876111 on Build GRCh38
rs775454138 Chr.8: 142876279-142876279 on Build GRCh38
rs375833424 Chr.8: 142876249-142876249 on Build GRCh38
rs1047606645 Chr.8: 142876411-142876411 on Build GRCh38
rs61752761 Chr.8: 142876488-142876488 on Build GRCh38
rs12674916 Chr.8: 142876542-142876542 on Build GRCh38
rs564209617 Chr.8: 142876154-142876154 on Build GRCh38
rs747773897 Chr.8: 142876206-142876206 on Build GRCh38
rs773739528 Chr.8: 142876217-142876217 on Build GRCh38
rs202091168 Chr.8: 142876296-142876296 on Build GRCh38
rs370713534 Chr.8: 142876408-142876408 on Build GRCh38
rs34570566 Chr.8: 142876322-142876322 on Build GRCh38
rs779910865 Chr.8: 142876155-142876155 on Build GRCh38
rs764253552 Chr.8: 142876229-142876229 on Build GRCh38
rs4535 Chr.8: 142876409-142876409 on Build GRCh38
rs751047685 Chr.8: 142876360-142876360 on Build GRCh38
rs536023008 Chr.8: 142876342-142876342 on Build GRCh38
rs6411 Chr.8: 142876232-142876232 on Build GRCh38
rs528362028 Chr.8: 142876412-142876412 on Build GRCh38
rs369207471 Chr.8: 142876401-142876401 on Build GRCh38
rs780468070 Chr.8: 142876426-142876426 on Build GRCh38
rs387907574 Chr.8: 142876267-142876267 on Build GRCh38
rs866826540 Chr.8: 142876128-142876128 on Build GRCh38
rs5292 Chr.8: 142876318-142876318 on Build GRCh38
rs775128501 Chr.8: 142876353-142876354 on Build GRCh38
rs752755752 Chr.8: 142876223-142876223 on Build GRCh38
rs1005898526 Chr.8: 142876274-142876274 on Build GRCh38
rs528479332 Chr.8: 142876515-142876515 on Build GRCh38
rs375892072 Chr.8: 142876305-142876305 on Build GRCh38
rs778864300 Chr.8: 142876295-142876295 on Build GRCh38
rs148707144 Chr.8: 142876265-142876265 on Build GRCh38
rs562813618 Chr.8: 142876478-142876478 on Build GRCh38
rs568705604 Chr.8: 142876334-142876334 on Build GRCh38
rs546225536 Chr.8: 142876312-142876312 on Build GRCh38
rs756992317 Chr.8: 142876367-142876367 on Build GRCh38
rs200994388 Chr.8: 142876302-142876302 on Build GRCh38
rs5290 Chr.8: 142876370-142876370 on Build GRCh38
rs191126800 Chr.8: 142876340-142876340 on Build GRCh38
rs956330489 Chr.8: 142876131-142876131 on Build GRCh38
rs61752762 Chr.8: 142876487-142876487 on Build GRCh38
rs746327005 Chr.8: 142876231-142876231 on Build GRCh38
rs10105643 Chr.8: 142876470-142876470 on Build GRCh38
rs748626226 Chr.8: 142876446-142876446 on Build GRCh38
rs756565917 Chr.8: 142876382-142876382 on Build GRCh38
rs546316273 Chr.8: 142876307-142876307 on Build GRCh38
rs550479477 Chr.8: 142876268-142876268 on Build GRCh38
rs763444205 Chr.8: 142876327-142876327 on Build GRCh38
rs759207560 Chr.8: 142876518-142876519 on Build GRCh38
rs996518742 Chr.8: 142876130-142876130 on Build GRCh38
rs769081498 Chr.8: 142876391-142876391 on Build GRCh38
rs147004517 Chr.8: 142876394-142876394 on Build GRCh38
rs141368413 Chr.8: 142876372-142876372 on Build GRCh38
rs918676158 Chr.8: 142876525-142876525 on Build GRCh38
rs199962349 Chr.8: 142876271-142876271 on Build GRCh38
rs372768323 Chr.8: 142876404-142876404 on Build GRCh38
rs770295234 Chr.8: 142876272-142876272 on Build GRCh38
rs773902049 Chr.8: 142876193-142876193 on Build GRCh38
rs1018211644 Chr.8: 142876506-142876506 on Build GRCh38
rs753774484 Chr.8: 142876241-142876241 on Build GRCh38
rs376065546 Chr.8: 142876407-142876407 on Build GRCh38
rs7822986 Chr.8: 142876440-142876440 on Build GRCh38
rs770651158 Chr.8: 142876304-142876304 on Build GRCh38
rs544670837 Chr.8: 142876350-142876350 on Build GRCh38
rs911309577 Chr.8: 142876194-142876194 on Build GRCh38
rs947423052 Chr.8: 142876160-142876160 on Build GRCh38
rs763711819 Chr.8: 142876358-142876358 on Build GRCh38
rs777316392 Chr.8: 142876219-142876219 on Build GRCh38
rs368239110 Chr.8: 142876218-142876218 on Build GRCh38
rs759428193 Chr.8: 142876349-142876349 on Build GRCh38
rs780265554 Chr.8: 142876383-142876383 on Build GRCh38
rs777419135 Chr.8: 142876298-142876298 on Build GRCh38
rs746628473 Chr.8: 142876163-142876163 on Build GRCh38
rs539928761 Chr.8: 142876214-142876214 on Build GRCh38
rs5291 Chr.8: 142876353-142876353 on Build GRCh38
rs200473174 Chr.8: 142876402-142876402 on Build GRCh38
rs914574742 Chr.8: 142876158-142876158 on Build GRCh38
rs369228207 Chr.8: 142876147-142876147 on Build GRCh38
rs1010821757 Chr.8: 142876496-142876496 on Build GRCh38
rs769310764 Chr.8: 142876288-142876288 on Build GRCh38
rs943182652 Chr.8: 142876198-142876198 on Build GRCh38
rs61752760 Chr.8: 142876498-142876498 on Build GRCh38
rs774441602 Chr.8: 142876392-142876392 on Build GRCh38
rs758580553 Chr.8: 142876227-142876227 on Build GRCh38
rs372647044 Chr.8: 142876250-142876250 on Build GRCh38

View Full Product Details