Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33253219-33253763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG1
Entrez Gene ID
573
Gene Name
BCL2 associated athanogene 1
Gene Aliases
BAG-1, HAP, RAP46
UniGene
Hs.377484
Gene Chromosome Location
Chr.9: 33252471-33264761 on Build GRCh38
Transcripts
NM_001172415.1 NM_004323.5

SNP Detail

SNP ID Chromosome Location
rs903261776 Chr.9: 33253252-33253252 on Build GRCh38
rs998927530 Chr.9: 33253541-33253541 on Build GRCh38
rs921757051 Chr.9: 33253593-33253593 on Build GRCh38
rs995146342 Chr.9: 33253280-33253280 on Build GRCh38
rs561657833 Chr.9: 33253442-33253442 on Build GRCh38
rs369142849 Chr.9: 33253564-33253564 on Build GRCh38
rs527248567 Chr.9: 33253443-33253443 on Build GRCh38
rs532920947 Chr.9: 33253549-33253549 on Build GRCh38
rs957290508 Chr.9: 33253307-33253307 on Build GRCh38
rs138680830 Chr.9: 33253465-33253465 on Build GRCh38
rs991373833 Chr.9: 33253319-33253319 on Build GRCh38
rs1040513378 Chr.9: 33253511-33253511 on Build GRCh38
rs971331628 Chr.9: 33253575-33253575 on Build GRCh38
rs188163329 Chr.9: 33253372-33253372 on Build GRCh38
rs973824056 Chr.9: 33253655-33253655 on Build GRCh38
rs961436043 Chr.9: 33253651-33253651 on Build GRCh38
rs1019488296 Chr.9: 33253245-33253245 on Build GRCh38
rs1009048635 Chr.9: 33253240-33253240 on Build GRCh38
rs973711228 Chr.9: 33253408-33253408 on Build GRCh38
rs706115 Chr.9: 33253607-33253607 on Build GRCh38
rs140069634 Chr.9: 33253675-33253675 on Build GRCh38
rs1004926499 Chr.9: 33253660-33253660 on Build GRCh38
rs919565716 Chr.9: 33253430-33253430 on Build GRCh38
rs578036647 Chr.9: 33253687-33253687 on Build GRCh38
rs752011130 Chr.9: 33253499-33253499 on Build GRCh38
rs1024027892 Chr.9: 33253373-33253373 on Build GRCh38
rs978073627 Chr.9: 33253548-33253548 on Build GRCh38
rs1001269373 Chr.9: 33253698-33253698 on Build GRCh38
rs534985939 Chr.9: 33253644-33253644 on Build GRCh38
rs769127390 Chr.9: 33253487-33253487 on Build GRCh38
rs570495278 Chr.9: 33253490-33253490 on Build GRCh38
rs556053159 Chr.9: 33253613-33253613 on Build GRCh38
rs908321350 Chr.9: 33253357-33253357 on Build GRCh38
rs1049409867 Chr.9: 33253630-33253630 on Build GRCh38
rs780586858 Chr.9: 33253488-33253488 on Build GRCh38
rs910803191 Chr.9: 33253483-33253483 on Build GRCh38
rs562575726 Chr.9: 33253327-33253327 on Build GRCh38
rs763857728 Chr.9: 33253638-33253638 on Build GRCh38
rs971148962 Chr.9: 33253396-33253396 on Build GRCh38
rs985086316 Chr.9: 33253691-33253691 on Build GRCh38
rs570203808 Chr.9: 33253565-33253565 on Build GRCh38
rs565972346 Chr.9: 33253445-33253445 on Build GRCh38
rs1015387586 Chr.9: 33253612-33253612 on Build GRCh38
rs541763213 Chr.9: 33253431-33253431 on Build GRCh38
rs915536651 Chr.9: 33253492-33253492 on Build GRCh38
rs981295975 Chr.9: 33253332-33253332 on Build GRCh38
rs762651796 Chr.9: 33253615-33253615 on Build GRCh38
rs191119067 Chr.9: 33253625-33253625 on Build GRCh38
rs150332360 Chr.9: 33253690-33253690 on Build GRCh38
rs886101415 Chr.9: 33253369-33253369 on Build GRCh38
rs911949992 Chr.9: 33253550-33253550 on Build GRCh38
rs903270815 Chr.9: 33253522-33253522 on Build GRCh38
rs1045442899 Chr.9: 33253678-33253678 on Build GRCh38
rs1054471288 Chr.9: 33253543-33253543 on Build GRCh38
rs1022873350 Chr.9: 33253326-33253326 on Build GRCh38
rs1032974304 Chr.9: 33253290-33253290 on Build GRCh38
rs999271678 Chr.9: 33253286-33253286 on Build GRCh38
rs910805790 Chr.9: 33253728-33253728 on Build GRCh38

View Full Product Details