Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34338618-34339115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDT2
Entrez Gene ID
318
Gene Name
nudix hydrolase 2
Gene Aliases
APAH1
UniGene
Hs.493767
Gene Chromosome Location
Chr.9: 34329506-34343713 on Build GRCh38
Transcripts
NM_001161.4 NM_001244390.1 NM_147172.2 NM_147173.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4488908 c.33C>T p.F11F Chr.9: 34339072-34339072 on Build GRCh38
5876153 c.58G>T p.D20Y Chr.9: 34339097-34339097 on Build GRCh38
1108442 c.35G>A p.R12Q Chr.9: 34339074-34339074 on Build GRCh38
3906999 c.4G>A p.A2T Chr.9: 34339043-34339043 on Build GRCh38
6193880 c.28A>T p.I10F Chr.9: 34339067-34339067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769837247 Chr.9: 34339074-34339074 on Build GRCh38
rs763844219 Chr.9: 34339084-34339084 on Build GRCh38
rs771240044 Chr.9: 34339055-34339055 on Build GRCh38
rs1036339269 Chr.9: 34338763-34338763 on Build GRCh38
rs1033994800 Chr.9: 34338669-34338669 on Build GRCh38
rs933513308 Chr.9: 34338815-34338815 on Build GRCh38
rs901873711 Chr.9: 34338665-34338665 on Build GRCh38
rs886528360 Chr.9: 34338698-34338699 on Build GRCh38
rs146917126 Chr.9: 34338649-34338649 on Build GRCh38
rs147053559 Chr.9: 34338767-34338767 on Build GRCh38
rs922149553 Chr.9: 34338804-34338804 on Build GRCh38
rs1013502224 Chr.9: 34338947-34338947 on Build GRCh38
rs1802826 Chr.9: 34338730-34338730 on Build GRCh38
rs148118852 Chr.9: 34339046-34339046 on Build GRCh38
rs2275263 Chr.9: 34338816-34338816 on Build GRCh38
rs141762241 Chr.9: 34338720-34338720 on Build GRCh38
rs895020101 Chr.9: 34338946-34338946 on Build GRCh38
rs1002101133 Chr.9: 34338966-34338966 on Build GRCh38
rs148119952 Chr.9: 34339073-34339073 on Build GRCh38
rs988003900 Chr.9: 34338830-34338830 on Build GRCh38
rs774090026 Chr.9: 34339085-34339085 on Build GRCh38
rs1010454099 Chr.9: 34338999-34338999 on Build GRCh38
rs762784885 Chr.9: 34339081-34339081 on Build GRCh38
rs745748055 Chr.9: 34339007-34339007 on Build GRCh38
rs749851057 Chr.9: 34338999-34338999 on Build GRCh38
rs775276310 Chr.9: 34339076-34339076 on Build GRCh38
rs80275122 Chr.9: 34338777-34338777 on Build GRCh38
rs963424862 Chr.9: 34339025-34339025 on Build GRCh38
rs1042107855 Chr.9: 34338850-34338850 on Build GRCh38
rs867803849 Chr.9: 34339053-34339053 on Build GRCh38
rs775831901 Chr.9: 34338987-34338987 on Build GRCh38
rs966438524 Chr.9: 34338805-34338805 on Build GRCh38
rs76223630 Chr.9: 34338920-34338920 on Build GRCh38
rs187418807 Chr.9: 34338662-34338662 on Build GRCh38
rs913529300 Chr.9: 34338779-34338779 on Build GRCh38
rs368433197 Chr.9: 34338817-34338817 on Build GRCh38
rs1006276540 Chr.9: 34338706-34338706 on Build GRCh38
rs975041063 Chr.9: 34338799-34338799 on Build GRCh38
rs35394154 Chr.9: 34338998-34338999 on Build GRCh38
rs758653286 Chr.9: 34338648-34338648 on Build GRCh38
rs751376883 Chr.9: 34338661-34338661 on Build GRCh38
rs1037772783 Chr.9: 34338935-34338935 on Build GRCh38
rs540116916 Chr.9: 34339044-34339044 on Build GRCh38
rs757274696 Chr.9: 34338716-34338716 on Build GRCh38
rs997920193 Chr.9: 34338688-34338688 on Build GRCh38
rs538190975 Chr.9: 34338857-34338857 on Build GRCh38
rs913398940 Chr.9: 34338839-34338839 on Build GRCh38
rs979119926 Chr.9: 34338817-34338818 on Build GRCh38
rs983529725 Chr.9: 34338742-34338742 on Build GRCh38
rs778192361 Chr.9: 34339036-34339036 on Build GRCh38
rs1048204326 Chr.9: 34338886-34338886 on Build GRCh38
rs780709713 Chr.9: 34338997-34338997 on Build GRCh38
rs370422239 Chr.9: 34339018-34339018 on Build GRCh38
rs139196212 Chr.9: 34338685-34338685 on Build GRCh38
rs566192163 Chr.9: 34338710-34338710 on Build GRCh38
rs945161912 Chr.9: 34338846-34338846 on Build GRCh38
rs933215575 Chr.9: 34338882-34338882 on Build GRCh38
rs1024849847 Chr.9: 34338961-34338961 on Build GRCh38
rs906057794 Chr.9: 34338951-34338951 on Build GRCh38
rs371496158 Chr.9: 34338988-34338988 on Build GRCh38
rs772554957 Chr.9: 34339032-34339032 on Build GRCh38
rs1026440385 Chr.9: 34338697-34338697 on Build GRCh38
rs943187395 Chr.9: 34338841-34338841 on Build GRCh38
rs746674514 Chr.9: 34339015-34339017 on Build GRCh38
rs776795302 Chr.9: 34339067-34339067 on Build GRCh38
rs116357346 Chr.9: 34338895-34338895 on Build GRCh38

View Full Product Details