Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34520953-34521448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAI1
Entrez Gene ID
27019
Gene Name
dynein axonemal intermediate chain 1
Gene Aliases
CILD1, DIC1, ICS1, PCD
UniGene
Hs.112667
Gene Chromosome Location
Chr.9: 34458752-34520989 on Build GRCh38
Transcripts
NM_001281428.1 NM_012144.3 XM_006716758.3 XM_011517848.1 XM_017014625.1 NM_198573.2

Gene Symbol
ENHO
Entrez Gene ID
375704
Gene Name
energy homeostasis associated
Gene Aliases
C9orf165, UNQ470
UniGene
Hs.522085
Gene Chromosome Location
Chr.9: 34521042-34523039 on Build GRCh38
Transcripts
NM_001281428.1 NM_012144.3 XM_006716758.3 XM_011517848.1 XM_017014625.1 NM_198573.2

SNP Detail

SNP ID Chromosome Location
rs956312899 Chr.9: 34521181-34521181 on Build GRCh38
rs532006177 Chr.9: 34521300-34521300 on Build GRCh38
rs72735260 Chr.9: 34521190-34521190 on Build GRCh38
rs186235852 Chr.9: 34521151-34521151 on Build GRCh38
rs113770141 Chr.9: 34521014-34521014 on Build GRCh38
rs1055225673 Chr.9: 34521143-34521143 on Build GRCh38
rs1024887479 Chr.9: 34521187-34521187 on Build GRCh38
rs765234440 Chr.9: 34521401-34521402 on Build GRCh38
rs975538635 Chr.9: 34521242-34521242 on Build GRCh38
rs374071200 Chr.9: 34521227-34521227 on Build GRCh38
rs913443319 Chr.9: 34520988-34520988 on Build GRCh38
rs921241649 Chr.9: 34521276-34521276 on Build GRCh38
rs561916315 Chr.9: 34521134-34521134 on Build GRCh38
rs1055838117 Chr.9: 34521000-34521000 on Build GRCh38
rs570484051 Chr.9: 34521212-34521212 on Build GRCh38
rs769475832 Chr.9: 34521378-34521378 on Build GRCh38
rs563654142 Chr.9: 34521002-34521002 on Build GRCh38
rs878873066 Chr.9: 34521304-34521304 on Build GRCh38
rs770833093 Chr.9: 34521346-34521346 on Build GRCh38
rs761086839 Chr.9: 34521412-34521412 on Build GRCh38
rs760547330 Chr.9: 34521314-34521315 on Build GRCh38
rs913495886 Chr.9: 34521312-34521312 on Build GRCh38
rs778936646 Chr.9: 34521257-34521257 on Build GRCh38
rs774883423 Chr.9: 34521388-34521388 on Build GRCh38
rs994498031 Chr.9: 34520999-34520999 on Build GRCh38
rs755174552 Chr.9: 34521428-34521428 on Build GRCh38
rs1000599024 Chr.9: 34521114-34521114 on Build GRCh38
rs1016075106 Chr.9: 34521218-34521218 on Build GRCh38
rs183306584 Chr.9: 34521015-34521015 on Build GRCh38
rs955406066 Chr.9: 34521298-34521298 on Build GRCh38
rs548523968 Chr.9: 34521084-34521084 on Build GRCh38
rs555651543 Chr.9: 34521358-34521358 on Build GRCh38
rs950578405 Chr.9: 34521188-34521188 on Build GRCh38
rs1024184179 Chr.9: 34521036-34521036 on Build GRCh38
rs761619222 Chr.9: 34521306-34521306 on Build GRCh38
rs1111375 Chr.9: 34521150-34521150 on Build GRCh38
rs749438308 Chr.9: 34521175-34521175 on Build GRCh38
rs987718169 Chr.9: 34521309-34521309 on Build GRCh38
rs551721224 Chr.9: 34521287-34521287 on Build GRCh38
rs898936382 Chr.9: 34520985-34520985 on Build GRCh38
rs766730004 Chr.9: 34521419-34521419 on Build GRCh38
rs773711872 Chr.9: 34521404-34521404 on Build GRCh38
rs746789461 Chr.9: 34521336-34521336 on Build GRCh38
rs190606607 Chr.9: 34521198-34521198 on Build GRCh38
rs936439422 Chr.9: 34521409-34521409 on Build GRCh38
rs746083874 Chr.9: 34521001-34521001 on Build GRCh38
rs777696216 Chr.9: 34521335-34521335 on Build GRCh38
rs754104019 Chr.9: 34521422-34521422 on Build GRCh38
rs569333934 Chr.9: 34521299-34521299 on Build GRCh38
rs76043341 Chr.9: 34521238-34521238 on Build GRCh38
rs767915634 Chr.9: 34521399-34521399 on Build GRCh38
rs769855531 Chr.9: 34521044-34521044 on Build GRCh38
rs566352810 Chr.9: 34521359-34521359 on Build GRCh38
rs998684819 Chr.9: 34521152-34521152 on Build GRCh38
rs903605718 Chr.9: 34521112-34521112 on Build GRCh38
rs150406670 Chr.9: 34521301-34521301 on Build GRCh38

View Full Product Details