Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15407428-15407941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIR
Entrez Gene ID
8544
Gene Name
pirin
Gene Aliases
-
UniGene
Hs.495728
Gene Chromosome Location
Chr.X: 15384799-15493588 on Build GRCh38
Transcripts
NM_001018109.2 NM_003662.3 NR_037859.2

Gene Symbol
PIR-FIGF
Entrez Gene ID
100532742
Gene Name
PIR-FIGF readthrough
Gene Aliases
-
UniGene
Hs.495728
Gene Chromosome Location
Chr.X: 15345591-15491309 on Build GRCh38
Transcripts
NM_001018109.2 NM_003662.3 NR_037859.2

SNP Detail

SNP ID Chromosome Location
rs60378841 Chr.X: 15407791-15407791 on Build GRCh38
rs751639462 Chr.X: 15407503-15407503 on Build GRCh38
rs41311829 Chr.X: 15407633-15407633 on Build GRCh38
rs772848756 Chr.X: 15407647-15407647 on Build GRCh38
rs964634172 Chr.X: 15407671-15407674 on Build GRCh38
rs766509434 Chr.X: 15407501-15407501 on Build GRCh38
rs923115646 Chr.X: 15407894-15407894 on Build GRCh38
rs774494117 Chr.X: 15407615-15407615 on Build GRCh38
rs745385130 Chr.X: 15407588-15407588 on Build GRCh38
rs761722625 Chr.X: 15407480-15407480 on Build GRCh38
rs746273316 Chr.X: 15407856-15407859 on Build GRCh38
rs754988948 Chr.X: 15407517-15407517 on Build GRCh38
rs60953218 Chr.X: 15407581-15407581 on Build GRCh38
rs779495488 Chr.X: 15407586-15407586 on Build GRCh38
rs984384977 Chr.X: 15407629-15407629 on Build GRCh38
rs763139509 Chr.X: 15407491-15407491 on Build GRCh38
rs761614438 Chr.X: 15407642-15407643 on Build GRCh38
rs763759399 Chr.X: 15407465-15407465 on Build GRCh38
rs529697458 Chr.X: 15407474-15407474 on Build GRCh38
rs898914352 Chr.X: 15407877-15407877 on Build GRCh38
rs140893586 Chr.X: 15407646-15407646 on Build GRCh38
rs34107232 Chr.X: 15407799-15407800 on Build GRCh38
rs112327224 Chr.X: 15407739-15407739 on Build GRCh38
rs1037818396 Chr.X: 15407836-15407836 on Build GRCh38
rs754455361 Chr.X: 15407556-15407556 on Build GRCh38
rs200350682 Chr.X: 15407529-15407529 on Build GRCh38
rs776673863 Chr.X: 15407476-15407476 on Build GRCh38
rs767802108 Chr.X: 15407524-15407524 on Build GRCh38
rs777976196 Chr.X: 15407543-15407543 on Build GRCh38
rs934618113 Chr.X: 15407475-15407475 on Build GRCh38
rs373415087 Chr.X: 15407913-15407913 on Build GRCh38
rs747067533 Chr.X: 15407718-15407718 on Build GRCh38
rs1026057163 Chr.X: 15407604-15407604 on Build GRCh38
rs181641471 Chr.X: 15407820-15407820 on Build GRCh38
rs752822548 Chr.X: 15407528-15407528 on Build GRCh38
rs752704052 Chr.X: 15407515-15407515 on Build GRCh38
rs993155542 Chr.X: 15407890-15407890 on Build GRCh38

View Full Product Details