Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 24310717-24311211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUPT20HL2
Entrez Gene ID
170067
Gene Name
SPT20 homolog, SAGA complex component-like 2
Gene Aliases
FAM48B1, FAM48B2
UniGene
Hs.716893
Gene Chromosome Location
Chr.X: 24310862-24313315 on Build GRCh38
Transcripts
NM_001136233.1

SNP Detail

SNP ID Chromosome Location
rs758396332 Chr.X: 24310888-24310888 on Build GRCh38
rs368733626 Chr.X: 24311055-24311055 on Build GRCh38
rs765411333 Chr.X: 24311175-24311175 on Build GRCh38
rs759533555 Chr.X: 24311141-24311141 on Build GRCh38
rs1035715207 Chr.X: 24310743-24310743 on Build GRCh38
rs781156706 Chr.X: 24311041-24311043 on Build GRCh38
rs745923718 Chr.X: 24311040-24311041 on Build GRCh38
rs1043875890 Chr.X: 24311165-24311165 on Build GRCh38
rs753390639 Chr.X: 24310814-24310814 on Build GRCh38
rs780322229 Chr.X: 24311188-24311188 on Build GRCh38
rs772880244 Chr.X: 24311124-24311124 on Build GRCh38
rs751447245 Chr.X: 24311011-24311011 on Build GRCh38
rs746355786 Chr.X: 24311020-24311020 on Build GRCh38
rs748533546 Chr.X: 24311102-24311102 on Build GRCh38
rs912432864 Chr.X: 24311053-24311053 on Build GRCh38
rs370240670 Chr.X: 24311028-24311028 on Build GRCh38
rs934596561 Chr.X: 24311062-24311062 on Build GRCh38
rs752444377 Chr.X: 24311172-24311172 on Build GRCh38
rs771157603 Chr.X: 24310951-24310951 on Build GRCh38
rs769335945 Chr.X: 24311103-24311103 on Build GRCh38
rs778931051 Chr.X: 24310954-24310954 on Build GRCh38
rs930355872 Chr.X: 24311073-24311073 on Build GRCh38
rs776076322 Chr.X: 24310964-24310964 on Build GRCh38
rs780846657 Chr.X: 24310780-24310780 on Build GRCh38
rs772724290 Chr.X: 24310958-24310958 on Build GRCh38
rs779909988 Chr.X: 24310900-24310900 on Build GRCh38
rs751361194 Chr.X: 24310902-24310902 on Build GRCh38
rs138896785 Chr.X: 24310800-24310800 on Build GRCh38
rs761406472 Chr.X: 24310816-24310816 on Build GRCh38
rs766333160 Chr.X: 24311006-24311006 on Build GRCh38
rs1004300572 Chr.X: 24310859-24310859 on Build GRCh38
rs764939761 Chr.X: 24311000-24311000 on Build GRCh38
rs750628206 Chr.X: 24311183-24311183 on Build GRCh38
rs745943572 Chr.X: 24311032-24311032 on Build GRCh38
rs774784150 Chr.X: 24311018-24311018 on Build GRCh38
rs374184071 Chr.X: 24311054-24311054 on Build GRCh38
rs776927632 Chr.X: 24311084-24311084 on Build GRCh38
rs962406951 Chr.X: 24310921-24310921 on Build GRCh38
rs758515349 Chr.X: 24311042-24311042 on Build GRCh38
rs753868783 Chr.X: 24311150-24311150 on Build GRCh38
rs765921384 Chr.X: 24311128-24311128 on Build GRCh38
rs772611443 Chr.X: 24311002-24311002 on Build GRCh38
rs201018804 Chr.X: 24310997-24310997 on Build GRCh38
rs540022132 Chr.X: 24311170-24311170 on Build GRCh38
rs758673663 Chr.X: 24311186-24311186 on Build GRCh38
rs867424848 Chr.X: 24311089-24311089 on Build GRCh38
rs762381583 Chr.X: 24311003-24311003 on Build GRCh38
rs896743920 Chr.X: 24310783-24310783 on Build GRCh38
rs772673703 Chr.X: 24311030-24311030 on Build GRCh38
rs866584666 Chr.X: 24311051-24311051 on Build GRCh38
rs773816760 Chr.X: 24311140-24311140 on Build GRCh38
rs747200265 Chr.X: 24311034-24311057 on Build GRCh38
rs761413178 Chr.X: 24310998-24310998 on Build GRCh38
rs749524991 Chr.X: 24310941-24310941 on Build GRCh38
rs749917407 Chr.X: 24310870-24310870 on Build GRCh38
rs372315919 Chr.X: 24311057-24311057 on Build GRCh38
rs777995858 Chr.X: 24310931-24310931 on Build GRCh38
rs771131192 Chr.X: 24311037-24311038 on Build GRCh38
rs754730029 Chr.X: 24310929-24310929 on Build GRCh38
rs367865263 Chr.X: 24310868-24310868 on Build GRCh38
rs565076395 Chr.X: 24310810-24310810 on Build GRCh38
rs755875032 Chr.X: 24311174-24311174 on Build GRCh38
rs747411372 Chr.X: 24310987-24310987 on Build GRCh38

View Full Product Details