Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47144779-47145275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFB11
Entrez Gene ID
54539
Gene Name
NADH:ubiquinone oxidoreductase subunit B11
Gene Aliases
CI-ESSS, ESSS, LSDMCA3, NP17.3, Np15, P17.3
UniGene
Hs.521969
Gene Chromosome Location
Chr.X: 47142216-47145210 on Build GRCh38
Transcripts
NM_001135998.2 NM_019056.6 NM_001204466.1 NM_001204467.1 NM_001204468.1 NM_005676.4 NM_152856.2 XM_005272677.4 XM_017029884.1

Gene Symbol
RBM10
Entrez Gene ID
8241
Gene Name
RNA binding motif protein 10
Gene Aliases
DXS8237E, GPATC9, GPATCH9, S1-1, TARPS, ZRANB5
UniGene
Hs.401509
Gene Chromosome Location
Chr.X: 47145196-47186815 on Build GRCh38
Transcripts
NM_001135998.2 NM_019056.6 NM_001204466.1 NM_001204467.1 NM_001204468.1 NM_005676.4 NM_152856.2 XM_005272677.4 XM_017029884.1

SNP Detail

SNP ID Chromosome Location
rs915630002 Chr.X: 47144873-47144873 on Build GRCh38
rs183970296 Chr.X: 47144814-47144814 on Build GRCh38
rs782567288 Chr.X: 47144911-47144911 on Build GRCh38
rs782234203 Chr.X: 47145088-47145088 on Build GRCh38
rs782425981 Chr.X: 47145205-47145205 on Build GRCh38
rs190515179 Chr.X: 47145143-47145143 on Build GRCh38
rs902881813 Chr.X: 47144926-47144926 on Build GRCh38
rs990995623 Chr.X: 47145238-47145238 on Build GRCh38
rs982120881 Chr.X: 47145171-47145171 on Build GRCh38
rs890388649 Chr.X: 47144946-47144946 on Build GRCh38
rs1026299551 Chr.X: 47145091-47145091 on Build GRCh38
rs928103746 Chr.X: 47144836-47144836 on Build GRCh38
rs782685207 Chr.X: 47145062-47145062 on Build GRCh38
rs915435549 Chr.X: 47145243-47145243 on Build GRCh38
rs782472450 Chr.X: 47144954-47144954 on Build GRCh38
rs998717491 Chr.X: 47144928-47144928 on Build GRCh38
rs182036815 Chr.X: 47145007-47145007 on Build GRCh38
rs938164170 Chr.X: 47144845-47144845 on Build GRCh38
rs928177383 Chr.X: 47145183-47145183 on Build GRCh38
rs1017860542 Chr.X: 47144974-47144974 on Build GRCh38
rs947140029 Chr.X: 47145246-47145246 on Build GRCh38
rs188262684 Chr.X: 47144932-47144932 on Build GRCh38
rs947158759 Chr.X: 47144909-47144909 on Build GRCh38
rs782647790 Chr.X: 47144970-47144970 on Build GRCh38
rs184835888 Chr.X: 47145018-47145018 on Build GRCh38

View Full Product Details