Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101897790-101898302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZMAT1
Entrez Gene ID
84460
Gene Name
zinc finger matrin-type 1
Gene Aliases
-
UniGene
Hs.496512
Gene Chromosome Location
Chr.X: 101882288-101932135 on Build GRCh38
Transcripts
NM_001011657.3 NM_001282400.1 NM_001282401.1 XM_005262212.4 XM_005262213.4 XM_005262214.3 XM_005262216.4 XM_017029903.1 XM_017029904.1

SNP Detail

SNP ID Chromosome Location
rs752686074 Chr.X: 101897885-101897885 on Build GRCh38
rs774920916 Chr.X: 101898003-101898003 on Build GRCh38
rs975245036 Chr.X: 101898072-101898072 on Build GRCh38
rs763989703 Chr.X: 101898086-101898086 on Build GRCh38
rs140421354 Chr.X: 101898164-101898164 on Build GRCh38
rs763510075 Chr.X: 101897825-101897825 on Build GRCh38
rs201939211 Chr.X: 101898197-101898197 on Build GRCh38
rs370545442 Chr.X: 101898233-101898233 on Build GRCh38
rs759295308 Chr.X: 101898216-101898216 on Build GRCh38
rs761584692 Chr.X: 101898243-101898243 on Build GRCh38
rs946400932 Chr.X: 101898223-101898224 on Build GRCh38
rs781450942 Chr.X: 101897936-101897936 on Build GRCh38
rs143644365 Chr.X: 101898199-101898199 on Build GRCh38
rs752193964 Chr.X: 101898076-101898076 on Build GRCh38
rs769324501 Chr.X: 101898156-101898156 on Build GRCh38
rs776379207 Chr.X: 101898032-101898032 on Build GRCh38
rs999943136 Chr.X: 101898017-101898017 on Build GRCh38
rs141058940 Chr.X: 101898007-101898007 on Build GRCh38
rs189207863 Chr.X: 101898096-101898096 on Build GRCh38
rs762949047 Chr.X: 101898083-101898083 on Build GRCh38
rs779928068 Chr.X: 101897987-101897987 on Build GRCh38
rs769943813 Chr.X: 101897962-101897962 on Build GRCh38
rs373508177 Chr.X: 101898234-101898234 on Build GRCh38
rs762092655 Chr.X: 101897906-101897906 on Build GRCh38
rs749825296 Chr.X: 101897994-101897994 on Build GRCh38
rs1009755189 Chr.X: 101897920-101897920 on Build GRCh38
rs746026620 Chr.X: 101897939-101897939 on Build GRCh38
rs764554559 Chr.X: 101897832-101897832 on Build GRCh38
rs775023613 Chr.X: 101898220-101898220 on Build GRCh38
rs372762382 Chr.X: 101897852-101897852 on Build GRCh38
rs757094572 Chr.X: 101898120-101898120 on Build GRCh38
rs764071851 Chr.X: 101898239-101898239 on Build GRCh38
rs374727904 Chr.X: 101897820-101897820 on Build GRCh38
rs767354453 Chr.X: 101898245-101898245 on Build GRCh38
rs756229077 Chr.X: 101898143-101898143 on Build GRCh38
rs759044077 Chr.X: 101898042-101898042 on Build GRCh38
rs749374346 Chr.X: 101898151-101898151 on Build GRCh38
rs144779698 Chr.X: 101898021-101898021 on Build GRCh38
rs184829669 Chr.X: 101897849-101897849 on Build GRCh38
rs1030012426 Chr.X: 101898080-101898080 on Build GRCh38
rs750171320 Chr.X: 101898246-101898246 on Build GRCh38
rs377126814 Chr.X: 101897854-101897854 on Build GRCh38
rs748627441 Chr.X: 101898196-101898196 on Build GRCh38
rs776083570 Chr.X: 101897823-101897823 on Build GRCh38
rs982683258 Chr.X: 101898167-101898167 on Build GRCh38
rs776644966 Chr.X: 101897818-101897818 on Build GRCh38
rs756424747 Chr.X: 101898258-101898258 on Build GRCh38
rs146221291 Chr.X: 101897929-101897929 on Build GRCh38
rs766676141 Chr.X: 101898267-101898267 on Build GRCh38
rs144995159 Chr.X: 101898000-101898000 on Build GRCh38
rs780807122 Chr.X: 101898127-101898127 on Build GRCh38
rs751408064 Chr.X: 101898101-101898101 on Build GRCh38
rs1026172762 Chr.X: 101897883-101897883 on Build GRCh38
rs745931910 Chr.X: 101897819-101897819 on Build GRCh38
rs974572009 Chr.X: 101898247-101898247 on Build GRCh38
rs769331880 Chr.X: 101898218-101898218 on Build GRCh38
rs760460789 Chr.X: 101897838-101897838 on Build GRCh38
rs764612970 Chr.X: 101897862-101897862 on Build GRCh38
rs369050619 Chr.X: 101897830-101897830 on Build GRCh38
rs764817398 Chr.X: 101898064-101898064 on Build GRCh38
rs780237290 Chr.X: 101898144-101898144 on Build GRCh38
rs777607161 Chr.X: 101897916-101897916 on Build GRCh38
rs146743965 Chr.X: 101898139-101898139 on Build GRCh38

View Full Product Details