Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 135286355-135286853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF75D
Entrez Gene ID
7626
Gene Name
zinc finger protein 75D
Gene Aliases
D8C6, ZKSCAN24, ZNF75, ZNF82, ZSCAN28
UniGene
Hs.371717
Gene Chromosome Location
Chr.X: 135248589-135344109 on Build GRCh38
Transcripts
NM_001185063.2 NM_007131.4 NR_110381.1 XM_005262469.1 XM_005262471.3 XM_011531391.1 XM_011531395.2 XM_011531396.2 XM_011531397.2 XM_017029818.1 XM_017029819.1 XM_017029820.1

SNP Detail

SNP ID Chromosome Location
rs78687584 Chr.X: 135286693-135286693 on Build GRCh38
rs191481645 Chr.X: 135286505-135286505 on Build GRCh38
rs188684966 Chr.X: 135286770-135286770 on Build GRCh38
rs782085233 Chr.X: 135286810-135286810 on Build GRCh38
rs1129095 Chr.X: 135286670-135286670 on Build GRCh38
rs7358020 Chr.X: 135286562-135286562 on Build GRCh38
rs7358017 Chr.X: 135286486-135286486 on Build GRCh38
rs782477002 Chr.X: 135286812-135286812 on Build GRCh38
rs144595751 Chr.X: 135286820-135286820 on Build GRCh38
rs183676952 Chr.X: 135286554-135286554 on Build GRCh38
rs782584498 Chr.X: 135286649-135286649 on Build GRCh38
rs374641046 Chr.X: 135286528-135286528 on Build GRCh38
rs181111937 Chr.X: 135286790-135286790 on Build GRCh38
rs782433679 Chr.X: 135286475-135286475 on Build GRCh38
rs782336671 Chr.X: 135286772-135286772 on Build GRCh38
rs782389656 Chr.X: 135286510-135286510 on Build GRCh38

View Full Product Details