Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 147925453-147925953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMR1
Entrez Gene ID
2332
Gene Name
fragile X mental retardation 1
Gene Aliases
FMRP, FRAXA, POF, POF1
UniGene
Hs.103183
Gene Chromosome Location
Chr.X: 147911951-147951127 on Build GRCh38
Transcripts
NM_001185075.1 NM_001185076.1 NM_001185081.1 NM_001185082.1 NM_002024.5 NR_033699.1 NR_033700.1

SNP Detail

SNP ID Chromosome Location
rs781832896 Chr.X: 147925663-147925663 on Build GRCh38
rs782202548 Chr.X: 147925623-147925623 on Build GRCh38
rs782393400 Chr.X: 147925633-147925633 on Build GRCh38
rs781790587 Chr.X: 147925497-147925497 on Build GRCh38
rs781784498 Chr.X: 147925637-147925637 on Build GRCh38
rs183745963 Chr.X: 147925532-147925532 on Build GRCh38
rs200455877 Chr.X: 147925642-147925642 on Build GRCh38
rs782451556 Chr.X: 147925642-147925643 on Build GRCh38
rs782696616 Chr.X: 147925630-147925630 on Build GRCh38
rs782233272 Chr.X: 147925637-147925637 on Build GRCh38
rs782791326 Chr.X: 147925502-147925502 on Build GRCh38
rs782142501 Chr.X: 147925524-147925525 on Build GRCh38
rs186344906 Chr.X: 147925913-147925913 on Build GRCh38
rs3737554 Chr.X: 147925650-147925650 on Build GRCh38
rs782639149 Chr.X: 147925634-147925634 on Build GRCh38
rs782551523 Chr.X: 147925500-147925500 on Build GRCh38
rs782438699 Chr.X: 147925628-147925628 on Build GRCh38
rs25731 Chr.X: 147925702-147925702 on Build GRCh38
rs782284934 Chr.X: 147925632-147925632 on Build GRCh38
rs924092966 Chr.X: 147925507-147925507 on Build GRCh38
rs781999118 Chr.X: 147925491-147925491 on Build GRCh38
rs964061264 Chr.X: 147925863-147925863 on Build GRCh38
rs782130500 Chr.X: 147925651-147925651 on Build GRCh38
rs782516655 Chr.X: 147925872-147925872 on Build GRCh38
rs1038876282 Chr.X: 147925706-147925706 on Build GRCh38
rs781810881 Chr.X: 147925694-147925694 on Build GRCh38
rs1018112097 Chr.X: 147925856-147925856 on Build GRCh38
rs782073569 Chr.X: 147925678-147925678 on Build GRCh38
rs376206337 Chr.X: 147925761-147925761 on Build GRCh38
rs781935092 Chr.X: 147925638-147925638 on Build GRCh38
rs370314343 Chr.X: 147925857-147925857 on Build GRCh38
rs955203203 Chr.X: 147925809-147925809 on Build GRCh38
rs782602321 Chr.X: 147925592-147925592 on Build GRCh38
rs781866021 Chr.X: 147925522-147925522 on Build GRCh38
rs376676904 Chr.X: 147925514-147925514 on Build GRCh38
rs782293684 Chr.X: 147925903-147925903 on Build GRCh38
rs372981558 Chr.X: 147925643-147925643 on Build GRCh38
rs181856740 Chr.X: 147925876-147925876 on Build GRCh38
rs1008099883 Chr.X: 147925822-147925822 on Build GRCh38
rs113627441 Chr.X: 147925880-147925880 on Build GRCh38
rs188414110 Chr.X: 147925803-147925803 on Build GRCh38
rs375330839 Chr.X: 147925658-147925658 on Build GRCh38
rs73606778 Chr.X: 147925911-147925911 on Build GRCh38
rs782720052 Chr.X: 147925683-147925683 on Build GRCh38
rs994643830 Chr.X: 147925778-147925778 on Build GRCh38
rs895152089 Chr.X: 147925692-147925692 on Build GRCh38
rs898557968 Chr.X: 147925760-147925760 on Build GRCh38
rs782617656 Chr.X: 147925727-147925727 on Build GRCh38

View Full Product Details