Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186312567-186313099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRG4
Entrez Gene ID
10216
Gene Name
proteoglycan 4
Gene Aliases
CACP, HAPO, JCAP, MSF, SZP
UniGene
Hs.647723
Gene Chromosome Location
Chr.1: 186296273-186314562 on Build GRCh38
Transcripts
NM_001127708.2 NM_001127709.2 NM_001127710.2 NM_001303232.1 NM_005807.4 XM_017000002.1 XM_017000003.1 NM_003292.2

Gene Symbol
TPR
Entrez Gene ID
7175
Gene Name
translocated promoter region, nuclear basket protein
Gene Aliases
-
UniGene
Hs.279640
Gene Chromosome Location
Chr.1: 186311652-186375325 on Build GRCh38
Transcripts
NM_001127708.2 NM_001127709.2 NM_001127710.2 NM_001303232.1 NM_005807.4 XM_017000002.1 XM_017000003.1 NM_003292.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
229437 c.4039C>T p.P1347S Chr.1: 186312816-186312816 on Build GRCh38
1263014 c.4090G>C p.G1364R Chr.1: 186312867-186312867 on Build GRCh38
1689431 c.4029G>A p.W1343* Chr.1: 186312806-186312806 on Build GRCh38
4026142 c.4051A>G p.T1351A Chr.1: 186312828-186312828 on Build GRCh38
7188616 c.4117+2T>C p.? Chr.1: 186312896-186312896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141044991 Chr.1: 186312785-186312785 on Build GRCh38
rs544807583 Chr.1: 186313074-186313074 on Build GRCh38
rs774980792 Chr.1: 186312975-186312975 on Build GRCh38
rs112053315 Chr.1: 186312676-186312676 on Build GRCh38
rs111255655 Chr.1: 186312744-186312744 on Build GRCh38
rs745562530 Chr.1: 186312835-186312835 on Build GRCh38
rs922311017 Chr.1: 186312778-186312778 on Build GRCh38
rs145515394 Chr.1: 186312866-186312866 on Build GRCh38
rs1005683005 Chr.1: 186313025-186313025 on Build GRCh38
rs945948802 Chr.1: 186312789-186312789 on Build GRCh38
rs767617029 Chr.1: 186312721-186312722 on Build GRCh38
rs571489866 Chr.1: 186312898-186312898 on Build GRCh38
rs984711489 Chr.1: 186312702-186312702 on Build GRCh38
rs751490999 Chr.1: 186312946-186312946 on Build GRCh38
rs775987208 Chr.1: 186312851-186312851 on Build GRCh38
rs758151167 Chr.1: 186312929-186312929 on Build GRCh38
rs766994588 Chr.1: 186312941-186312941 on Build GRCh38
rs370271535 Chr.1: 186312792-186312792 on Build GRCh38
rs935827105 Chr.1: 186312839-186312839 on Build GRCh38
rs78280711 Chr.1: 186312772-186312772 on Build GRCh38
rs925876514 Chr.1: 186312825-186312825 on Build GRCh38
rs1046432362 Chr.1: 186312926-186312926 on Build GRCh38
rs776117124 Chr.1: 186312742-186312742 on Build GRCh38
rs774754880 Chr.1: 186312874-186312874 on Build GRCh38
rs140393971 Chr.1: 186312822-186312822 on Build GRCh38
rs770269033 Chr.1: 186312837-186312837 on Build GRCh38
rs770596036 Chr.1: 186312812-186312812 on Build GRCh38
rs745367093 Chr.1: 186312883-186312883 on Build GRCh38
rs538996915 Chr.1: 186313013-186313013 on Build GRCh38
rs764493899 Chr.1: 186312910-186312911 on Build GRCh38
rs1025800422 Chr.1: 186312667-186312667 on Build GRCh38
rs767690523 Chr.1: 186312769-186312769 on Build GRCh38
rs973022775 Chr.1: 186312611-186312611 on Build GRCh38
rs577666227 Chr.1: 186312602-186312602 on Build GRCh38
rs752727412 Chr.1: 186312723-186312723 on Build GRCh38
rs761807767 Chr.1: 186312761-186312761 on Build GRCh38
rs947544337 Chr.1: 186312599-186312599 on Build GRCh38
rs770320395 Chr.1: 186312740-186312740 on Build GRCh38
rs746479632 Chr.1: 186312811-186312811 on Build GRCh38
rs758290902 Chr.1: 186312803-186312803 on Build GRCh38
rs760956044 Chr.1: 186312893-186312893 on Build GRCh38
rs756629372 Chr.1: 186312725-186312726 on Build GRCh38
rs977597377 Chr.1: 186313000-186313000 on Build GRCh38
rs752573420 Chr.1: 186312796-186312796 on Build GRCh38
rs761925655 Chr.1: 186312879-186312879 on Build GRCh38
rs557224189 Chr.1: 186313019-186313019 on Build GRCh38
rs1043900180 Chr.1: 186312656-186312656 on Build GRCh38
rs767635485 Chr.1: 186312881-186312881 on Build GRCh38
rs80196755 Chr.1: 186312884-186312884 on Build GRCh38
rs994410258 Chr.1: 186312999-186312999 on Build GRCh38
rs529646599 Chr.1: 186312631-186312631 on Build GRCh38
rs901526136 Chr.1: 186313048-186313048 on Build GRCh38
rs764632698 Chr.1: 186312948-186312948 on Build GRCh38
rs935018794 Chr.1: 186312700-186312700 on Build GRCh38
rs750688165 Chr.1: 186312777-186312777 on Build GRCh38
rs751586838 Chr.1: 186312748-186312748 on Build GRCh38
rs201243788 Chr.1: 186312726-186312726 on Build GRCh38
rs548274907 Chr.1: 186312687-186312687 on Build GRCh38
rs1025870826 Chr.1: 186313004-186313004 on Build GRCh38
rs373873150 Chr.1: 186312718-186312718 on Build GRCh38
rs748574099 Chr.1: 186312739-186312739 on Build GRCh38
rs752479638 Chr.1: 186312925-186312925 on Build GRCh38
rs377031640 Chr.1: 186312766-186312766 on Build GRCh38
rs371575509 Chr.1: 186312804-186312804 on Build GRCh38
rs1040074963 Chr.1: 186313033-186313033 on Build GRCh38
rs750079793 Chr.1: 186312941-186312941 on Build GRCh38
rs992158698 Chr.1: 186312947-186312947 on Build GRCh38
rs201960232 Chr.1: 186312938-186312938 on Build GRCh38
rs200043407 Chr.1: 186312867-186312867 on Build GRCh38
rs764074601 Chr.1: 186312743-186312743 on Build GRCh38

View Full Product Details