Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200969701-200970193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF21B
Entrez Gene ID
23046
Gene Name
kinesin family member 21B
Gene Aliases
-
UniGene
Hs.169182
Gene Chromosome Location
Chr.1: 200969386-201023700 on Build GRCh38
Transcripts
NM_001252100.1 NM_001252102.1 NM_001252103.1 NM_017596.3

SNP Detail

SNP ID Chromosome Location
rs769220182 Chr.1: 200969981-200969981 on Build GRCh38
rs1024481293 Chr.1: 200970089-200970089 on Build GRCh38
rs1027437309 Chr.1: 200969977-200969977 on Build GRCh38
rs575150687 Chr.1: 200969825-200969825 on Build GRCh38
rs573509015 Chr.1: 200970145-200970145 on Build GRCh38
rs536908002 Chr.1: 200969776-200969776 on Build GRCh38
rs996958653 Chr.1: 200969869-200969869 on Build GRCh38
rs936056571 Chr.1: 200969813-200969813 on Build GRCh38
rs142990738 Chr.1: 200970003-200970003 on Build GRCh38
rs991744958 Chr.1: 200969729-200969729 on Build GRCh38
rs915057092 Chr.1: 200969844-200969844 on Build GRCh38
rs769595920 Chr.1: 200969955-200969955 on Build GRCh38
rs559904826 Chr.1: 200970073-200970073 on Build GRCh38
rs796712258 Chr.1: 200970112-200970112 on Build GRCh38
rs893835666 Chr.1: 200969914-200969914 on Build GRCh38
rs577295801 Chr.1: 200969999-200969999 on Build GRCh38
rs994570750 Chr.1: 200969888-200969888 on Build GRCh38
rs749878398 Chr.1: 200969871-200969871 on Build GRCh38
rs556584045 Chr.1: 200969790-200969790 on Build GRCh38
rs567900545 Chr.1: 200969757-200969757 on Build GRCh38
rs971421701 Chr.1: 200969736-200969736 on Build GRCh38
rs537459124 Chr.1: 200969852-200969852 on Build GRCh38
rs778056885 Chr.1: 200969733-200969733 on Build GRCh38
rs1050356871 Chr.1: 200969848-200969848 on Build GRCh38
rs1033494852 Chr.1: 200970101-200970101 on Build GRCh38
rs756311883 Chr.1: 200969760-200969760 on Build GRCh38
rs747155111 Chr.1: 200969849-200969849 on Build GRCh38
rs931836269 Chr.1: 200969847-200969847 on Build GRCh38
rs1021337107 Chr.1: 200970034-200970034 on Build GRCh38
rs920282258 Chr.1: 200969764-200969764 on Build GRCh38
rs972020847 Chr.1: 200970104-200970104 on Build GRCh38
rs780268783 Chr.1: 200969857-200969857 on Build GRCh38
rs1013542304 Chr.1: 200970017-200970017 on Build GRCh38
rs977713768 Chr.1: 200970113-200970113 on Build GRCh38
rs1032415263 Chr.1: 200969885-200969885 on Build GRCh38
rs973592534 Chr.1: 200970119-200970119 on Build GRCh38
rs557768432 Chr.1: 200969998-200969998 on Build GRCh38
rs897514040 Chr.1: 200969882-200969882 on Build GRCh38
rs1031607920 Chr.1: 200970157-200970157 on Build GRCh38

View Full Product Details