Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202961781-202962286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5R1
Entrez Gene ID
51706
Gene Name
cytochrome b5 reductase 1
Gene Aliases
B5R.1, B5R1, B5R2, NQO3A2, humb5R2
UniGene
Hs.334832
Gene Chromosome Location
Chr.1: 202961873-202967276 on Build GRCh38
Transcripts
NM_016243.2

SNP Detail

SNP ID Chromosome Location
rs902740333 Chr.1: 202961831-202961831 on Build GRCh38
rs546003132 Chr.1: 202962124-202962124 on Build GRCh38
rs896539247 Chr.1: 202962060-202962060 on Build GRCh38
rs998593359 Chr.1: 202961924-202961924 on Build GRCh38
rs557076104 Chr.1: 202962087-202962087 on Build GRCh38
rs540526309 Chr.1: 202961837-202961837 on Build GRCh38
rs549841273 Chr.1: 202961803-202961803 on Build GRCh38
rs907814700 Chr.1: 202961991-202961991 on Build GRCh38
rs1001107105 Chr.1: 202962214-202962214 on Build GRCh38
rs141835757 Chr.1: 202961844-202961844 on Build GRCh38
rs938117584 Chr.1: 202962021-202962021 on Build GRCh38
rs765596831 Chr.1: 202961806-202961806 on Build GRCh38
rs900167086 Chr.1: 202962240-202962240 on Build GRCh38
rs181308363 Chr.1: 202962053-202962053 on Build GRCh38
rs577148176 Chr.1: 202962098-202962098 on Build GRCh38
rs1012289868 Chr.1: 202962249-202962249 on Build GRCh38
rs780004297 Chr.1: 202962261-202962264 on Build GRCh38
rs186500705 Chr.1: 202962160-202962160 on Build GRCh38
rs955545211 Chr.1: 202961879-202961879 on Build GRCh38
rs544452826 Chr.1: 202962089-202962089 on Build GRCh38
rs182661380 Chr.1: 202962256-202962256 on Build GRCh38
rs1017726014 Chr.1: 202962177-202962177 on Build GRCh38
rs878935736 Chr.1: 202962195-202962195 on Build GRCh38
rs556972438 Chr.1: 202962062-202962062 on Build GRCh38
rs1024738099 Chr.1: 202962250-202962250 on Build GRCh38
rs13107 Chr.1: 202962196-202962196 on Build GRCh38
rs113918756 Chr.1: 202962119-202962119 on Build GRCh38
rs572150243 Chr.1: 202962215-202962215 on Build GRCh38
rs932767660 Chr.1: 202962051-202962051 on Build GRCh38
rs919931359 Chr.1: 202962042-202962042 on Build GRCh38
rs535181011 Chr.1: 202961946-202961946 on Build GRCh38
rs541265190 Chr.1: 202962245-202962245 on Build GRCh38
rs941592851 Chr.1: 202961918-202961918 on Build GRCh38
rs902794227 Chr.1: 202962192-202962192 on Build GRCh38
rs997103815 Chr.1: 202962251-202962251 on Build GRCh38
rs12973 Chr.1: 202962162-202962162 on Build GRCh38
rs111712145 Chr.1: 202961999-202961999 on Build GRCh38
rs139938675 Chr.1: 202962175-202962175 on Build GRCh38
rs561053445 Chr.1: 202962248-202962248 on Build GRCh38
rs376488232 Chr.1: 202962252-202962252 on Build GRCh38
rs1032460112 Chr.1: 202962239-202962239 on Build GRCh38
rs41313934 Chr.1: 202961993-202961993 on Build GRCh38
rs1042202314 Chr.1: 202962083-202962083 on Build GRCh38
rs894811412 Chr.1: 202962245-202962246 on Build GRCh38
rs189266517 Chr.1: 202961890-202961890 on Build GRCh38
rs41310909 Chr.1: 202961899-202961899 on Build GRCh38
rs959416150 Chr.1: 202961864-202961864 on Build GRCh38
rs1021226031 Chr.1: 202961807-202961807 on Build GRCh38
rs1029817978 Chr.1: 202961858-202961858 on Build GRCh38
rs891171353 Chr.1: 202962064-202962064 on Build GRCh38
rs150627492 Chr.1: 202962025-202962025 on Build GRCh38
rs538382202 Chr.1: 202961869-202961869 on Build GRCh38
rs753693184 Chr.1: 202961995-202961995 on Build GRCh38
rs764971017 Chr.1: 202962212-202962212 on Build GRCh38

View Full Product Details