Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202962981-202963488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5R1
Entrez Gene ID
51706
Gene Name
cytochrome b5 reductase 1
Gene Aliases
B5R.1, B5R1, B5R2, NQO3A2, humb5R2
UniGene
Hs.334832
Gene Chromosome Location
Chr.1: 202961873-202967276 on Build GRCh38
Transcripts
NM_016243.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6318654 c.692C>A p.A231D Chr.1: 202963119-202963119 on Build GRCh38
1134799 c.679G>A p.E227K Chr.1: 202963132-202963132 on Build GRCh38
3771591 c.739C>G p.P247A Chr.1: 202963072-202963072 on Build GRCh38
6262884 c.723C>A p.F241L Chr.1: 202963088-202963088 on Build GRCh38
902412 c.707G>A p.R236H Chr.1: 202963104-202963104 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201067717 Chr.1: 202963196-202963196 on Build GRCh38
rs948368845 Chr.1: 202963111-202963111 on Build GRCh38
rs778167846 Chr.1: 202963027-202963027 on Build GRCh38
rs747308777 Chr.1: 202963033-202963033 on Build GRCh38
rs987341630 Chr.1: 202963272-202963272 on Build GRCh38
rs1033820926 Chr.1: 202963071-202963071 on Build GRCh38
rs571169303 Chr.1: 202963405-202963405 on Build GRCh38
rs750972198 Chr.1: 202963412-202963412 on Build GRCh38
rs1035964624 Chr.1: 202963135-202963135 on Build GRCh38
rs201222473 Chr.1: 202963075-202963075 on Build GRCh38
rs2232851 Chr.1: 202963026-202963026 on Build GRCh38
rs940372782 Chr.1: 202963302-202963302 on Build GRCh38
rs903076293 Chr.1: 202963203-202963203 on Build GRCh38
rs761540645 Chr.1: 202963096-202963096 on Build GRCh38
rs561500440 Chr.1: 202963058-202963058 on Build GRCh38
rs920249528 Chr.1: 202963456-202963456 on Build GRCh38
rs770830800 Chr.1: 202963213-202963213 on Build GRCh38
rs760414191 Chr.1: 202963208-202963208 on Build GRCh38
rs757705469 Chr.1: 202963054-202963054 on Build GRCh38
rs930377052 Chr.1: 202963142-202963142 on Build GRCh38
rs921750919 Chr.1: 202963245-202963245 on Build GRCh38
rs754322830 Chr.1: 202963150-202963151 on Build GRCh38
rs144951127 Chr.1: 202963081-202963081 on Build GRCh38
rs773639496 Chr.1: 202963044-202963044 on Build GRCh38
rs111586136 Chr.1: 202963065-202963065 on Build GRCh38
rs1023680892 Chr.1: 202963324-202963326 on Build GRCh38
rs148221696 Chr.1: 202963144-202963144 on Build GRCh38
rs543752468 Chr.1: 202963005-202963005 on Build GRCh38
rs928238369 Chr.1: 202963409-202963409 on Build GRCh38
rs148992048 Chr.1: 202963141-202963141 on Build GRCh38
rs767290000 Chr.1: 202963100-202963100 on Build GRCh38
rs982216546 Chr.1: 202963360-202963360 on Build GRCh38
rs760633859 Chr.1: 202963105-202963105 on Build GRCh38
rs369275806 Chr.1: 202963117-202963117 on Build GRCh38
rs76324202 Chr.1: 202963266-202963266 on Build GRCh38
rs778456223 Chr.1: 202963186-202963186 on Build GRCh38
rs2232850 Chr.1: 202963277-202963277 on Build GRCh38
rs780457584 Chr.1: 202963150-202963150 on Build GRCh38
rs550865482 Chr.1: 202963353-202963353 on Build GRCh38
rs377439886 Chr.1: 202963200-202963200 on Build GRCh38
rs71635599 Chr.1: 202963370-202963370 on Build GRCh38
rs190928825 Chr.1: 202963265-202963265 on Build GRCh38
rs559078788 Chr.1: 202963132-202963132 on Build GRCh38
rs757619233 Chr.1: 202963143-202963143 on Build GRCh38
rs937062536 Chr.1: 202963391-202963391 on Build GRCh38
rs940375660 Chr.1: 202963126-202963126 on Build GRCh38
rs781727349 Chr.1: 202963061-202963061 on Build GRCh38
rs1055327823 Chr.1: 202963417-202963417 on Build GRCh38
rs539733576 Chr.1: 202963426-202963426 on Build GRCh38
rs764625862 Chr.1: 202963148-202963150 on Build GRCh38
rs368686887 Chr.1: 202963091-202963091 on Build GRCh38
rs764352174 Chr.1: 202963134-202963134 on Build GRCh38
rs773061564 Chr.1: 202963104-202963104 on Build GRCh38
rs999548975 Chr.1: 202963240-202963240 on Build GRCh38
rs766280193 Chr.1: 202963116-202963116 on Build GRCh38
rs756594390 Chr.1: 202963148-202963148 on Build GRCh38
rs375092634 Chr.1: 202963025-202963025 on Build GRCh38
rs369808847 Chr.1: 202963180-202963180 on Build GRCh38
rs2232849 Chr.1: 202963314-202963314 on Build GRCh38
rs765359335 Chr.1: 202963012-202963012 on Build GRCh38
rs895431831 Chr.1: 202963080-202963080 on Build GRCh38
rs748766650 Chr.1: 202963155-202963155 on Build GRCh38

View Full Product Details