Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202967042-202967511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5R1
Entrez Gene ID
51706
Gene Name
cytochrome b5 reductase 1
Gene Aliases
B5R.1, B5R1, B5R2, NQO3A2, humb5R2
UniGene
Hs.334832
Gene Chromosome Location
Chr.1: 202961873-202967276 on Build GRCh38
Transcripts
NM_016243.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
678097 c.15G>A p.T5T Chr.1: 202967191-202967191 on Build GRCh38
5379255 c.9C>T p.I3I Chr.1: 202967197-202967197 on Build GRCh38
4848295 c.6G>A p.G2G Chr.1: 202967200-202967200 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs933932362 Chr.1: 202967155-202967155 on Build GRCh38
rs763411132 Chr.1: 202967174-202967174 on Build GRCh38
rs1018050021 Chr.1: 202967080-202967080 on Build GRCh38
rs1034352073 Chr.1: 202967242-202967242 on Build GRCh38
rs868676143 Chr.1: 202967385-202967385 on Build GRCh38
rs12137082 Chr.1: 202967328-202967328 on Build GRCh38
rs767229373 Chr.1: 202967252-202967252 on Build GRCh38
rs1019613885 Chr.1: 202967462-202967462 on Build GRCh38
rs1008517986 Chr.1: 202967452-202967452 on Build GRCh38
rs776046416 Chr.1: 202967172-202967172 on Build GRCh38
rs927452169 Chr.1: 202967245-202967245 on Build GRCh38
rs577918422 Chr.1: 202967201-202967201 on Build GRCh38
rs181846999 Chr.1: 202967079-202967079 on Build GRCh38
rs777752626 Chr.1: 202967148-202967148 on Build GRCh38
rs977070147 Chr.1: 202967116-202967116 on Build GRCh38
rs777190901 Chr.1: 202967224-202967224 on Build GRCh38
rs559163490 Chr.1: 202967271-202967271 on Build GRCh38
rs766135205 Chr.1: 202967208-202967208 on Build GRCh38
rs770166677 Chr.1: 202967238-202967238 on Build GRCh38
rs1010211258 Chr.1: 202967463-202967463 on Build GRCh38
rs909639240 Chr.1: 202967400-202967400 on Build GRCh38
rs200734862 Chr.1: 202967160-202967160 on Build GRCh38
rs575425988 Chr.1: 202967363-202967363 on Build GRCh38
rs372193496 Chr.1: 202967240-202967240 on Build GRCh38
rs746140681 Chr.1: 202967235-202967235 on Build GRCh38
rs983756337 Chr.1: 202967183-202967183 on Build GRCh38
rs893275069 Chr.1: 202967380-202967380 on Build GRCh38
rs59832792 Chr.1: 202967445-202967445 on Build GRCh38
rs576550492 Chr.1: 202967228-202967228 on Build GRCh38
rs764147451 Chr.1: 202967152-202967153 on Build GRCh38
rs909514212 Chr.1: 202967104-202967104 on Build GRCh38
rs533730101 Chr.1: 202967087-202967087 on Build GRCh38
rs536344173 Chr.1: 202967212-202967212 on Build GRCh38
rs574625364 Chr.1: 202967349-202967349 on Build GRCh38
rs112384957 Chr.1: 202967190-202967190 on Build GRCh38
rs563328987 Chr.1: 202967327-202967327 on Build GRCh38
rs947765532 Chr.1: 202967207-202967207 on Build GRCh38
rs397863058 Chr.1: 202967365-202967365 on Build GRCh38
rs11544743 Chr.1: 202967215-202967215 on Build GRCh38
rs1024408311 Chr.1: 202967268-202967268 on Build GRCh38
rs761438720 Chr.1: 202967251-202967251 on Build GRCh38
rs111910733 Chr.1: 202967189-202967189 on Build GRCh38
rs941047335 Chr.1: 202967125-202967125 on Build GRCh38
rs538979563 Chr.1: 202967266-202967266 on Build GRCh38
rs776919033 Chr.1: 202967151-202967151 on Build GRCh38
rs781418798 Chr.1: 202967229-202967229 on Build GRCh38
rs371484143 Chr.1: 202967194-202967194 on Build GRCh38
rs770155729 Chr.1: 202967161-202967161 on Build GRCh38
rs376595591 Chr.1: 202967290-202967290 on Build GRCh38
rs754935900 Chr.1: 202967213-202967213 on Build GRCh38
rs374636337 Chr.1: 202967150-202967150 on Build GRCh38
rs760283435 Chr.1: 202967257-202967257 on Build GRCh38
rs934391101 Chr.1: 202967351-202967351 on Build GRCh38
rs769057887 Chr.1: 202967244-202967244 on Build GRCh38
rs772974642 Chr.1: 202967254-202967254 on Build GRCh38
rs35356717 Chr.1: 202967399-202967399 on Build GRCh38
rs748126018 Chr.1: 202967142-202967142 on Build GRCh38
rs767282339 Chr.1: 202967197-202967197 on Build GRCh38
rs749791275 Chr.1: 202967243-202967243 on Build GRCh38
rs34591036 Chr.1: 202967361-202967361 on Build GRCh38
rs1028534794 Chr.1: 202967416-202967416 on Build GRCh38
rs34807965 Chr.1: 202967366-202967366 on Build GRCh38
rs951191004 Chr.1: 202967076-202967076 on Build GRCh38
rs755957956 Chr.1: 202967205-202967205 on Build GRCh38
rs772167743 Chr.1: 202967144-202967144 on Build GRCh38
rs199856907 Chr.1: 202967191-202967191 on Build GRCh38
rs567416582 Chr.1: 202967204-202967204 on Build GRCh38
rs773933353 Chr.1: 202967248-202967248 on Build GRCh38

View Full Product Details