Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203798238-203798749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZC3H11A
Entrez Gene ID
9877
Gene Name
zinc finger CCCH-type containing 11A
Gene Aliases
ZC3HDC11A
UniGene
Hs.532399
Gene Chromosome Location
Chr.1: 203795537-203854128 on Build GRCh38
Transcripts
NM_001319238.1 NM_001319239.1 NM_014827.5 XM_005245643.2 XM_005245644.3 XM_006711673.2 XM_011510199.1 XM_011510200.1 XM_011510201.1 XM_011510202.1 XM_017002956.1 XM_017002957.1 XM_017002958.1 XM_017002959.1 XM_017002960.1 XM_017002961.1 XM_017002962.1 XM_017002963.1 XM_017002964.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6336027 c.790G>T p.A264S Chr.1: 203798312-203798312 on Build GRCh38
7341876 c.1192G>A p.E398K Chr.1: 203798714-203798714 on Build GRCh38
5626022 c.903A>G p.L301L Chr.1: 203798425-203798425 on Build GRCh38
3803539 c.1157C>T p.A386V Chr.1: 203798679-203798679 on Build GRCh38
3943345 c.743T>G p.L248R Chr.1: 203798265-203798265 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573597462 Chr.1: 203798326-203798326 on Build GRCh38
rs576073615 Chr.1: 203798468-203798468 on Build GRCh38
rs750401100 Chr.1: 203798705-203798705 on Build GRCh38
rs374322432 Chr.1: 203798540-203798540 on Build GRCh38
rs770480012 Chr.1: 203798403-203798403 on Build GRCh38
rs752482003 Chr.1: 203798270-203798270 on Build GRCh38
rs969212245 Chr.1: 203798274-203798274 on Build GRCh38
rs77585755 Chr.1: 203798675-203798675 on Build GRCh38
rs976453033 Chr.1: 203798696-203798696 on Build GRCh38
rs767740097 Chr.1: 203798725-203798725 on Build GRCh38
rs777550685 Chr.1: 203798346-203798346 on Build GRCh38
rs564728552 Chr.1: 203798496-203798496 on Build GRCh38
rs947435131 Chr.1: 203798441-203798441 on Build GRCh38
rs370618612 Chr.1: 203798519-203798519 on Build GRCh38
rs758114772 Chr.1: 203798327-203798327 on Build GRCh38
rs570640442 Chr.1: 203798699-203798699 on Build GRCh38
rs545060878 Chr.1: 203798473-203798473 on Build GRCh38
rs542442615 Chr.1: 203798435-203798435 on Build GRCh38
rs775199780 Chr.1: 203798598-203798598 on Build GRCh38
rs923549475 Chr.1: 203798342-203798342 on Build GRCh38
rs910089894 Chr.1: 203798642-203798642 on Build GRCh38
rs781143251 Chr.1: 203798314-203798314 on Build GRCh38
rs913391110 Chr.1: 203798713-203798713 on Build GRCh38
rs775730425 Chr.1: 203798579-203798579 on Build GRCh38
rs890377253 Chr.1: 203798721-203798721 on Build GRCh38
rs941775801 Chr.1: 203798669-203798669 on Build GRCh38
rs892007134 Chr.1: 203798410-203798410 on Build GRCh38
rs746635288 Chr.1: 203798359-203798359 on Build GRCh38
rs537020297 Chr.1: 203798617-203798617 on Build GRCh38
rs555924237 Chr.1: 203798445-203798445 on Build GRCh38
rs764690334 Chr.1: 203798574-203798574 on Build GRCh38
rs1035979106 Chr.1: 203798505-203798505 on Build GRCh38
rs769877528 Chr.1: 203798640-203798641 on Build GRCh38
rs140585450 Chr.1: 203798700-203798700 on Build GRCh38
rs970215585 Chr.1: 203798551-203798551 on Build GRCh38
rs762568030 Chr.1: 203798635-203798635 on Build GRCh38
rs780630170 Chr.1: 203798419-203798419 on Build GRCh38
rs763814434 Chr.1: 203798645-203798645 on Build GRCh38
rs759091595 Chr.1: 203798573-203798573 on Build GRCh38
rs1025431641 Chr.1: 203798559-203798559 on Build GRCh38
rs746404400 Chr.1: 203798485-203798485 on Build GRCh38
rs745833264 Chr.1: 203798723-203798725 on Build GRCh38
rs1053272593 Chr.1: 203798405-203798405 on Build GRCh38
rs989086525 Chr.1: 203798708-203798709 on Build GRCh38
rs757124023 Chr.1: 203798266-203798266 on Build GRCh38
rs747382390 Chr.1: 203798492-203798492 on Build GRCh38
rs527242325 Chr.1: 203798654-203798654 on Build GRCh38
rs748387695 Chr.1: 203798443-203798443 on Build GRCh38
rs749320203 Chr.1: 203798392-203798392 on Build GRCh38
rs553693791 Chr.1: 203798279-203798279 on Build GRCh38
rs542266903 Chr.1: 203798546-203798546 on Build GRCh38
rs1001562186 Chr.1: 203798487-203798487 on Build GRCh38
rs980677910 Chr.1: 203798560-203798560 on Build GRCh38
rs983804082 Chr.1: 203798672-203798672 on Build GRCh38
rs6697388 Chr.1: 203798694-203798694 on Build GRCh38
rs762102398 Chr.1: 203798704-203798704 on Build GRCh38

View Full Product Details