Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244851203-244851747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPU
Entrez Gene ID
3192
Gene Name
heterogeneous nuclear ribonucleoprotein U
Gene Aliases
HNRNPU-AS1, HNRPU, SAF-A, SAFA, U21.1, hnRNP U
UniGene
Hs.106212
Gene Chromosome Location
Chr.1: 244842123-244864720 on Build GRCh38
Transcripts
NM_004501.3 NM_031844.2 XM_017001115.1 XM_017001116.1 XM_017001117.1

SNP Detail

SNP ID Chromosome Location
rs995352473 Chr.1: 244851280-244851280 on Build GRCh38
rs559973295 Chr.1: 244851690-244851690 on Build GRCh38
rs375378291 Chr.1: 244851657-244851658 on Build GRCh38
rs1028129629 Chr.1: 244851318-244851318 on Build GRCh38
rs546202352 Chr.1: 244851304-244851304 on Build GRCh38
rs772957676 Chr.1: 244851342-244851342 on Build GRCh38
rs916160142 Chr.1: 244851620-244851625 on Build GRCh38
rs1053160057 Chr.1: 244851679-244851679 on Build GRCh38
rs139342570 Chr.1: 244851408-244851408 on Build GRCh38
rs886973355 Chr.1: 244851526-244851526 on Build GRCh38
rs1046271411 Chr.1: 244851390-244851390 on Build GRCh38
rs961092732 Chr.1: 244851482-244851482 on Build GRCh38
rs920943747 Chr.1: 244851231-244851231 on Build GRCh38
rs946231623 Chr.1: 244851692-244851692 on Build GRCh38
rs184618218 Chr.1: 244851670-244851670 on Build GRCh38
rs985446812 Chr.1: 244851610-244851610 on Build GRCh38
rs527867316 Chr.1: 244851490-244851490 on Build GRCh38
rs767765330 Chr.1: 244851420-244851420 on Build GRCh38
rs193032660 Chr.1: 244851606-244851606 on Build GRCh38
rs1014916366 Chr.1: 244851340-244851340 on Build GRCh38
rs759739215 Chr.1: 244851371-244851371 on Build GRCh38
rs1012878259 Chr.1: 244851379-244851382 on Build GRCh38
rs1004964538 Chr.1: 244851578-244851578 on Build GRCh38
rs548144884 Chr.1: 244851558-244851558 on Build GRCh38
rs927564125 Chr.1: 244851362-244851362 on Build GRCh38
rs760810802 Chr.1: 244851582-244851582 on Build GRCh38
rs986959117 Chr.1: 244851255-244851257 on Build GRCh38
rs983820892 Chr.1: 244851338-244851338 on Build GRCh38
rs899653962 Chr.1: 244851275-244851275 on Build GRCh38
rs953883476 Chr.1: 244851474-244851475 on Build GRCh38
rs545233223 Chr.1: 244851483-244851484 on Build GRCh38
rs948761447 Chr.1: 244851636-244851636 on Build GRCh38
rs771652857 Chr.1: 244851468-244851469 on Build GRCh38
rs899768401 Chr.1: 244851481-244851481 on Build GRCh38
rs914743441 Chr.1: 244851678-244851678 on Build GRCh38
rs981427918 Chr.1: 244851632-244851632 on Build GRCh38
rs562694871 Chr.1: 244851398-244851398 on Build GRCh38
rs1008146616 Chr.1: 244851242-244851242 on Build GRCh38
rs367771932 Chr.1: 244851240-244851243 on Build GRCh38
rs1031528783 Chr.1: 244851422-244851423 on Build GRCh38
rs1034525657 Chr.1: 244851642-244851642 on Build GRCh38
rs981639424 Chr.1: 244851357-244851357 on Build GRCh38
rs892977991 Chr.1: 244851686-244851688 on Build GRCh38
rs907175637 Chr.1: 244851654-244851655 on Build GRCh38
rs952166210 Chr.1: 244851322-244851322 on Build GRCh38
rs375170946 Chr.1: 244851445-244851445 on Build GRCh38
rs371929666 Chr.1: 244851487-244851488 on Build GRCh38
rs774299712 Chr.1: 244851719-244851719 on Build GRCh38
rs930777326 Chr.1: 244851575-244851575 on Build GRCh38
rs757534329 Chr.1: 244851382-244851385 on Build GRCh38
rs958958427 Chr.1: 244851659-244851659 on Build GRCh38
rs1015445194 Chr.1: 244851612-244851612 on Build GRCh38
rs562315330 Chr.1: 244851451-244851451 on Build GRCh38
rs971357302 Chr.1: 244851619-244851619 on Build GRCh38
rs1041898398 Chr.1: 244851432-244851432 on Build GRCh38
rs995300145 Chr.1: 244851488-244851488 on Build GRCh38
rs533578473 Chr.1: 244851238-244851241 on Build GRCh38
rs943715526 Chr.1: 244851450-244851450 on Build GRCh38
rs1018228613 Chr.1: 244851260-244851260 on Build GRCh38
rs1036474501 Chr.1: 244851319-244851319 on Build GRCh38
rs752804161 Chr.1: 244851477-244851477 on Build GRCh38
rs770096424 Chr.1: 244851452-244851453 on Build GRCh38
rs187978986 Chr.1: 244851270-244851270 on Build GRCh38
rs937488878 Chr.1: 244851374-244851374 on Build GRCh38
rs1011412169 Chr.1: 244851712-244851712 on Build GRCh38
rs1026826619 Chr.1: 244851503-244851503 on Build GRCh38
rs930912849 Chr.1: 244851330-244851330 on Build GRCh38
rs1043631 Chr.1: 244851395-244851395 on Build GRCh38

View Full Product Details