Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 824950-825498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LARP4B
Entrez Gene ID
23185
Gene Name
La ribonucleoprotein domain family member 4B
Gene Aliases
KIAA0217, LARP5
UniGene
Hs.159069
Gene Chromosome Location
Chr.10: 806914-931900 on Build GRCh38
Transcripts
NM_015155.2 XM_005252432.3 XM_005252435.3 XM_011519435.2 XM_017015987.1 XM_017015988.1 XM_017015989.1 XM_017015990.1 XM_017015992.1 XM_017015993.1 XM_017015994.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3770702 c.1393C>T p.Q465* Chr.10: 825156-825156 on Build GRCh38
5002724 c.134G>T p.R45L Chr.10: 825111-825111 on Build GRCh38
1349350 c.1262C>T p.A421V Chr.10: 825287-825287 on Build GRCh38
5801616 c.1389delG p.I464fs*28 Chr.10: 825160-825160 on Build GRCh38
178078 c.1456G>A p.E486K Chr.10: 825093-825093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374286524 Chr.10: 825096-825096 on Build GRCh38
rs138081905 Chr.10: 825287-825287 on Build GRCh38
rs746027286 Chr.10: 825055-825055 on Build GRCh38
rs776830731 Chr.10: 824998-824998 on Build GRCh38
rs372744681 Chr.10: 825240-825240 on Build GRCh38
rs747911468 Chr.10: 825021-825021 on Build GRCh38
rs1041086834 Chr.10: 825382-825382 on Build GRCh38
rs916710282 Chr.10: 825120-825120 on Build GRCh38
rs756355969 Chr.10: 825366-825366 on Build GRCh38
rs2026567 Chr.10: 825338-825338 on Build GRCh38
rs1042068969 Chr.10: 825324-825324 on Build GRCh38
rs974860961 Chr.10: 825022-825022 on Build GRCh38
rs776027544 Chr.10: 825082-825082 on Build GRCh38
rs369825497 Chr.10: 825026-825026 on Build GRCh38
rs761616993 Chr.10: 825332-825333 on Build GRCh38
rs201396176 Chr.10: 825355-825355 on Build GRCh38
rs769751907 Chr.10: 825057-825057 on Build GRCh38
rs965156681 Chr.10: 825010-825010 on Build GRCh38
rs145294175 Chr.10: 825206-825206 on Build GRCh38
rs954803522 Chr.10: 825466-825466 on Build GRCh38
rs778817653 Chr.10: 825019-825019 on Build GRCh38
rs765670503 Chr.10: 825252-825252 on Build GRCh38
rs944109654 Chr.10: 825369-825369 on Build GRCh38
rs754997742 Chr.10: 825142-825142 on Build GRCh38
rs765216921 Chr.10: 825149-825149 on Build GRCh38
rs140223312 Chr.10: 825170-825170 on Build GRCh38
rs759058698 Chr.10: 825294-825294 on Build GRCh38
rs761550184 Chr.10: 825351-825351 on Build GRCh38
rs750590095 Chr.10: 825197-825197 on Build GRCh38
rs776303473 Chr.10: 825293-825293 on Build GRCh38
rs577633540 Chr.10: 825127-825127 on Build GRCh38
rs186064987 Chr.10: 825033-825033 on Build GRCh38
rs572575150 Chr.10: 825340-825340 on Build GRCh38
rs774724850 Chr.10: 825094-825094 on Build GRCh38
rs1032202268 Chr.10: 824996-824996 on Build GRCh38
rs908111254 Chr.10: 825100-825100 on Build GRCh38
rs367827564 Chr.10: 825113-825113 on Build GRCh38
rs964576286 Chr.10: 825364-825364 on Build GRCh38
rs772963374 Chr.10: 825286-825286 on Build GRCh38
rs779321717 Chr.10: 825260-825260 on Build GRCh38
rs781536895 Chr.10: 825178-825178 on Build GRCh38
rs1030432527 Chr.10: 825464-825464 on Build GRCh38
rs765964052 Chr.10: 825116-825116 on Build GRCh38
rs769116294 Chr.10: 825092-825092 on Build GRCh38
rs937724310 Chr.10: 825200-825200 on Build GRCh38
rs748542556 Chr.10: 825050-825050 on Build GRCh38
rs79897367 Chr.10: 825107-825107 on Build GRCh38
rs758124632 Chr.10: 825172-825172 on Build GRCh38
rs748609302 Chr.10: 825268-825268 on Build GRCh38
rs749767169 Chr.10: 825086-825086 on Build GRCh38
rs539520658 Chr.10: 824983-824983 on Build GRCh38
rs772308241 Chr.10: 825276-825276 on Build GRCh38
rs751332874 Chr.10: 825361-825361 on Build GRCh38
rs767199153 Chr.10: 825106-825106 on Build GRCh38
rs996237648 Chr.10: 825384-825384 on Build GRCh38
rs770665715 Chr.10: 825288-825288 on Build GRCh38
rs780140835 Chr.10: 825218-825218 on Build GRCh38
rs936990157 Chr.10: 825193-825193 on Build GRCh38
rs375939556 Chr.10: 825348-825348 on Build GRCh38
rs1038273461 Chr.10: 825141-825141 on Build GRCh38
rs773720322 Chr.10: 825444-825444 on Build GRCh38

View Full Product Details