Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13107962-13108477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPTN
Entrez Gene ID
10133
Gene Name
optineurin
Gene Aliases
ALS12, FIP2, GLC1E, HIP7, HYPL, NRP, TFIIIA-INTP
UniGene
Hs.332706
Gene Chromosome Location
Chr.10: 13100082-13138276 on Build GRCh38
Transcripts
NM_001008211.1 NM_001008212.1 NM_001008213.1 NM_021980.4

SNP Detail

SNP ID Chromosome Location
rs1046572572 Chr.10: 13108259-13108259 on Build GRCh38
rs928095026 Chr.10: 13108016-13108016 on Build GRCh38
rs913955400 Chr.10: 13108092-13108092 on Build GRCh38
rs749376810 Chr.10: 13108364-13108364 on Build GRCh38
rs942608021 Chr.10: 13108328-13108328 on Build GRCh38
rs34546097 Chr.10: 13108374-13108374 on Build GRCh38
rs901012866 Chr.10: 13108354-13108354 on Build GRCh38
rs1036788514 Chr.10: 13108190-13108190 on Build GRCh38
rs976531324 Chr.10: 13108153-13108153 on Build GRCh38
rs1004395261 Chr.10: 13107997-13107997 on Build GRCh38
rs868561312 Chr.10: 13108243-13108243 on Build GRCh38
rs149005252 Chr.10: 13108283-13108283 on Build GRCh38
rs1032785486 Chr.10: 13108048-13108048 on Build GRCh38
rs1030720652 Chr.10: 13108156-13108156 on Build GRCh38
rs1050844257 Chr.10: 13108287-13108287 on Build GRCh38
rs566940047 Chr.10: 13108071-13108071 on Build GRCh38
rs185323491 Chr.10: 13108165-13108165 on Build GRCh38
rs528934051 Chr.10: 13108288-13108288 on Build GRCh38
rs1023921327 Chr.10: 13108185-13108185 on Build GRCh38
rs544897020 Chr.10: 13108201-13108201 on Build GRCh38
rs982341658 Chr.10: 13108195-13108195 on Build GRCh38
rs769100096 Chr.10: 13108372-13108372 on Build GRCh38
rs41291309 Chr.10: 13108265-13108265 on Build GRCh38
rs984480297 Chr.10: 13108433-13108433 on Build GRCh38
rs751354321 Chr.10: 13108061-13108061 on Build GRCh38
rs11258191 Chr.10: 13108040-13108040 on Build GRCh38
rs1036356795 Chr.10: 13108038-13108038 on Build GRCh38
rs989120625 Chr.10: 13108177-13108177 on Build GRCh38
rs953902224 Chr.10: 13108433-13108433 on Build GRCh38
rs909765276 Chr.10: 13108436-13108436 on Build GRCh38
rs1013777043 Chr.10: 13108236-13108236 on Build GRCh38
rs893549026 Chr.10: 13108402-13108402 on Build GRCh38
rs553528449 Chr.10: 13108058-13108058 on Build GRCh38
rs965160856 Chr.10: 13108094-13108094 on Build GRCh38
rs965405239 Chr.10: 13108336-13108336 on Build GRCh38
rs746080607 Chr.10: 13108189-13108189 on Build GRCh38
rs921206032 Chr.10: 13108155-13108155 on Build GRCh38
rs542461161 Chr.10: 13108301-13108301 on Build GRCh38
rs1010597996 Chr.10: 13108420-13108420 on Build GRCh38
rs570836408 Chr.10: 13108045-13108045 on Build GRCh38
rs372030158 Chr.10: 13108019-13108019 on Build GRCh38
rs1048291398 Chr.10: 13108167-13108167 on Build GRCh38
rs1040915811 Chr.10: 13108353-13108353 on Build GRCh38
rs575885963 Chr.10: 13108186-13108186 on Build GRCh38
rs77158107 Chr.10: 13108055-13108055 on Build GRCh38
rs940776250 Chr.10: 13108020-13108020 on Build GRCh38
rs577586552 Chr.10: 13108277-13108277 on Build GRCh38
rs41291307 Chr.10: 13108110-13108110 on Build GRCh38
rs920655727 Chr.10: 13108282-13108282 on Build GRCh38
rs780893081 Chr.10: 13108157-13108157 on Build GRCh38
rs539807267 Chr.10: 13108278-13108278 on Build GRCh38
rs2304706 Chr.10: 13108044-13108044 on Build GRCh38
rs1015350836 Chr.10: 13107998-13107998 on Build GRCh38

View Full Product Details