Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 15557936-15558451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA8
Entrez Gene ID
8516
Gene Name
integrin subunit alpha 8
Gene Aliases
-
UniGene
Hs.171311
Gene Chromosome Location
Chr.10: 15513949-15720335 on Build GRCh38
Transcripts
NM_001291494.1 NM_003638.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1702092 c.2656G>A p.D886N Chr.10: 15558184-15558184 on Build GRCh38
7401366 c.2748G>C p.Q916H Chr.10: 15558092-15558092 on Build GRCh38
5647789 c.2642C>T p.A881V Chr.10: 15558198-15558198 on Build GRCh38
6372072 c.2717G>T p.R906M Chr.10: 15558123-15558123 on Build GRCh38
75041 c.2684G>A p.R895Q Chr.10: 15558156-15558156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891727606 Chr.10: 15557995-15557995 on Build GRCh38
rs561240801 Chr.10: 15558413-15558413 on Build GRCh38
rs142740699 Chr.10: 15558167-15558167 on Build GRCh38
rs745543817 Chr.10: 15558252-15558252 on Build GRCh38
rs991641007 Chr.10: 15558279-15558279 on Build GRCh38
rs908662081 Chr.10: 15558019-15558019 on Build GRCh38
rs751431332 Chr.10: 15558241-15558241 on Build GRCh38
rs149745919 Chr.10: 15558089-15558089 on Build GRCh38
rs770613585 Chr.10: 15558166-15558166 on Build GRCh38
rs776662841 Chr.10: 15558213-15558213 on Build GRCh38
rs532745123 Chr.10: 15558131-15558131 on Build GRCh38
rs941448903 Chr.10: 15558044-15558044 on Build GRCh38
rs752710916 Chr.10: 15558157-15558157 on Build GRCh38
rs748711612 Chr.10: 15558178-15558178 on Build GRCh38
rs545929379 Chr.10: 15557967-15557967 on Build GRCh38
rs763796174 Chr.10: 15558239-15558239 on Build GRCh38
rs537111237 Chr.10: 15558164-15558164 on Build GRCh38
rs767955888 Chr.10: 15558128-15558128 on Build GRCh38
rs758364474 Chr.10: 15558234-15558234 on Build GRCh38
rs988487078 Chr.10: 15558255-15558255 on Build GRCh38
rs145550180 Chr.10: 15558136-15558136 on Build GRCh38
rs762238451 Chr.10: 15558321-15558321 on Build GRCh38
rs754889447 Chr.10: 15558154-15558154 on Build GRCh38
rs746700510 Chr.10: 15558086-15558086 on Build GRCh38
rs1015519808 Chr.10: 15558138-15558138 on Build GRCh38
rs543077714 Chr.10: 15558370-15558370 on Build GRCh38
rs760880676 Chr.10: 15558207-15558207 on Build GRCh38
rs1050215814 Chr.10: 15558424-15558424 on Build GRCh38
rs754912131 Chr.10: 15558025-15558025 on Build GRCh38
rs750561487 Chr.10: 15558133-15558133 on Build GRCh38
rs983224802 Chr.10: 15558322-15558322 on Build GRCh38
rs528700348 Chr.10: 15558018-15558018 on Build GRCh38
rs561484690 Chr.10: 15558016-15558016 on Build GRCh38
rs371707660 Chr.10: 15558224-15558224 on Build GRCh38
rs917511443 Chr.10: 15558292-15558292 on Build GRCh38
rs912876441 Chr.10: 15558270-15558270 on Build GRCh38
rs369329220 Chr.10: 15558216-15558216 on Build GRCh38
rs575551737 Chr.10: 15558369-15558369 on Build GRCh38
rs780833154 Chr.10: 15558171-15558171 on Build GRCh38
rs781029235 Chr.10: 15558246-15558246 on Build GRCh38
rs571642992 Chr.10: 15558114-15558114 on Build GRCh38
rs746651259 Chr.10: 15558110-15558111 on Build GRCh38
rs991714682 Chr.10: 15557978-15557978 on Build GRCh38
rs747943982 Chr.10: 15558069-15558069 on Build GRCh38
rs1009018078 Chr.10: 15558145-15558145 on Build GRCh38
rs539023139 Chr.10: 15558350-15558350 on Build GRCh38
rs776289109 Chr.10: 15558090-15558090 on Build GRCh38
rs371931316 Chr.10: 15558208-15558208 on Build GRCh38
rs777250717 Chr.10: 15558082-15558082 on Build GRCh38
rs1050276861 Chr.10: 15558117-15558117 on Build GRCh38
rs375026864 Chr.10: 15558242-15558242 on Build GRCh38
rs762120352 Chr.10: 15558115-15558115 on Build GRCh38
rs200167471 Chr.10: 15558172-15558172 on Build GRCh38
rs1043393742 Chr.10: 15558284-15558284 on Build GRCh38
rs114268426 Chr.10: 15558354-15558354 on Build GRCh38
rs185082435 Chr.10: 15558309-15558309 on Build GRCh38
rs373750835 Chr.10: 15558078-15558078 on Build GRCh38
rs534845463 Chr.10: 15558222-15558222 on Build GRCh38
rs180754486 Chr.10: 15558141-15558141 on Build GRCh38
rs368768931 Chr.10: 15558095-15558095 on Build GRCh38
rs778667154 Chr.10: 15558028-15558028 on Build GRCh38
rs138187704 Chr.10: 15558106-15558106 on Build GRCh38
rs756774200 Chr.10: 15558113-15558114 on Build GRCh38
rs774677009 Chr.10: 15558175-15558175 on Build GRCh38
rs374258924 Chr.10: 15558139-15558139 on Build GRCh38
rs1008242881 Chr.10: 15558394-15558394 on Build GRCh38
rs961624156 Chr.10: 15558142-15558142 on Build GRCh38
rs368464139 Chr.10: 15558156-15558156 on Build GRCh38
rs567428315 Chr.10: 15558179-15558179 on Build GRCh38
rs1037507775 Chr.10: 15558053-15558053 on Build GRCh38
rs9333214 Chr.10: 15557993-15557993 on Build GRCh38
rs745638635 Chr.10: 15558273-15558273 on Build GRCh38
rs1011877077 Chr.10: 15558009-15558009 on Build GRCh38
rs2277204 Chr.10: 15558272-15558272 on Build GRCh38

View Full Product Details