Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 15782028-15782546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM188A
Entrez Gene ID
80013
Gene Name
family with sequence similarity 188 member A
Gene Aliases
C10orf97, CARP, DERP5, MST126, MSTP126, my042
UniGene
Hs.158870
Gene Chromosome Location
Chr.10: 15778169-15860533 on Build GRCh38
Transcripts
NM_001318330.1 NM_024948.3 XM_005252600.2 XM_005252602.2 XM_006717508.2 XM_011519690.2 XM_011519691.2 XM_011519693.1 XM_011519694.1 XM_017016671.1 XM_017016672.1 XM_017016673.1 XM_017016674.1 XM_017016675.1 XM_017016676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
299209 c.1136C>A p.S379Y Chr.10: 15782207-15782207 on Build GRCh38
6410660 c.1118G>T p.G373V Chr.10: 15782225-15782225 on Build GRCh38
3436250 c.1136C>T p.S379F Chr.10: 15782207-15782207 on Build GRCh38
7304897 c.1119C>T p.G373G Chr.10: 15782224-15782224 on Build GRCh38
3686666 c.1180A>G p.N394D Chr.10: 15782163-15782163 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1010106256 Chr.10: 15782317-15782317 on Build GRCh38
rs146321227 Chr.10: 15782508-15782508 on Build GRCh38
rs771851051 Chr.10: 15782292-15782292 on Build GRCh38
rs138087852 Chr.10: 15782255-15782259 on Build GRCh38
rs961744514 Chr.10: 15782420-15782420 on Build GRCh38
rs534487716 Chr.10: 15782149-15782149 on Build GRCh38
rs999825777 Chr.10: 15782428-15782428 on Build GRCh38
rs757635397 Chr.10: 15782267-15782267 on Build GRCh38
rs768314299 Chr.10: 15782164-15782164 on Build GRCh38
rs7912258 Chr.10: 15782093-15782093 on Build GRCh38
rs371725801 Chr.10: 15782135-15782135 on Build GRCh38
rs761607721 Chr.10: 15782104-15782104 on Build GRCh38
rs757434545 Chr.10: 15782132-15782132 on Build GRCh38
rs113526994 Chr.10: 15782491-15782491 on Build GRCh38
rs563927443 Chr.10: 15782503-15782503 on Build GRCh38
rs544921036 Chr.10: 15782425-15782425 on Build GRCh38
rs532835336 Chr.10: 15782063-15782063 on Build GRCh38
rs1038787542 Chr.10: 15782343-15782343 on Build GRCh38
rs771783690 Chr.10: 15782199-15782199 on Build GRCh38
rs767300788 Chr.10: 15782109-15782109 on Build GRCh38
rs967045440 Chr.10: 15782136-15782136 on Build GRCh38
rs573407794 Chr.10: 15782348-15782348 on Build GRCh38
rs372317948 Chr.10: 15782447-15782447 on Build GRCh38
rs574853100 Chr.10: 15782349-15782349 on Build GRCh38
rs868125661 Chr.10: 15782498-15782498 on Build GRCh38
rs181239213 Chr.10: 15782251-15782251 on Build GRCh38
rs750712940 Chr.10: 15782275-15782275 on Build GRCh38
rs548759483 Chr.10: 15782087-15782087 on Build GRCh38
rs374654897 Chr.10: 15782202-15782202 on Build GRCh38
rs183711073 Chr.10: 15782493-15782493 on Build GRCh38
rs530388712 Chr.10: 15782487-15782487 on Build GRCh38
rs188522968 Chr.10: 15782074-15782074 on Build GRCh38
rs952958561 Chr.10: 15782456-15782456 on Build GRCh38
rs375044749 Chr.10: 15782141-15782141 on Build GRCh38
rs199510240 Chr.10: 15782260-15782265 on Build GRCh38
rs11818243 Chr.10: 15782086-15782086 on Build GRCh38
rs953102135 Chr.10: 15782117-15782117 on Build GRCh38
rs1048419967 Chr.10: 15782256-15782256 on Build GRCh38
rs749912869 Chr.10: 15782114-15782114 on Build GRCh38
rs958979123 Chr.10: 15782472-15782472 on Build GRCh38
rs1033131404 Chr.10: 15782462-15782462 on Build GRCh38
rs895670975 Chr.10: 15782075-15782075 on Build GRCh38
rs749288780 Chr.10: 15782159-15782159 on Build GRCh38
rs751838176 Chr.10: 15782478-15782478 on Build GRCh38
rs191126254 Chr.10: 15782355-15782355 on Build GRCh38
rs769446285 Chr.10: 15782348-15782348 on Build GRCh38
rs374883099 Chr.10: 15782121-15782121 on Build GRCh38
rs542266736 Chr.10: 15782419-15782419 on Build GRCh38
rs543116495 Chr.10: 15782323-15782323 on Build GRCh38
rs534741031 Chr.10: 15782333-15782333 on Build GRCh38
rs565077933 Chr.10: 15782479-15782480 on Build GRCh38
rs552726614 Chr.10: 15782195-15782195 on Build GRCh38
rs746462837 Chr.10: 15782384-15782384 on Build GRCh38
rs557655924 Chr.10: 15782069-15782069 on Build GRCh38
rs747987545 Chr.10: 15782182-15782182 on Build GRCh38
rs767717048 Chr.10: 15782268-15782268 on Build GRCh38
rs1057222934 Chr.10: 15782485-15782485 on Build GRCh38
rs774240028 Chr.10: 15782173-15782173 on Build GRCh38
rs941343587 Chr.10: 15782335-15782335 on Build GRCh38
rs764556987 Chr.10: 15782123-15782123 on Build GRCh38
rs370057014 Chr.10: 15782165-15782165 on Build GRCh38
rs775826391 Chr.10: 15782229-15782229 on Build GRCh38
rs369350483 Chr.10: 15782122-15782122 on Build GRCh38
rs781400472 Chr.10: 15782134-15782134 on Build GRCh38
rs760088852 Chr.10: 15782218-15782218 on Build GRCh38
rs879016312 Chr.10: 15782096-15782099 on Build GRCh38
rs751754202 Chr.10: 15782131-15782131 on Build GRCh38
rs904465296 Chr.10: 15782324-15782324 on Build GRCh38
rs149583944 Chr.10: 15782477-15782478 on Build GRCh38
rs763561663 Chr.10: 15782231-15782231 on Build GRCh38
rs553408659 Chr.10: 15782342-15782342 on Build GRCh38
rs879327486 Chr.10: 15782101-15782101 on Build GRCh38
rs995992576 Chr.10: 15782079-15782079 on Build GRCh38
rs372752445 Chr.10: 15782422-15782422 on Build GRCh38
rs563278484 Chr.10: 15782430-15782430 on Build GRCh38
rs185779128 Chr.10: 15782276-15782276 on Build GRCh38
rs765977882 Chr.10: 15782222-15782222 on Build GRCh38
rs982888916 Chr.10: 15782328-15782328 on Build GRCh38
rs1028684820 Chr.10: 15782112-15782112 on Build GRCh38
rs756277604 Chr.10: 15782138-15782138 on Build GRCh38
rs927254798 Chr.10: 15782478-15782478 on Build GRCh38
rs1041438190 Chr.10: 15782424-15782424 on Build GRCh38
rs538534854 Chr.10: 15782259-15782259 on Build GRCh38
rs1033231237 Chr.10: 15782244-15782244 on Build GRCh38
rs973386537 Chr.10: 15782140-15782140 on Build GRCh38
rs754604264 Chr.10: 15782119-15782119 on Build GRCh38
rs369821482 Chr.10: 15782301-15782301 on Build GRCh38

View Full Product Details