Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 30438824-30439319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K8
Entrez Gene ID
1326
Gene Name
mitogen-activated protein kinase kinase kinase 8
Gene Aliases
AURA2, COT, EST, ESTF, MEKK8, TPL2, Tpl-2, c-COT
UniGene
Hs.432453
Gene Chromosome Location
Chr.10: 30434021-30461833 on Build GRCh38
Transcripts
NM_001244134.1 NM_001320961.1 NM_005204.3 XM_017015707.1 XM_017015708.1 XM_017015709.1 XM_017015710.1 XM_017015711.1 XM_017015712.1 XM_017015713.1 XM_017015714.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7194133 c.31A>G p.K11E Chr.10: 30438969-30438969 on Build GRCh38
7194134 c.31A>G p.K11E Chr.10: 30438969-30438969 on Build GRCh38
5551934 c.74A>G p.D25G Chr.10: 30439012-30439012 on Build GRCh38
427467 c.275C>G p.S92C Chr.10: 30439213-30439213 on Build GRCh38
7227207 c.311A>G p.Q104R Chr.10: 30439249-30439249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761222379 Chr.10: 30438851-30438851 on Build GRCh38
rs964024255 Chr.10: 30438887-30438887 on Build GRCh38
rs139640494 Chr.10: 30439032-30439032 on Build GRCh38
rs755873862 Chr.10: 30439055-30439055 on Build GRCh38
rs1042058 Chr.10: 30439172-30439172 on Build GRCh38
rs777682936 Chr.10: 30438977-30438977 on Build GRCh38
rs757105191 Chr.10: 30439066-30439066 on Build GRCh38
rs753839798 Chr.10: 30439063-30439063 on Build GRCh38
rs144299935 Chr.10: 30439293-30439293 on Build GRCh38
rs780995987 Chr.10: 30438958-30438958 on Build GRCh38
rs753513338 Chr.10: 30439152-30439152 on Build GRCh38
rs914933119 Chr.10: 30439170-30439170 on Build GRCh38
rs745882056 Chr.10: 30439079-30439079 on Build GRCh38
rs762517753 Chr.10: 30438912-30438912 on Build GRCh38
rs377410996 Chr.10: 30439128-30439128 on Build GRCh38
rs760273502 Chr.10: 30439146-30439146 on Build GRCh38
rs778965236 Chr.10: 30439002-30439002 on Build GRCh38
rs8176968 Chr.10: 30438859-30438859 on Build GRCh38
rs577031274 Chr.10: 30439067-30439067 on Build GRCh38
rs550846611 Chr.10: 30438885-30438885 on Build GRCh38
rs748402123 Chr.10: 30439239-30439239 on Build GRCh38
rs1016753845 Chr.10: 30438952-30438952 on Build GRCh38
rs938853569 Chr.10: 30439242-30439242 on Build GRCh38
rs747361922 Chr.10: 30439014-30439014 on Build GRCh38
rs1003139110 Chr.10: 30438876-30438876 on Build GRCh38
rs767144818 Chr.10: 30439041-30439041 on Build GRCh38
rs150001848 Chr.10: 30439107-30439107 on Build GRCh38
rs777243002 Chr.10: 30439023-30439023 on Build GRCh38
rs762418488 Chr.10: 30439030-30439030 on Build GRCh38
rs886421503 Chr.10: 30438902-30438902 on Build GRCh38
rs757273381 Chr.10: 30438953-30438953 on Build GRCh38
rs372120772 Chr.10: 30438980-30438980 on Build GRCh38
rs886724478 Chr.10: 30439278-30439278 on Build GRCh38
rs754151678 Chr.10: 30438899-30438899 on Build GRCh38
rs767198025 Chr.10: 30439138-30439138 on Build GRCh38
rs747488469 Chr.10: 30438927-30438928 on Build GRCh38
rs745432732 Chr.10: 30439289-30439289 on Build GRCh38
rs752555792 Chr.10: 30439043-30439043 on Build GRCh38
rs916277317 Chr.10: 30439126-30439126 on Build GRCh38
rs765934792 Chr.10: 30439031-30439031 on Build GRCh38
rs755208662 Chr.10: 30439216-30439216 on Build GRCh38
rs747304103 Chr.10: 30439243-30439243 on Build GRCh38
rs146728212 Chr.10: 30439114-30439114 on Build GRCh38
rs55962705 Chr.10: 30439160-30439160 on Build GRCh38
rs763911668 Chr.10: 30439151-30439151 on Build GRCh38
rs748719604 Chr.10: 30439127-30439127 on Build GRCh38
rs781623061 Chr.10: 30439226-30439226 on Build GRCh38
rs780656361 Chr.10: 30439236-30439236 on Build GRCh38
rs145696701 Chr.10: 30439181-30439181 on Build GRCh38
rs771091007 Chr.10: 30438934-30438934 on Build GRCh38
rs549474 Chr.10: 30439168-30439168 on Build GRCh38
rs773924433 Chr.10: 30439131-30439131 on Build GRCh38
rs779058455 Chr.10: 30439076-30439076 on Build GRCh38
rs539535216 Chr.10: 30439056-30439056 on Build GRCh38
rs780414317 Chr.10: 30439106-30439106 on Build GRCh38
rs374430565 Chr.10: 30439035-30439035 on Build GRCh38
rs751200275 Chr.10: 30438918-30438918 on Build GRCh38
rs772123870 Chr.10: 30439007-30439007 on Build GRCh38
rs575445977 Chr.10: 30439294-30439294 on Build GRCh38
rs758966181 Chr.10: 30439134-30439134 on Build GRCh38
rs947835153 Chr.10: 30439171-30439171 on Build GRCh38
rs775103410 Chr.10: 30439140-30439140 on Build GRCh38
rs780469254 Chr.10: 30439187-30439187 on Build GRCh38
rs752421142 Chr.10: 30438961-30438961 on Build GRCh38
rs370656437 Chr.10: 30439166-30439166 on Build GRCh38
rs769129181 Chr.10: 30439288-30439288 on Build GRCh38
rs781423722 Chr.10: 30438986-30438989 on Build GRCh38
rs150579938 Chr.10: 30439020-30439020 on Build GRCh38
rs756102760 Chr.10: 30438974-30438974 on Build GRCh38
rs766832145 Chr.10: 30438897-30438897 on Build GRCh38
rs1036237977 Chr.10: 30439098-30439098 on Build GRCh38
rs77186746 Chr.10: 30439097-30439097 on Build GRCh38
rs765985240 Chr.10: 30438913-30438913 on Build GRCh38
rs776014707 Chr.10: 30439009-30439009 on Build GRCh38
rs751851768 Chr.10: 30439208-30439208 on Build GRCh38
rs117396201 Chr.10: 30439113-30439113 on Build GRCh38
rs1008537173 Chr.10: 30438884-30438884 on Build GRCh38
rs552211501 Chr.10: 30438857-30438857 on Build GRCh38
rs1004915792 Chr.10: 30438930-30438930 on Build GRCh38
rs746136036 Chr.10: 30439004-30439004 on Build GRCh38

View Full Product Details