Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95237892-95238392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDLIM1
Entrez Gene ID
9124
Gene Name
PDZ and LIM domain 1
Gene Aliases
CLIM1, CLP-36, CLP36, HEL-S-112, hCLIM1
UniGene
Hs.368525
Gene Chromosome Location
Chr.10: 95237568-95291148 on Build GRCh38
Transcripts
NM_020992.3 XM_011540330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5738998 c.835C>T p.R279C Chr.10: 95238080-95238080 on Build GRCh38
921773 c.941G>A p.R314Q Chr.10: 95237974-95237974 on Build GRCh38
5837536 c.869G>A p.G290D Chr.10: 95238046-95238046 on Build GRCh38
3752067 c.852T>C p.Y284Y Chr.10: 95238063-95238063 on Build GRCh38
1287058 c.824G>T p.R275L Chr.10: 95238091-95238091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370809135 Chr.10: 95238079-95238079 on Build GRCh38
rs533972034 Chr.10: 95238073-95238073 on Build GRCh38
rs770821975 Chr.10: 95238097-95238097 on Build GRCh38
rs991342288 Chr.10: 95238337-95238337 on Build GRCh38
rs987816667 Chr.10: 95238280-95238280 on Build GRCh38
rs759680023 Chr.10: 95237970-95237971 on Build GRCh38
rs139651549 Chr.10: 95238355-95238355 on Build GRCh38
rs755653017 Chr.10: 95238039-95238039 on Build GRCh38
rs748931617 Chr.10: 95238112-95238113 on Build GRCh38
rs538320746 Chr.10: 95238266-95238266 on Build GRCh38
rs758650219 Chr.10: 95238069-95238069 on Build GRCh38
rs748177765 Chr.10: 95238086-95238086 on Build GRCh38
rs772608190 Chr.10: 95238113-95238114 on Build GRCh38
rs558211723 Chr.10: 95238329-95238329 on Build GRCh38
rs752196380 Chr.10: 95238022-95238022 on Build GRCh38
rs575749051 Chr.10: 95238252-95238252 on Build GRCh38
rs1049814 Chr.10: 95238063-95238063 on Build GRCh38
rs969662294 Chr.10: 95238206-95238206 on Build GRCh38
rs958828494 Chr.10: 95238336-95238336 on Build GRCh38
rs752361625 Chr.10: 95238114-95238114 on Build GRCh38
rs752901362 Chr.10: 95238067-95238070 on Build GRCh38
rs1011505051 Chr.10: 95238219-95238219 on Build GRCh38
rs766224079 Chr.10: 95237968-95237968 on Build GRCh38
rs1035521059 Chr.10: 95238293-95238293 on Build GRCh38
rs967419655 Chr.10: 95238274-95238274 on Build GRCh38
rs777513042 Chr.10: 95238109-95238110 on Build GRCh38
rs781513523 Chr.10: 95238153-95238153 on Build GRCh38
rs775525917 Chr.10: 95237940-95237940 on Build GRCh38
rs376960366 Chr.10: 95237943-95237943 on Build GRCh38
rs1051244241 Chr.10: 95238110-95238110 on Build GRCh38
rs768989089 Chr.10: 95238080-95238080 on Build GRCh38
rs1023049259 Chr.10: 95238199-95238199 on Build GRCh38
rs113032402 Chr.10: 95238112-95238112 on Build GRCh38
rs1002942107 Chr.10: 95238278-95238278 on Build GRCh38
rs781768621 Chr.10: 95238267-95238267 on Build GRCh38
rs867034939 Chr.10: 95237979-95237979 on Build GRCh38
rs772978940 Chr.10: 95238092-95238092 on Build GRCh38
rs372510009 Chr.10: 95237962-95237962 on Build GRCh38
rs759668682 Chr.10: 95238158-95238172 on Build GRCh38
rs762604256 Chr.10: 95237967-95237967 on Build GRCh38
rs762045152 Chr.10: 95237956-95237956 on Build GRCh38
rs772502005 Chr.10: 95237933-95237933 on Build GRCh38
rs755402441 Chr.10: 95238148-95238148 on Build GRCh38
rs902059987 Chr.10: 95237965-95237965 on Build GRCh38
rs1020262181 Chr.10: 95238264-95238264 on Build GRCh38
rs761096185 Chr.10: 95238114-95238115 on Build GRCh38
rs745318815 Chr.10: 95238111-95238112 on Build GRCh38
rs776996985 Chr.10: 95238116-95238117 on Build GRCh38
rs754601619 Chr.10: 95237975-95237975 on Build GRCh38
rs753329631 Chr.10: 95238126-95238126 on Build GRCh38
rs145614632 Chr.10: 95237959-95237959 on Build GRCh38
rs1049921 Chr.10: 95237913-95237913 on Build GRCh38
rs781630484 Chr.10: 95238085-95238085 on Build GRCh38
rs779232015 Chr.10: 95238043-95238043 on Build GRCh38
rs779538958 Chr.10: 95237926-95237926 on Build GRCh38
rs762912831 Chr.10: 95238095-95238095 on Build GRCh38
rs765276765 Chr.10: 95238161-95238161 on Build GRCh38
rs758919967 Chr.10: 95238137-95238137 on Build GRCh38
rs397846225 Chr.10: 95238358-95238358 on Build GRCh38
rs991849569 Chr.10: 95237992-95237992 on Build GRCh38
rs757689568 Chr.10: 95237918-95237918 on Build GRCh38
rs377676948 Chr.10: 95238117-95238117 on Build GRCh38
rs547415127 Chr.10: 95238099-95238099 on Build GRCh38
rs774118628 Chr.10: 95238102-95238102 on Build GRCh38
rs562977612 Chr.10: 95237952-95237952 on Build GRCh38
rs765506392 Chr.10: 95237981-95237981 on Build GRCh38
rs757899611 Chr.10: 95238194-95238194 on Build GRCh38
rs759390698 Chr.10: 95238113-95238113 on Build GRCh38
rs757170222 Chr.10: 95238110-95238111 on Build GRCh38
rs758236230 Chr.10: 95238073-95238074 on Build GRCh38
rs752319886 Chr.10: 95237974-95237974 on Build GRCh38
rs202155307 Chr.10: 95238115-95238115 on Build GRCh38
rs199932530 Chr.10: 95238040-95238040 on Build GRCh38
rs769937368 Chr.10: 95238091-95238091 on Build GRCh38
rs746153920 Chr.10: 95237928-95237928 on Build GRCh38
rs369246498 Chr.10: 95238144-95238144 on Build GRCh38
rs780767932 Chr.10: 95238141-95238141 on Build GRCh38
rs35704711 Chr.10: 95238355-95238355 on Build GRCh38
rs147722437 Chr.10: 95237997-95237997 on Build GRCh38

View Full Product Details