Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95238377-95238795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDLIM1
Entrez Gene ID
9124
Gene Name
PDZ and LIM domain 1
Gene Aliases
CLIM1, CLP-36, CLP36, HEL-S-112, hCLIM1
UniGene
Hs.368525
Gene Chromosome Location
Chr.10: 95237568-95291148 on Build GRCh38
Transcripts
NM_020992.3 XM_011540330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5750002 c.742G>T p.A248S Chr.10: 95238629-95238629 on Build GRCh38
4617056 c.760G>A p.A254T Chr.10: 95238611-95238611 on Build GRCh38
5776994 c.704C>A p.S235* Chr.10: 95238667-95238667 on Build GRCh38
1971001 c.749C>T p.S250L Chr.10: 95238622-95238622 on Build GRCh38
4455340 c.737A>T p.K246I Chr.10: 95238634-95238634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051544262 Chr.10: 95238515-95238515 on Build GRCh38
rs776764839 Chr.10: 95238692-95238692 on Build GRCh38
rs954398297 Chr.10: 95238722-95238722 on Build GRCh38
rs2860998 Chr.10: 95238762-95238762 on Build GRCh38
rs918948791 Chr.10: 95238455-95238455 on Build GRCh38
rs540814669 Chr.10: 95238434-95238434 on Build GRCh38
rs372785139 Chr.10: 95238709-95238709 on Build GRCh38
rs565368926 Chr.10: 95238625-95238625 on Build GRCh38
rs772884124 Chr.10: 95238728-95238728 on Build GRCh38
rs1039888169 Chr.10: 95238533-95238533 on Build GRCh38
rs1026506635 Chr.10: 95238500-95238500 on Build GRCh38
rs371469888 Chr.10: 95238608-95238608 on Build GRCh38
rs768192692 Chr.10: 95238732-95238732 on Build GRCh38
rs933041564 Chr.10: 95238502-95238502 on Build GRCh38
rs768352607 Chr.10: 95238700-95238700 on Build GRCh38
rs761352394 Chr.10: 95238704-95238704 on Build GRCh38
rs760345620 Chr.10: 95238602-95238602 on Build GRCh38
rs145894144 Chr.10: 95238626-95238626 on Build GRCh38
rs375596774 Chr.10: 95238573-95238573 on Build GRCh38
rs578136086 Chr.10: 95238426-95238426 on Build GRCh38
rs914777607 Chr.10: 95238765-95238765 on Build GRCh38
rs143667794 Chr.10: 95238528-95238528 on Build GRCh38
rs759267863 Chr.10: 95238582-95238582 on Build GRCh38
rs887821201 Chr.10: 95238504-95238504 on Build GRCh38
rs547524648 Chr.10: 95238695-95238695 on Build GRCh38
rs778775058 Chr.10: 95238629-95238629 on Build GRCh38
rs190654687 Chr.10: 95238693-95238693 on Build GRCh38
rs745836853 Chr.10: 95238635-95238635 on Build GRCh38
rs753099217 Chr.10: 95238733-95238733 on Build GRCh38
rs371730712 Chr.10: 95238621-95238621 on Build GRCh38
rs746172112 Chr.10: 95238418-95238418 on Build GRCh38
rs777883967 Chr.10: 95238549-95238549 on Build GRCh38
rs200857998 Chr.10: 95238586-95238586 on Build GRCh38
rs2901832 Chr.10: 95238701-95238701 on Build GRCh38
rs993740662 Chr.10: 95238456-95238456 on Build GRCh38
rs770850336 Chr.10: 95238566-95238566 on Build GRCh38
rs1034830136 Chr.10: 95238688-95238688 on Build GRCh38
rs368533526 Chr.10: 95238594-95238594 on Build GRCh38
rs562841980 Chr.10: 95238536-95238536 on Build GRCh38
rs374874749 Chr.10: 95238622-95238622 on Build GRCh38
rs753377309 Chr.10: 95238603-95238603 on Build GRCh38
rs12771349 Chr.10: 95238436-95238436 on Build GRCh38
rs574232989 Chr.10: 95238509-95238509 on Build GRCh38
rs370876200 Chr.10: 95238565-95238565 on Build GRCh38
rs766930662 Chr.10: 95238617-95238617 on Build GRCh38
rs947631562 Chr.10: 95238770-95238770 on Build GRCh38
rs375724368 Chr.10: 95238516-95238516 on Build GRCh38
rs772039791 Chr.10: 95238669-95238669 on Build GRCh38
rs769944923 Chr.10: 95238545-95238545 on Build GRCh38
rs201069556 Chr.10: 95238624-95238624 on Build GRCh38
rs372416055 Chr.10: 95238425-95238425 on Build GRCh38
rs200587356 Chr.10: 95238730-95238730 on Build GRCh38
rs963337945 Chr.10: 95238428-95238428 on Build GRCh38

View Full Product Details